Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154703722-154704223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNN3
Entrez Gene ID
3782
Gene Name
potassium calcium-activated channel subfamily N member 3
Gene Aliases
KCa2.3, SK3, SKCA3, hSK3
UniGene
Hs.490765
Gene Chromosome Location
Chr.1: 154697462-154870278 on Build GRCh38
Transcripts
NM_001204087.1 NM_002249.5 NM_170782.2

SNP Detail

SNP ID Chromosome Location
rs985755439 Chr.1: 154703953-154703953 on Build GRCh38
rs978467174 Chr.1: 154704067-154704067 on Build GRCh38
rs1005585970 Chr.1: 154703854-154703854 on Build GRCh38
rs869135104 Chr.1: 154704055-154704056 on Build GRCh38
rs1029885919 Chr.1: 154703942-154703942 on Build GRCh38
rs571875977 Chr.1: 154704157-154704157 on Build GRCh38
rs889089168 Chr.1: 154703901-154703901 on Build GRCh38
rs928187187 Chr.1: 154704178-154704178 on Build GRCh38
rs1018146196 Chr.1: 154703929-154703929 on Build GRCh38
rs914881201 Chr.1: 154703781-154703781 on Build GRCh38
rs1017015714 Chr.1: 154703860-154703860 on Build GRCh38
rs773996607 Chr.1: 154704065-154704065 on Build GRCh38
rs911368111 Chr.1: 154704029-154704029 on Build GRCh38
rs1030058505 Chr.1: 154704183-154704183 on Build GRCh38
rs4418588 Chr.1: 154703887-154703887 on Build GRCh38
rs760410737 Chr.1: 154704172-154704172 on Build GRCh38
rs955833383 Chr.1: 154703946-154703946 on Build GRCh38
rs934286078 Chr.1: 154704076-154704076 on Build GRCh38
rs573396210 Chr.1: 154704164-154704164 on Build GRCh38
rs184187248 Chr.1: 154703844-154703844 on Build GRCh38
rs530968072 Chr.1: 154703809-154703809 on Build GRCh38
rs191995548 Chr.1: 154703791-154703791 on Build GRCh38
rs1044353154 Chr.1: 154704145-154704145 on Build GRCh38
rs536347336 Chr.1: 154704068-154704068 on Build GRCh38
rs1395565 Chr.1: 154703932-154703932 on Build GRCh38
rs895504685 Chr.1: 154704136-154704136 on Build GRCh38
rs955430749 Chr.1: 154704185-154704185 on Build GRCh38
rs946905570 Chr.1: 154704045-154704045 on Build GRCh38
rs572599429 Chr.1: 154703917-154703917 on Build GRCh38
rs949994051 Chr.1: 154704137-154704137 on Build GRCh38
rs1053214289 Chr.1: 154704095-154704095 on Build GRCh38
rs906821451 Chr.1: 154703795-154703795 on Build GRCh38
rs553490376 Chr.1: 154704153-154704153 on Build GRCh38
rs986110008 Chr.1: 154704192-154704192 on Build GRCh38
rs763349610 Chr.1: 154703927-154703927 on Build GRCh38
rs956767312 Chr.1: 154704079-154704079 on Build GRCh38
rs189738131 Chr.1: 154704155-154704155 on Build GRCh38
rs897885269 Chr.1: 154703835-154703835 on Build GRCh38
rs750033575 Chr.1: 154704171-154704171 on Build GRCh38
rs780553532 Chr.1: 154703925-154703925 on Build GRCh38
rs934316989 Chr.1: 154703758-154703758 on Build GRCh38
rs989209137 Chr.1: 154704146-154704146 on Build GRCh38
rs895735243 Chr.1: 154703792-154703792 on Build GRCh38
rs775942458 Chr.1: 154703881-154703881 on Build GRCh38
rs528689724 Chr.1: 154704152-154704152 on Build GRCh38
rs138404969 Chr.1: 154703805-154703805 on Build GRCh38

View Full Product Details