Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154704253-154704749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNN3
Entrez Gene ID
3782
Gene Name
potassium calcium-activated channel subfamily N member 3
Gene Aliases
KCa2.3, SK3, SKCA3, hSK3
UniGene
Hs.490765
Gene Chromosome Location
Chr.1: 154697462-154870278 on Build GRCh38
Transcripts
NM_001204087.1 NM_002249.5 NM_170782.2

SNP Detail

SNP ID Chromosome Location
rs960379433 Chr.1: 154704439-154704439 on Build GRCh38
rs989756628 Chr.1: 154704355-154704356 on Build GRCh38
rs190526678 Chr.1: 154704441-154704441 on Build GRCh38
rs543408052 Chr.1: 154704523-154704523 on Build GRCh38
rs910617159 Chr.1: 154704672-154704672 on Build GRCh38
rs941171875 Chr.1: 154704350-154704350 on Build GRCh38
rs971438469 Chr.1: 154704612-154704612 on Build GRCh38
rs994647294 Chr.1: 154704368-154704368 on Build GRCh38
rs577697040 Chr.1: 154704506-154704506 on Build GRCh38
rs543086600 Chr.1: 154704625-154704625 on Build GRCh38
rs563294837 Chr.1: 154704540-154704540 on Build GRCh38
rs998027773 Chr.1: 154704318-154704318 on Build GRCh38
rs979807849 Chr.1: 154704652-154704652 on Build GRCh38
rs916891666 Chr.1: 154704295-154704295 on Build GRCh38
rs1001138400 Chr.1: 154704478-154704478 on Build GRCh38
rs573557217 Chr.1: 154704355-154704355 on Build GRCh38
rs573623902 Chr.1: 154704685-154704685 on Build GRCh38
rs927586276 Chr.1: 154704435-154704435 on Build GRCh38
rs575545181 Chr.1: 154704547-154704547 on Build GRCh38
rs988963137 Chr.1: 154704552-154704552 on Build GRCh38
rs917010599 Chr.1: 154704610-154704610 on Build GRCh38
rs1044667820 Chr.1: 154704322-154704322 on Build GRCh38
rs186230111 Chr.1: 154704293-154704293 on Build GRCh38
rs552720774 Chr.1: 154704375-154704375 on Build GRCh38
rs927193076 Chr.1: 154704698-154704698 on Build GRCh38
rs113740378 Chr.1: 154704699-154704699 on Build GRCh38
rs561279084 Chr.1: 154704678-154704678 on Build GRCh38
rs545711746 Chr.1: 154704369-154704369 on Build GRCh38
rs949784100 Chr.1: 154704307-154704307 on Build GRCh38
rs1040447058 Chr.1: 154704682-154704682 on Build GRCh38
rs972302007 Chr.1: 154704517-154704517 on Build GRCh38
rs968307251 Chr.1: 154704470-154704470 on Build GRCh38
rs1051223852 Chr.1: 154704395-154704395 on Build GRCh38
rs537078584 Chr.1: 154704305-154704305 on Build GRCh38
rs927222752 Chr.1: 154704340-154704340 on Build GRCh38
rs372381828 Chr.1: 154704333-154704333 on Build GRCh38
rs943483674 Chr.1: 154704677-154704677 on Build GRCh38
rs956926051 Chr.1: 154704510-154704510 on Build GRCh38
rs186228734 Chr.1: 154704417-154704417 on Build GRCh38
rs941263667 Chr.1: 154704713-154704713 on Build GRCh38
rs189092599 Chr.1: 154704360-154704360 on Build GRCh38
rs1018508551 Chr.1: 154704459-154704459 on Build GRCh38
rs930818988 Chr.1: 154704543-154704543 on Build GRCh38
rs182039496 Chr.1: 154704416-154704416 on Build GRCh38

View Full Product Details