Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155913701-155914246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0907
Entrez Gene ID
22889
Gene Name
KIAA0907
Gene Aliases
BLOM7
UniGene
Hs.24656
Gene Chromosome Location
Chr.1: 155913043-155934442 on Build GRCh38
Transcripts
NM_014949.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1196557 c.1767G>C p.Q589H Chr.1: 155914199-155914199 on Build GRCh38
6323447 c.1785C>T p.Y595Y Chr.1: 155914181-155914181 on Build GRCh38
3475081 c.1841C>T p.P614L Chr.1: 155914125-155914125 on Build GRCh38
346544 c.1792C>A p.P598T Chr.1: 155914174-155914174 on Build GRCh38
3740700 c.1809A>C p.R603R Chr.1: 155914157-155914157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188703027 Chr.1: 155914043-155914043 on Build GRCh38
rs938235971 Chr.1: 155913890-155913890 on Build GRCh38
rs970411757 Chr.1: 155913834-155913834 on Build GRCh38
rs763371915 Chr.1: 155913797-155913797 on Build GRCh38
rs1053643069 Chr.1: 155914118-155914118 on Build GRCh38
rs748469942 Chr.1: 155914146-155914148 on Build GRCh38
rs767098666 Chr.1: 155913947-155913947 on Build GRCh38
rs768580229 Chr.1: 155914119-155914119 on Build GRCh38
rs553515581 Chr.1: 155913974-155913974 on Build GRCh38
rs573749474 Chr.1: 155913858-155913858 on Build GRCh38
rs938312255 Chr.1: 155913828-155913828 on Build GRCh38
rs774035097 Chr.1: 155913935-155913935 on Build GRCh38
rs1032150220 Chr.1: 155913742-155913742 on Build GRCh38
rs781098786 Chr.1: 155914158-155914158 on Build GRCh38
rs927209870 Chr.1: 155913781-155913781 on Build GRCh38
rs143700648 Chr.1: 155913748-155913748 on Build GRCh38
rs775765915 Chr.1: 155913752-155913752 on Build GRCh38
rs3834092 Chr.1: 155913962-155913964 on Build GRCh38
rs546104273 Chr.1: 155914053-155914053 on Build GRCh38
rs199705309 Chr.1: 155914178-155914178 on Build GRCh38
rs980035122 Chr.1: 155913761-155913761 on Build GRCh38
rs541631749 Chr.1: 155913941-155913941 on Build GRCh38
rs746605738 Chr.1: 155914115-155914115 on Build GRCh38
rs997155887 Chr.1: 155914091-155914091 on Build GRCh38
rs775700856 Chr.1: 155914203-155914203 on Build GRCh38
rs564207619 Chr.1: 155914065-155914065 on Build GRCh38
rs777182758 Chr.1: 155914046-155914046 on Build GRCh38
rs754347254 Chr.1: 155914093-155914093 on Build GRCh38
rs769820823 Chr.1: 155914163-155914163 on Build GRCh38
rs781765779 Chr.1: 155914080-155914082 on Build GRCh38
rs759730294 Chr.1: 155914180-155914180 on Build GRCh38
rs772030059 Chr.1: 155914182-155914182 on Build GRCh38
rs756470265 Chr.1: 155914160-155914160 on Build GRCh38
rs750889119 Chr.1: 155914085-155914085 on Build GRCh38
rs757742850 Chr.1: 155914094-155914094 on Build GRCh38
rs1009352433 Chr.1: 155914188-155914188 on Build GRCh38
rs779912969 Chr.1: 155914114-155914114 on Build GRCh38
rs760819238 Chr.1: 155914220-155914220 on Build GRCh38
rs555487621 Chr.1: 155913851-155913851 on Build GRCh38
rs929715384 Chr.1: 155913874-155913874 on Build GRCh38
rs537041969 Chr.1: 155913770-155913770 on Build GRCh38
rs1047797839 Chr.1: 155913997-155913997 on Build GRCh38
rs749995222 Chr.1: 155914096-155914096 on Build GRCh38
rs890508580 Chr.1: 155914017-155914017 on Build GRCh38
rs887951042 Chr.1: 155913906-155913906 on Build GRCh38
rs757812201 Chr.1: 155914106-155914106 on Build GRCh38
rs764425097 Chr.1: 155913822-155913822 on Build GRCh38
rs781560246 Chr.1: 155913869-155913869 on Build GRCh38
rs973554028 Chr.1: 155913934-155913934 on Build GRCh38

View Full Product Details