Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 137011169-137011674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL20RA
Entrez Gene ID
53832
Gene Name
interleukin 20 receptor subunit alpha
Gene Aliases
CRF2-8, IL-20R-alpha, IL-20R1, IL-20RA
UniGene
Hs.445868
Gene Chromosome Location
Chr.6: 136999971-137045366 on Build GRCh38
Transcripts
NM_001278722.1 NM_001278723.1 NM_001278724.1 NM_014432.3 XM_006715506.2 XM_011535904.2 XM_017010954.1 XM_017010955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6685205 c.313G>A p.E105K Chr.6: 137011364-137011364 on Build GRCh38
5782179 c.302C>G p.T101S Chr.6: 137011375-137011375 on Build GRCh38
4761795 c.278C>G p.T93S Chr.6: 137011399-137011399 on Build GRCh38
5869391 c.347G>A p.W116* Chr.6: 137011330-137011330 on Build GRCh38
3015582 c.321G>A p.Q107Q Chr.6: 137011356-137011356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752780342 Chr.6: 137011437-137011437 on Build GRCh38
rs932987552 Chr.6: 137011622-137011622 on Build GRCh38
rs940830502 Chr.6: 137011496-137011496 on Build GRCh38
rs759862967 Chr.6: 137011425-137011425 on Build GRCh38
rs1026442841 Chr.6: 137011250-137011250 on Build GRCh38
rs951137061 Chr.6: 137011251-137011251 on Build GRCh38
rs559032166 Chr.6: 137011591-137011591 on Build GRCh38
rs745596581 Chr.6: 137011349-137011349 on Build GRCh38
rs775111805 Chr.6: 137011371-137011371 on Build GRCh38
rs767473706 Chr.6: 137011431-137011431 on Build GRCh38
rs757149703 Chr.6: 137011512-137011512 on Build GRCh38
rs771466183 Chr.6: 137011365-137011365 on Build GRCh38
rs773805262 Chr.6: 137011343-137011343 on Build GRCh38
rs756206874 Chr.6: 137011448-137011448 on Build GRCh38
rs752656609 Chr.6: 137011470-137011470 on Build GRCh38
rs765178251 Chr.6: 137011458-137011461 on Build GRCh38
rs771954 Chr.6: 137011608-137011608 on Build GRCh38
rs907409720 Chr.6: 137011524-137011524 on Build GRCh38
rs373657721 Chr.6: 137011489-137011489 on Build GRCh38
rs142983578 Chr.6: 137011358-137011358 on Build GRCh38
rs777629614 Chr.6: 137011466-137011466 on Build GRCh38
rs757038168 Chr.6: 137011500-137011500 on Build GRCh38
rs777717458 Chr.6: 137011323-137011323 on Build GRCh38
rs574128582 Chr.6: 137011484-137011484 on Build GRCh38
rs751824156 Chr.6: 137011295-137011295 on Build GRCh38
rs773882416 Chr.6: 137011402-137011402 on Build GRCh38
rs776995192 Chr.6: 137011252-137011252 on Build GRCh38
rs763356187 Chr.6: 137011406-137011406 on Build GRCh38
rs1043454420 Chr.6: 137011600-137011600 on Build GRCh38
rs770725433 Chr.6: 137011336-137011336 on Build GRCh38
rs142251595 Chr.6: 137011414-137011414 on Build GRCh38
rs767055831 Chr.6: 137011247-137011247 on Build GRCh38
rs758867778 Chr.6: 137011270-137011270 on Build GRCh38
rs199954231 Chr.6: 137011301-137011301 on Build GRCh38
rs566142607 Chr.6: 137011203-137011203 on Build GRCh38
rs758912551 Chr.6: 137011243-137011243 on Build GRCh38
rs769575065 Chr.6: 137011226-137011226 on Build GRCh38
rs759397154 Chr.6: 137011418-137011418 on Build GRCh38
rs138262239 Chr.6: 137011317-137011317 on Build GRCh38
rs749033134 Chr.6: 137011324-137011324 on Build GRCh38
rs189003450 Chr.6: 137011263-137011263 on Build GRCh38
rs866902353 Chr.6: 137011327-137011327 on Build GRCh38
rs748023995 Chr.6: 137011307-137011307 on Build GRCh38
rs770387625 Chr.6: 137011378-137011378 on Build GRCh38
rs899384920 Chr.6: 137011546-137011546 on Build GRCh38
rs371538746 Chr.6: 137011258-137011258 on Build GRCh38
rs184019040 Chr.6: 137011228-137011228 on Build GRCh38
rs758215934 Chr.6: 137011609-137011609 on Build GRCh38
rs907997562 Chr.6: 137011494-137011494 on Build GRCh38
rs1018784597 Chr.6: 137011625-137011625 on Build GRCh38
rs368376233 Chr.6: 137011239-137011239 on Build GRCh38
rs770529273 Chr.6: 137011238-137011238 on Build GRCh38
rs758393772 Chr.6: 137011504-137011505 on Build GRCh38
rs138995272 Chr.6: 137011409-137011409 on Build GRCh38
rs937627237 Chr.6: 137011544-137011544 on Build GRCh38
rs535023311 Chr.6: 137011207-137011207 on Build GRCh38
rs754948145 Chr.6: 137011297-137011297 on Build GRCh38
rs184335342 Chr.6: 137011616-137011616 on Build GRCh38
rs975112431 Chr.6: 137011359-137011359 on Build GRCh38
rs748120995 Chr.6: 137011225-137011225 on Build GRCh38
rs1055170687 Chr.6: 137011565-137011565 on Build GRCh38
rs543176650 Chr.6: 137011515-137011515 on Build GRCh38
rs1010553075 Chr.6: 137011579-137011579 on Build GRCh38
rs374954861 Chr.6: 137011294-137011294 on Build GRCh38
rs894799821 Chr.6: 137011571-137011571 on Build GRCh38
rs146129043 Chr.6: 137011372-137011372 on Build GRCh38
rs572557971 Chr.6: 137011605-137011605 on Build GRCh38
rs966324760 Chr.6: 137011639-137011639 on Build GRCh38

View Full Product Details