Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 137206831-137207337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNGR1
Entrez Gene ID
3459
Gene Name
interferon gamma receptor 1
Gene Aliases
CD119, IFNGR, IMD27A, IMD27B
UniGene
Hs.520414
Gene Chromosome Location
Chr.6: 137197484-137220351 on Build GRCh38
Transcripts
NM_000416.2 XM_006715470.2 XM_006715471.2 XM_011535793.2 XM_011535794.1 XM_017010827.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3662162 c.92C>A p.T31K Chr.6: 137207071-137207071 on Build GRCh38
6172814 c.61A>T p.T21S Chr.6: 137207072-137207072 on Build GRCh38
1210293 c.112G>T p.E38* Chr.6: 137207051-137207051 on Build GRCh38
6683920 c.165G>T p.Q55H Chr.6: 137206998-137206998 on Build GRCh38
6536082 c.192G>C p.K64N Chr.6: 137206971-137206971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950242339 Chr.6: 137206871-137206871 on Build GRCh38
rs866902738 Chr.6: 137207032-137207032 on Build GRCh38
rs777965499 Chr.6: 137207030-137207030 on Build GRCh38
rs181130211 Chr.6: 137207227-137207227 on Build GRCh38
rs781724626 Chr.6: 137207024-137207024 on Build GRCh38
rs574914821 Chr.6: 137207144-137207144 on Build GRCh38
rs1004961994 Chr.6: 137207155-137207155 on Build GRCh38
rs914935462 Chr.6: 137207301-137207301 on Build GRCh38
rs767897049 Chr.6: 137207106-137207106 on Build GRCh38
rs765012731 Chr.6: 137207093-137207093 on Build GRCh38
rs779191331 Chr.6: 137207010-137207019 on Build GRCh38
rs763004493 Chr.6: 137206956-137206956 on Build GRCh38
rs966886128 Chr.6: 137207163-137207163 on Build GRCh38
rs981854221 Chr.6: 137207259-137207259 on Build GRCh38
rs796276822 Chr.6: 137206897-137206897 on Build GRCh38
rs587776854 Chr.6: 137207059-137207060 on Build GRCh38
rs980254566 Chr.6: 137207230-137207230 on Build GRCh38
rs757939072 Chr.6: 137207099-137207099 on Build GRCh38
rs561031447 Chr.6: 137206999-137206999 on Build GRCh38
rs957616224 Chr.6: 137207276-137207276 on Build GRCh38
rs17175322 Chr.6: 137206982-137206982 on Build GRCh38
rs770973014 Chr.6: 137207046-137207046 on Build GRCh38
rs41505745 Chr.6: 137206945-137206945 on Build GRCh38
rs774237252 Chr.6: 137206959-137206959 on Build GRCh38
rs763644961 Chr.6: 137206996-137206996 on Build GRCh38
rs41477052 Chr.6: 137207122-137207122 on Build GRCh38
rs917438524 Chr.6: 137206870-137206870 on Build GRCh38
rs937676852 Chr.6: 137206950-137206950 on Build GRCh38
rs771064624 Chr.6: 137206957-137206957 on Build GRCh38
rs778617419 Chr.6: 137207004-137207004 on Build GRCh38
rs372285230 Chr.6: 137207200-137207200 on Build GRCh38
rs121913174 Chr.6: 137207153-137207153 on Build GRCh38
rs759517006 Chr.6: 137206960-137206960 on Build GRCh38
rs1024384720 Chr.6: 137207188-137207188 on Build GRCh38
rs758999421 Chr.6: 137207121-137207121 on Build GRCh38
rs34046625 Chr.6: 137206937-137206937 on Build GRCh38
rs995097591 Chr.6: 137206979-137206979 on Build GRCh38
rs538147969 Chr.6: 137207027-137207027 on Build GRCh38
rs373456911 Chr.6: 137207082-137207082 on Build GRCh38
rs749578839 Chr.6: 137207296-137207298 on Build GRCh38
rs762424077 Chr.6: 137207052-137207052 on Build GRCh38
rs573548206 Chr.6: 137206929-137206929 on Build GRCh38
rs780704447 Chr.6: 137206918-137206918 on Build GRCh38
rs749976277 Chr.6: 137207007-137207007 on Build GRCh38
rs587776855 Chr.6: 137206962-137206962 on Build GRCh38
rs587776853 Chr.6: 137207032-137207032 on Build GRCh38
rs202070612 Chr.6: 137207115-137207115 on Build GRCh38
rs139375382 Chr.6: 137207129-137207129 on Build GRCh38
rs555180897 Chr.6: 137207063-137207063 on Build GRCh38
rs755352307 Chr.6: 137206915-137206915 on Build GRCh38
rs754564084 Chr.6: 137207161-137207161 on Build GRCh38
rs957619516 Chr.6: 137207123-137207123 on Build GRCh38
rs990388986 Chr.6: 137207290-137207290 on Build GRCh38
rs531198728 Chr.6: 137207131-137207131 on Build GRCh38
rs17175329 Chr.6: 137206948-137206948 on Build GRCh38
rs761509783 Chr.6: 137207089-137207089 on Build GRCh38
rs945137618 Chr.6: 137207053-137207053 on Build GRCh38
rs370074491 Chr.6: 137207071-137207071 on Build GRCh38
rs937667659 Chr.6: 137207278-137207278 on Build GRCh38
rs745988398 Chr.6: 137207067-137207067 on Build GRCh38
rs148104171 Chr.6: 137207028-137207028 on Build GRCh38
rs121912715 Chr.6: 137206981-137206981 on Build GRCh38
rs375024435 Chr.6: 137207011-137207011 on Build GRCh38
rs771850288 Chr.6: 137207070-137207070 on Build GRCh38
rs753311707 Chr.6: 137207111-137207111 on Build GRCh38
rs749548839 Chr.6: 137207033-137207033 on Build GRCh38
rs199742409 Chr.6: 137207084-137207084 on Build GRCh38
rs757426347 Chr.6: 137207126-137207126 on Build GRCh38
rs753213766 Chr.6: 137207070-137207078 on Build GRCh38
rs544637873 Chr.6: 137206998-137206998 on Build GRCh38
rs906180253 Chr.6: 137206879-137206879 on Build GRCh38
rs760535699 Chr.6: 137206983-137206983 on Build GRCh38
rs778156170 Chr.6: 137207117-137207117 on Build GRCh38
rs568137266 Chr.6: 137207165-137207165 on Build GRCh38
rs764023379 Chr.6: 137207081-137207081 on Build GRCh38
rs56367090 Chr.6: 137207262-137207262 on Build GRCh38
rs771738514 Chr.6: 137207248-137207249 on Build GRCh38
rs781363944 Chr.6: 137206939-137206939 on Build GRCh38
rs867775070 Chr.6: 137206966-137206966 on Build GRCh38
rs749886514 Chr.6: 137207097-137207097 on Build GRCh38

View Full Product Details