Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 139178072-139178576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECA
Entrez Gene ID
51696
Gene Name
hdc homolog, cell cycle regulator
Gene Aliases
HDC, HDCL, HHDC, dJ225E12.1
UniGene
Hs.197644
Gene Chromosome Location
Chr.6: 139135112-139180809 on Build GRCh38
Transcripts
NM_016217.2

SNP Detail

SNP ID Chromosome Location
rs574688172 Chr.6: 139178314-139178315 on Build GRCh38
rs905524951 Chr.6: 139178194-139178194 on Build GRCh38
rs911131352 Chr.6: 139178386-139178386 on Build GRCh38
rs869026718 Chr.6: 139178133-139178137 on Build GRCh38
rs1014306499 Chr.6: 139178478-139178478 on Build GRCh38
rs970335131 Chr.6: 139178311-139178311 on Build GRCh38
rs993906207 Chr.6: 139178501-139178501 on Build GRCh38
rs896962300 Chr.6: 139178405-139178405 on Build GRCh38
rs922243939 Chr.6: 139178136-139178136 on Build GRCh38
rs963791575 Chr.6: 139178532-139178532 on Build GRCh38
rs565216844 Chr.6: 139178339-139178339 on Build GRCh38
rs77566634 Chr.6: 139178304-139178304 on Build GRCh38
rs950042512 Chr.6: 139178377-139178377 on Build GRCh38
rs889702209 Chr.6: 139178165-139178165 on Build GRCh38
rs201062294 Chr.6: 139178516-139178516 on Build GRCh38
rs891724095 Chr.6: 139178156-139178156 on Build GRCh38
rs933928532 Chr.6: 139178148-139178148 on Build GRCh38
rs72980699 Chr.6: 139178410-139178410 on Build GRCh38
rs188161312 Chr.6: 139178341-139178341 on Build GRCh38
rs397785067 Chr.6: 139178516-139178516 on Build GRCh38
rs533573779 Chr.6: 139178328-139178328 on Build GRCh38
rs777198169 Chr.6: 139178423-139178423 on Build GRCh38
rs895898146 Chr.6: 139178464-139178464 on Build GRCh38
rs752890315 Chr.6: 139178249-139178249 on Build GRCh38
rs547966810 Chr.6: 139178500-139178500 on Build GRCh38
rs1038623112 Chr.6: 139178397-139178397 on Build GRCh38
rs960049741 Chr.6: 139178293-139178293 on Build GRCh38
rs868082433 Chr.6: 139178506-139178506 on Build GRCh38
rs774854464 Chr.6: 139178102-139178102 on Build GRCh38
rs763922079 Chr.6: 139178360-139178360 on Build GRCh38
rs181003522 Chr.6: 139178388-139178388 on Build GRCh38
rs1047869609 Chr.6: 139178427-139178427 on Build GRCh38
rs1026335554 Chr.6: 139178270-139178270 on Build GRCh38
rs550396554 Chr.6: 139178164-139178164 on Build GRCh38
rs776205151 Chr.6: 139178499-139178500 on Build GRCh38
rs535835661 Chr.6: 139178223-139178223 on Build GRCh38
rs545509869 Chr.6: 139178200-139178200 on Build GRCh38
rs1017710749 Chr.6: 139178495-139178495 on Build GRCh38
rs376099308 Chr.6: 139178495-139178495 on Build GRCh38
rs979343183 Chr.6: 139178326-139178326 on Build GRCh38
rs368670941 Chr.6: 139178359-139178359 on Build GRCh38
rs933687984 Chr.6: 139178357-139178357 on Build GRCh38
rs1026466491 Chr.6: 139178291-139178291 on Build GRCh38
rs35622637 Chr.6: 139178499-139178499 on Build GRCh38
rs922337353 Chr.6: 139178353-139178353 on Build GRCh38
rs898220125 Chr.6: 139178249-139178250 on Build GRCh38
rs899253420 Chr.6: 139178222-139178222 on Build GRCh38
rs185007344 Chr.6: 139178338-139178338 on Build GRCh38
rs995023558 Chr.6: 139178250-139178251 on Build GRCh38
rs974679100 Chr.6: 139178310-139178310 on Build GRCh38
rs577770543 Chr.6: 139178436-139178436 on Build GRCh38
rs547701919 Chr.6: 139178300-139178300 on Build GRCh38
rs988627455 Chr.6: 139178342-139178342 on Build GRCh38
rs1022928131 Chr.6: 139178485-139178485 on Build GRCh38
rs1014249218 Chr.6: 139178299-139178299 on Build GRCh38
rs767581484 Chr.6: 139178191-139178191 on Build GRCh38
rs1000424672 Chr.6: 139178547-139178547 on Build GRCh38
rs756983224 Chr.6: 139178417-139178417 on Build GRCh38
rs951065407 Chr.6: 139178298-139178299 on Build GRCh38
rs935842765 Chr.6: 139178122-139178122 on Build GRCh38
rs545288129 Chr.6: 139178471-139178471 on Build GRCh38

View Full Product Details