Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 142752545-142753091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIVEP2
Entrez Gene ID
3097
Gene Name
human immunodeficiency virus type I enhancer binding protein 2
Gene Aliases
HIV-EP2, MBP-2, MIBP1, MRD43, SHN2, ZAS2, ZNF40B
UniGene
Hs.510172
Gene Chromosome Location
Chr.6: 142751467-142945201 on Build GRCh38
Transcripts
NM_006734.3 XM_017010805.1

SNP Detail

SNP ID Chromosome Location
rs1014566531 Chr.6: 142752667-142752667 on Build GRCh38
rs190204297 Chr.6: 142753049-142753049 on Build GRCh38
rs886706430 Chr.6: 142752993-142752993 on Build GRCh38
rs779098986 Chr.6: 142752784-142752784 on Build GRCh38
rs898918423 Chr.6: 142752688-142752688 on Build GRCh38
rs755082434 Chr.6: 142752736-142752736 on Build GRCh38
rs58500519 Chr.6: 142752997-142752998 on Build GRCh38
rs553259891 Chr.6: 142753054-142753054 on Build GRCh38
rs746250164 Chr.6: 142752939-142752939 on Build GRCh38
rs919627364 Chr.6: 142752875-142752875 on Build GRCh38
rs763153791 Chr.6: 142753066-142753066 on Build GRCh38
rs1025262494 Chr.6: 142752813-142752813 on Build GRCh38
rs535987024 Chr.6: 142753058-142753060 on Build GRCh38
rs959598594 Chr.6: 142752766-142752766 on Build GRCh38
rs1004585439 Chr.6: 142752664-142752664 on Build GRCh38
rs184870965 Chr.6: 142753040-142753040 on Build GRCh38
rs978070260 Chr.6: 142752890-142752890 on Build GRCh38
rs981259141 Chr.6: 142752663-142752663 on Build GRCh38
rs761192737 Chr.6: 142753057-142753060 on Build GRCh38
rs552089515 Chr.6: 142753009-142753009 on Build GRCh38
rs148467224 Chr.6: 142752992-142752993 on Build GRCh38
rs971377738 Chr.6: 142752836-142752836 on Build GRCh38
rs1020167115 Chr.6: 142752613-142752613 on Build GRCh38
rs747727466 Chr.6: 142752580-142752582 on Build GRCh38
rs994567349 Chr.6: 142752700-142752700 on Build GRCh38
rs990518693 Chr.6: 142752610-142752613 on Build GRCh38
rs923902822 Chr.6: 142752975-142752975 on Build GRCh38
rs748603830 Chr.6: 142752814-142752814 on Build GRCh38
rs776915196 Chr.6: 142752847-142752847 on Build GRCh38
rs931000208 Chr.6: 142752928-142752928 on Build GRCh38
rs197478 Chr.6: 142752774-142752774 on Build GRCh38
rs1047139092 Chr.6: 142752963-142752963 on Build GRCh38
rs181815786 Chr.6: 142752972-142752972 on Build GRCh38
rs957390426 Chr.6: 142752586-142752586 on Build GRCh38
rs1012813663 Chr.6: 142752770-142752770 on Build GRCh38
rs1029441264 Chr.6: 142752575-142752575 on Build GRCh38
rs890367198 Chr.6: 142752638-142752638 on Build GRCh38
rs547765078 Chr.6: 142752858-142752858 on Build GRCh38
rs189185342 Chr.6: 142752869-142752869 on Build GRCh38
rs967239509 Chr.6: 142752618-142752621 on Build GRCh38
rs529667309 Chr.6: 142752841-142752841 on Build GRCh38
rs777330554 Chr.6: 142753060-142753061 on Build GRCh38
rs991247459 Chr.6: 142752862-142752862 on Build GRCh38
rs758934606 Chr.6: 142752996-142752996 on Build GRCh38

View Full Product Details