Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 142764615-142765130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIVEP2
Entrez Gene ID
3097
Gene Name
human immunodeficiency virus type I enhancer binding protein 2
Gene Aliases
HIV-EP2, MBP-2, MIBP1, MRD43, SHN2, ZAS2, ZNF40B
UniGene
Hs.510172
Gene Chromosome Location
Chr.6: 142751467-142945201 on Build GRCh38
Transcripts
NM_006734.3 XM_017010805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7202649 c.5406A>G p.E1802E Chr.6: 142764911-142764911 on Build GRCh38
339500 c.5478T>C p.P1826P Chr.6: 142764839-142764839 on Build GRCh38
5743155 c.5386C>T p.R1796W Chr.6: 142764931-142764931 on Build GRCh38
4690494 c.5440C>T p.L1814F Chr.6: 142764877-142764877 on Build GRCh38
5061456 c.5393delA p.K1798fs*45 Chr.6: 142764924-142764924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952863483 Chr.6: 142764691-142764694 on Build GRCh38
rs751235003 Chr.6: 142764950-142764950 on Build GRCh38
rs917845332 Chr.6: 142764848-142764848 on Build GRCh38
rs755666518 Chr.6: 142764984-142764984 on Build GRCh38
rs746357756 Chr.6: 142764995-142764995 on Build GRCh38
rs995586906 Chr.6: 142764741-142764741 on Build GRCh38
rs1015522835 Chr.6: 142764653-142764653 on Build GRCh38
rs754149906 Chr.6: 142765053-142765053 on Build GRCh38
rs376839134 Chr.6: 142764704-142764704 on Build GRCh38
rs776650343 Chr.6: 142764868-142764868 on Build GRCh38
rs772915584 Chr.6: 142764909-142764909 on Build GRCh38
rs747262699 Chr.6: 142765000-142765000 on Build GRCh38
rs942936493 Chr.6: 142764763-142764763 on Build GRCh38
rs763723664 Chr.6: 142765063-142765063 on Build GRCh38
rs9496460 Chr.6: 142764662-142764662 on Build GRCh38
rs765045837 Chr.6: 142764756-142764756 on Build GRCh38
rs775848520 Chr.6: 142764803-142764803 on Build GRCh38
rs760888393 Chr.6: 142764966-142764966 on Build GRCh38
rs749820767 Chr.6: 142764764-142764764 on Build GRCh38
rs766071612 Chr.6: 142764942-142764942 on Build GRCh38
rs368426840 Chr.6: 142764761-142764761 on Build GRCh38
rs1035670663 Chr.6: 142764654-142764654 on Build GRCh38
rs750329426 Chr.6: 142764930-142764930 on Build GRCh38
rs375701227 Chr.6: 142765026-142765026 on Build GRCh38
rs760795371 Chr.6: 142764857-142764857 on Build GRCh38
rs376079707 Chr.6: 142764758-142764758 on Build GRCh38
rs571810265 Chr.6: 142764639-142764639 on Build GRCh38
rs371652249 Chr.6: 142764779-142764779 on Build GRCh38
rs61729356 Chr.6: 142764920-142764920 on Build GRCh38
rs538262135 Chr.6: 142764747-142764747 on Build GRCh38
rs771528173 Chr.6: 142764767-142764767 on Build GRCh38
rs370467838 Chr.6: 142764759-142764759 on Build GRCh38
rs754518106 Chr.6: 142764952-142764952 on Build GRCh38
rs752208914 Chr.6: 142764967-142764967 on Build GRCh38
rs572706523 Chr.6: 142764781-142764781 on Build GRCh38
rs374184023 Chr.6: 142764794-142764794 on Build GRCh38
rs113059816 Chr.6: 142764999-142764999 on Build GRCh38
rs760759241 Chr.6: 142764866-142764866 on Build GRCh38
rs972806739 Chr.6: 142764668-142764668 on Build GRCh38
rs142233940 Chr.6: 142765057-142765057 on Build GRCh38
rs761759600 Chr.6: 142764902-142764902 on Build GRCh38
rs747358218 Chr.6: 142764779-142764780 on Build GRCh38
rs936319029 Chr.6: 142765056-142765056 on Build GRCh38
rs183415440 Chr.6: 142764746-142764746 on Build GRCh38
rs757130454 Chr.6: 142764755-142764755 on Build GRCh38
rs771945209 Chr.6: 142764675-142764675 on Build GRCh38
rs779607778 Chr.6: 142764991-142764991 on Build GRCh38
rs187534217 Chr.6: 142764762-142764762 on Build GRCh38
rs918667965 Chr.6: 142764678-142764678 on Build GRCh38
rs771463099 Chr.6: 142764780-142764780 on Build GRCh38
rs766526010 Chr.6: 142765024-142765024 on Build GRCh38
rs772293727 Chr.6: 142764802-142764802 on Build GRCh38
rs267600845 Chr.6: 142764927-142764927 on Build GRCh38
rs561647507 Chr.6: 142764965-142764965 on Build GRCh38
rs762868060 Chr.6: 142764937-142764937 on Build GRCh38

View Full Product Details