Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 146357402-146357949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRM1
Entrez Gene ID
2911
Gene Name
glutamate metabotropic receptor 1
Gene Aliases
GPRC1A, MGLU1, MGLUR1, PPP1R85, SCAR13
UniGene
Hs.32945
Gene Chromosome Location
Chr.6: 146027634-146437598 on Build GRCh38
Transcripts
NM_001278064.1 NM_001278065.1 NM_001278066.1 NM_001278067.1 XM_011535782.1 XM_017010783.1 XM_017010784.1 XM_017010785.1 XM_017010786.1 XM_017010787.1 XM_017010788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
328202 c.1458C>A p.Y486* Chr.6: 146357550-146357550 on Build GRCh38
483498 c.1495A>T p.T499S Chr.6: 146357587-146357587 on Build GRCh38
4828284 c.1489G>A p.V497I Chr.6: 146357581-146357581 on Build GRCh38
3829243 c.1462G>A p.E488K Chr.6: 146357554-146357554 on Build GRCh38
3621333 c.1536C>T p.I512I Chr.6: 146357628-146357628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767873257 Chr.6: 146357586-146357586 on Build GRCh38
rs762021245 Chr.6: 146357717-146357717 on Build GRCh38
rs362890 Chr.6: 146357473-146357473 on Build GRCh38
rs746401653 Chr.6: 146357479-146357479 on Build GRCh38
rs897673908 Chr.6: 146357770-146357770 on Build GRCh38
rs982371967 Chr.6: 146357801-146357801 on Build GRCh38
rs1004217239 Chr.6: 146357904-146357904 on Build GRCh38
rs545819604 Chr.6: 146357815-146357815 on Build GRCh38
rs141568265 Chr.6: 146357580-146357580 on Build GRCh38
rs770859697 Chr.6: 146357518-146357518 on Build GRCh38
rs957663689 Chr.6: 146357775-146357775 on Build GRCh38
rs746603273 Chr.6: 146357563-146357563 on Build GRCh38
rs776639922 Chr.6: 146357526-146357526 on Build GRCh38
rs956760556 Chr.6: 146357648-146357648 on Build GRCh38
rs368772965 Chr.6: 146357616-146357616 on Build GRCh38
rs750766383 Chr.6: 146357732-146357732 on Build GRCh38
rs920632383 Chr.6: 146357853-146357854 on Build GRCh38
rs578186460 Chr.6: 146357810-146357810 on Build GRCh38
rs747969826 Chr.6: 146357447-146357447 on Build GRCh38
rs759510081 Chr.6: 146357536-146357536 on Build GRCh38
rs756633092 Chr.6: 146357607-146357607 on Build GRCh38
rs362891 Chr.6: 146357496-146357496 on Build GRCh38
rs371024122 Chr.6: 146357716-146357716 on Build GRCh38
rs558827511 Chr.6: 146357790-146357790 on Build GRCh38
rs930354783 Chr.6: 146357779-146357779 on Build GRCh38
rs35301667 Chr.6: 146357817-146357818 on Build GRCh38
rs865874656 Chr.6: 146357530-146357530 on Build GRCh38
rs750458105 Chr.6: 146357483-146357484 on Build GRCh38
rs377302722 Chr.6: 146357715-146357715 on Build GRCh38
rs867821187 Chr.6: 146357570-146357570 on Build GRCh38
rs146507714 Chr.6: 146357676-146357676 on Build GRCh38
rs973274570 Chr.6: 146357834-146357834 on Build GRCh38
rs868511150 Chr.6: 146357571-146357571 on Build GRCh38
rs758003743 Chr.6: 146357631-146357631 on Build GRCh38
rs903453655 Chr.6: 146357721-146357721 on Build GRCh38
rs362892 Chr.6: 146357746-146357746 on Build GRCh38
rs765142468 Chr.6: 146357557-146357557 on Build GRCh38
rs200049230 Chr.6: 146357497-146357497 on Build GRCh38
rs990569804 Chr.6: 146357792-146357792 on Build GRCh38
rs1050563587 Chr.6: 146357912-146357912 on Build GRCh38
rs775619516 Chr.6: 146357561-146357561 on Build GRCh38
rs775323670 Chr.6: 146357694-146357694 on Build GRCh38
rs906750770 Chr.6: 146357896-146357896 on Build GRCh38
rs362894 Chr.6: 146357816-146357816 on Build GRCh38
rs757559798 Chr.6: 146357581-146357581 on Build GRCh38
rs775922600 Chr.6: 146357865-146357865 on Build GRCh38
rs767985221 Chr.6: 146357484-146357484 on Build GRCh38
rs1027294196 Chr.6: 146357480-146357480 on Build GRCh38
rs773029489 Chr.6: 146357513-146357513 on Build GRCh38
rs899128781 Chr.6: 146357617-146357617 on Build GRCh38
rs397715407 Chr.6: 146357821-146357822 on Build GRCh38
rs777510399 Chr.6: 146357643-146357643 on Build GRCh38
rs911947413 Chr.6: 146357916-146357916 on Build GRCh38
rs796569247 Chr.6: 146357444-146357446 on Build GRCh38
rs1036162497 Chr.6: 146357768-146357768 on Build GRCh38
rs982263350 Chr.6: 146357595-146357595 on Build GRCh38
rs1045814701 Chr.6: 146357895-146357895 on Build GRCh38
rs770770147 Chr.6: 146357654-146357654 on Build GRCh38
rs767788577 Chr.6: 146357730-146357730 on Build GRCh38
rs569951616 Chr.6: 146357671-146357671 on Build GRCh38
rs760497336 Chr.6: 146357516-146357516 on Build GRCh38
rs71546209 Chr.6: 146357816-146357817 on Build GRCh38
rs539867859 Chr.6: 146357428-146357428 on Build GRCh38
rs183890088 Chr.6: 146357760-146357760 on Build GRCh38
rs932054515 Chr.6: 146357646-146357646 on Build GRCh38
rs376313056 Chr.6: 146357703-146357703 on Build GRCh38
rs750821948 Chr.6: 146357602-146357602 on Build GRCh38
rs564428059 Chr.6: 146357623-146357623 on Build GRCh38
rs202196179 Chr.6: 146357486-146357486 on Build GRCh38
rs989543701 Chr.6: 146357652-146357652 on Build GRCh38
rs362893 Chr.6: 146357793-146357793 on Build GRCh38
rs764264066 Chr.6: 146357564-146357564 on Build GRCh38
rs746660819 Chr.6: 146357653-146357653 on Build GRCh38

View Full Product Details