Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23253115-23253640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPNMB
Entrez Gene ID
10457
Gene Name
glycoprotein nmb
Gene Aliases
HGFIN, NMB
UniGene
Hs.190495
Gene Chromosome Location
Chr.7: 23246206-23275110 on Build GRCh38
Transcripts
NM_001005340.1 NM_002510.2 XM_005249578.2 XM_017011676.1 XM_017011677.1 XM_017011678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4187956 c.168A>C p.E56D Chr.7: 23253404-23253404 on Build GRCh38
3880265 c.199A>G p.M67V Chr.7: 23253435-23253435 on Build GRCh38
6548083 c.107C>T p.A36V Chr.7: 23253343-23253343 on Build GRCh38
1450054 c.133A>C p.N45H Chr.7: 23253369-23253369 on Build GRCh38
1088585 c.145T>A p.S49T Chr.7: 23253381-23253381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111839421 Chr.7: 23253325-23253325 on Build GRCh38
rs568499910 Chr.7: 23253150-23253150 on Build GRCh38
rs980080399 Chr.7: 23253197-23253197 on Build GRCh38
rs962919380 Chr.7: 23253576-23253576 on Build GRCh38
rs763426950 Chr.7: 23253416-23253416 on Build GRCh38
rs911158230 Chr.7: 23253585-23253585 on Build GRCh38
rs540911280 Chr.7: 23253360-23253360 on Build GRCh38
rs993346971 Chr.7: 23253149-23253149 on Build GRCh38
rs757911496 Chr.7: 23253487-23253487 on Build GRCh38
rs775707788 Chr.7: 23253414-23253414 on Build GRCh38
rs933921102 Chr.7: 23253200-23253200 on Build GRCh38
rs773027372 Chr.7: 23253281-23253281 on Build GRCh38
rs199348 Chr.7: 23253479-23253479 on Build GRCh38
rs994359321 Chr.7: 23253340-23253340 on Build GRCh38
rs548504911 Chr.7: 23253560-23253560 on Build GRCh38
rs375702302 Chr.7: 23253280-23253280 on Build GRCh38
rs777506338 Chr.7: 23253383-23253383 on Build GRCh38
rs572130500 Chr.7: 23253514-23253514 on Build GRCh38
rs765705175 Chr.7: 23253293-23253293 on Build GRCh38
rs538063242 Chr.7: 23253433-23253433 on Build GRCh38
rs1881203 Chr.7: 23253189-23253189 on Build GRCh38
rs918862502 Chr.7: 23253586-23253586 on Build GRCh38
rs767598502 Chr.7: 23253428-23253428 on Build GRCh38
rs760872488 Chr.7: 23253440-23253440 on Build GRCh38
rs764723608 Chr.7: 23253300-23253300 on Build GRCh38
rs761545520 Chr.7: 23253512-23253513 on Build GRCh38
rs746669002 Chr.7: 23253389-23253389 on Build GRCh38
rs568910498 Chr.7: 23253198-23253198 on Build GRCh38
rs759927081 Chr.7: 23253292-23253292 on Build GRCh38
rs972982234 Chr.7: 23253580-23253580 on Build GRCh38
rs369282413 Chr.7: 23253426-23253426 on Build GRCh38
rs751571243 Chr.7: 23253307-23253307 on Build GRCh38
rs771726642 Chr.7: 23253377-23253377 on Build GRCh38
rs368200780 Chr.7: 23253507-23253507 on Build GRCh38
rs747740369 Chr.7: 23253273-23253273 on Build GRCh38
rs568492591 Chr.7: 23253591-23253591 on Build GRCh38
rs537562192 Chr.7: 23253596-23253596 on Build GRCh38
rs750568068 Chr.7: 23253432-23253432 on Build GRCh38
rs933825438 Chr.7: 23253568-23253568 on Build GRCh38
rs200933390 Chr.7: 23253299-23253299 on Build GRCh38
rs1031486645 Chr.7: 23253527-23253527 on Build GRCh38
rs148889150 Chr.7: 23253541-23253541 on Build GRCh38
rs374315358 Chr.7: 23253365-23253365 on Build GRCh38
rs538947475 Chr.7: 23253268-23253268 on Build GRCh38
rs753722741 Chr.7: 23253348-23253348 on Build GRCh38
rs986993821 Chr.7: 23253613-23253613 on Build GRCh38
rs1016924423 Chr.7: 23253563-23253563 on Build GRCh38
rs115671841 Chr.7: 23253159-23253159 on Build GRCh38
rs769015018 Chr.7: 23253422-23253422 on Build GRCh38
rs755840415 Chr.7: 23253338-23253338 on Build GRCh38
rs770470655 Chr.7: 23253499-23253499 on Build GRCh38
rs773719457 Chr.7: 23253272-23253272 on Build GRCh38
rs542987086 Chr.7: 23253475-23253475 on Build GRCh38
rs891601610 Chr.7: 23253532-23253532 on Build GRCh38
rs942980922 Chr.7: 23253441-23253441 on Build GRCh38
rs574394407 Chr.7: 23253494-23253494 on Build GRCh38
rs752691373 Chr.7: 23253486-23253486 on Build GRCh38
rs750665895 Chr.7: 23253331-23253331 on Build GRCh38
rs766671694 Chr.7: 23253443-23253443 on Build GRCh38
rs776003401 Chr.7: 23253295-23253295 on Build GRCh38
rs112804048 Chr.7: 23253608-23253608 on Build GRCh38
rs143597331 Chr.7: 23253559-23253559 on Build GRCh38
rs138559016 Chr.7: 23253501-23253501 on Build GRCh38
rs1025810502 Chr.7: 23253402-23253402 on Build GRCh38
rs1024778819 Chr.7: 23253554-23253554 on Build GRCh38
rs746579231 Chr.7: 23253489-23253489 on Build GRCh38
rs78208743 Chr.7: 23253526-23253526 on Build GRCh38
rs199840471 Chr.7: 23253427-23253427 on Build GRCh38
rs773931147 Chr.7: 23253337-23253339 on Build GRCh38
rs907368787 Chr.7: 23253417-23253417 on Build GRCh38
rs574453231 Chr.7: 23253468-23253468 on Build GRCh38
rs956138531 Chr.7: 23253141-23253141 on Build GRCh38
rs185366425 Chr.7: 23253317-23253317 on Build GRCh38
rs778947910 Chr.7: 23253361-23253361 on Build GRCh38
rs180914138 Chr.7: 23253218-23253218 on Build GRCh38
rs770778100 Chr.7: 23253407-23253407 on Build GRCh38
rs779709220 Chr.7: 23253347-23253347 on Build GRCh38

View Full Product Details