Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23267425-23267972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPNMB
Entrez Gene ID
10457
Gene Name
glycoprotein nmb
Gene Aliases
HGFIN, NMB
UniGene
Hs.190495
Gene Chromosome Location
Chr.7: 23246206-23275110 on Build GRCh38
Transcripts
NM_001005340.1 NM_002510.2 XM_005249578.2 XM_017011676.1 XM_017011677.1 XM_017011678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6177312 c.1172A>G p.N391S Chr.7: 23267904-23267904 on Build GRCh38
6536184 c.1227A>G p.L409L Chr.7: 23267959-23267959 on Build GRCh38
4959155 c.1165G>A p.E389K Chr.7: 23267897-23267897 on Build GRCh38
4361734 c.1202C>T p.P401L Chr.7: 23267934-23267934 on Build GRCh38
3026161 c.1192G>A p.V398I Chr.7: 23267924-23267924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015846199 Chr.7: 23267714-23267714 on Build GRCh38
rs75523457 Chr.7: 23267789-23267789 on Build GRCh38
rs944937332 Chr.7: 23267795-23267795 on Build GRCh38
rs900531206 Chr.7: 23267601-23267601 on Build GRCh38
rs763141813 Chr.7: 23267910-23267910 on Build GRCh38
rs915707677 Chr.7: 23267775-23267775 on Build GRCh38
rs560187413 Chr.7: 23267713-23267713 on Build GRCh38
rs1015503692 Chr.7: 23267485-23267486 on Build GRCh38
rs773678089 Chr.7: 23267872-23267872 on Build GRCh38
rs749937697 Chr.7: 23267737-23267737 on Build GRCh38
rs370763005 Chr.7: 23267786-23267786 on Build GRCh38
rs1040587040 Chr.7: 23267798-23267798 on Build GRCh38
rs368472544 Chr.7: 23267638-23267638 on Build GRCh38
rs78993277 Chr.7: 23267777-23267777 on Build GRCh38
rs965621252 Chr.7: 23267536-23267536 on Build GRCh38
rs995557007 Chr.7: 23267492-23267492 on Build GRCh38
rs887893891 Chr.7: 23267668-23267668 on Build GRCh38
rs764341605 Chr.7: 23267918-23267918 on Build GRCh38
rs961251026 Chr.7: 23267715-23267715 on Build GRCh38
rs156426 Chr.7: 23267856-23267856 on Build GRCh38
rs112828318 Chr.7: 23267785-23267785 on Build GRCh38
rs79806006 Chr.7: 23267527-23267527 on Build GRCh38
rs373768935 Chr.7: 23267873-23267873 on Build GRCh38
rs913444105 Chr.7: 23267571-23267571 on Build GRCh38
rs375028722 Chr.7: 23267919-23267919 on Build GRCh38
rs996159556 Chr.7: 23267616-23267616 on Build GRCh38
rs754682519 Chr.7: 23267935-23267935 on Build GRCh38
rs781344096 Chr.7: 23267719-23267719 on Build GRCh38
rs922217895 Chr.7: 23267814-23267814 on Build GRCh38
rs370506567 Chr.7: 23267897-23267897 on Build GRCh38
rs961324911 Chr.7: 23267489-23267489 on Build GRCh38
rs901193841 Chr.7: 23267823-23267823 on Build GRCh38
rs1005381904 Chr.7: 23267682-23267682 on Build GRCh38
rs988959426 Chr.7: 23267791-23267791 on Build GRCh38
rs751383970 Chr.7: 23267540-23267540 on Build GRCh38
rs368337839 Chr.7: 23267923-23267923 on Build GRCh38
rs936172618 Chr.7: 23267538-23267538 on Build GRCh38
rs758683361 Chr.7: 23267844-23267844 on Build GRCh38
rs777529242 Chr.7: 23267846-23267846 on Build GRCh38
rs997021932 Chr.7: 23267824-23267824 on Build GRCh38
rs1055726547 Chr.7: 23267751-23267751 on Build GRCh38
rs372548115 Chr.7: 23267924-23267924 on Build GRCh38
rs546747856 Chr.7: 23267686-23267686 on Build GRCh38
rs370089558 Chr.7: 23267858-23267858 on Build GRCh38
rs533454955 Chr.7: 23267778-23267778 on Build GRCh38
rs156427 Chr.7: 23267524-23267524 on Build GRCh38
rs768482896 Chr.7: 23267868-23267868 on Build GRCh38
rs765353523 Chr.7: 23267895-23267895 on Build GRCh38
rs544077732 Chr.7: 23267570-23267570 on Build GRCh38
rs748845826 Chr.7: 23267938-23267938 on Build GRCh38
rs1049134952 Chr.7: 23267651-23267651 on Build GRCh38
rs756077015 Chr.7: 23267934-23267934 on Build GRCh38
rs1004196800 Chr.7: 23267792-23267792 on Build GRCh38
rs757113692 Chr.7: 23267847-23267847 on Build GRCh38
rs376499551 Chr.7: 23267857-23267857 on Build GRCh38
rs1027485823 Chr.7: 23267496-23267496 on Build GRCh38
rs916927518 Chr.7: 23267501-23267501 on Build GRCh38
rs541716092 Chr.7: 23267523-23267523 on Build GRCh38
rs997071484 Chr.7: 23267569-23267569 on Build GRCh38
rs571539276 Chr.7: 23267854-23267854 on Build GRCh38
rs570745219 Chr.7: 23267579-23267579 on Build GRCh38
rs757074815 Chr.7: 23267617-23267617 on Build GRCh38
rs991401626 Chr.7: 23267936-23267936 on Build GRCh38
rs534843459 Chr.7: 23267821-23267821 on Build GRCh38
rs989879675 Chr.7: 23267585-23267585 on Build GRCh38
rs894332897 Chr.7: 23267482-23267482 on Build GRCh38
rs977519986 Chr.7: 23267716-23267716 on Build GRCh38
rs759467497 Chr.7: 23267887-23267887 on Build GRCh38
rs777204409 Chr.7: 23267884-23267884 on Build GRCh38
rs1040326529 Chr.7: 23267597-23267597 on Build GRCh38
rs533045831 Chr.7: 23267613-23267613 on Build GRCh38
rs759368009 Chr.7: 23267728-23267728 on Build GRCh38
rs969698609 Chr.7: 23267771-23267771 on Build GRCh38
rs750383396 Chr.7: 23267930-23267930 on Build GRCh38
rs141070290 Chr.7: 23267855-23267855 on Build GRCh38
rs538408791 Chr.7: 23267816-23267816 on Build GRCh38
rs569456391 Chr.7: 23267811-23267811 on Build GRCh38
rs760321071 Chr.7: 23267459-23267459 on Build GRCh38

View Full Product Details