Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23267884-23268420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPNMB
Entrez Gene ID
10457
Gene Name
glycoprotein nmb
Gene Aliases
HGFIN, NMB
UniGene
Hs.190495
Gene Chromosome Location
Chr.7: 23246206-23275110 on Build GRCh38
Transcripts
NM_001005340.1 NM_002510.2 XM_005249578.2 XM_017011676.1 XM_017011677.1 XM_017011678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6177312 c.1172A>G p.N391S Chr.7: 23267904-23267904 on Build GRCh38
6536184 c.1227A>G p.L409L Chr.7: 23267959-23267959 on Build GRCh38
4959155 c.1165G>A p.E389K Chr.7: 23267897-23267897 on Build GRCh38
4361734 c.1202C>T p.P401L Chr.7: 23267934-23267934 on Build GRCh38
3026161 c.1192G>A p.V398I Chr.7: 23267924-23267924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991182380 Chr.7: 23268151-23268151 on Build GRCh38
rs778637813 Chr.7: 23268028-23268028 on Build GRCh38
rs10560802 Chr.7: 23268021-23268024 on Build GRCh38
rs915864570 Chr.7: 23267952-23267952 on Build GRCh38
rs959685970 Chr.7: 23268131-23268131 on Build GRCh38
rs976804547 Chr.7: 23268222-23268222 on Build GRCh38
rs763141813 Chr.7: 23267910-23267910 on Build GRCh38
rs892777764 Chr.7: 23268281-23268281 on Build GRCh38
rs79639478 Chr.7: 23268301-23268301 on Build GRCh38
rs564089804 Chr.7: 23268233-23268236 on Build GRCh38
rs369336246 Chr.7: 23268000-23268000 on Build GRCh38
rs1022868396 Chr.7: 23268154-23268154 on Build GRCh38
rs371991773 Chr.7: 23268235-23268238 on Build GRCh38
rs764341605 Chr.7: 23267918-23267918 on Build GRCh38
rs766179587 Chr.7: 23268021-23268023 on Build GRCh38
rs953627292 Chr.7: 23268309-23268309 on Build GRCh38
rs753346880 Chr.7: 23268014-23268014 on Build GRCh38
rs949861304 Chr.7: 23268164-23268164 on Build GRCh38
rs1003746656 Chr.7: 23268396-23268396 on Build GRCh38
rs746371384 Chr.7: 23267977-23267977 on Build GRCh38
rs879057730 Chr.7: 23268086-23268086 on Build GRCh38
rs375028722 Chr.7: 23267919-23267919 on Build GRCh38
rs754682519 Chr.7: 23267935-23267935 on Build GRCh38
rs922726081 Chr.7: 23268226-23268226 on Build GRCh38
rs140819013 Chr.7: 23267971-23267971 on Build GRCh38
rs896606058 Chr.7: 23268152-23268152 on Build GRCh38
rs1032655007 Chr.7: 23268261-23268261 on Build GRCh38
rs879656973 Chr.7: 23268279-23268279 on Build GRCh38
rs764251856 Chr.7: 23267996-23267996 on Build GRCh38
rs747832199 Chr.7: 23268036-23268036 on Build GRCh38
rs564253591 Chr.7: 23268117-23268117 on Build GRCh38
rs932643905 Chr.7: 23268246-23268246 on Build GRCh38
rs982036490 Chr.7: 23268080-23268081 on Build GRCh38
rs77906578 Chr.7: 23268190-23268190 on Build GRCh38
rs965809082 Chr.7: 23268300-23268300 on Build GRCh38
rs367548805 Chr.7: 23267991-23267991 on Build GRCh38
rs754545301 Chr.7: 23268019-23268019 on Build GRCh38
rs368337839 Chr.7: 23267923-23267923 on Build GRCh38
rs755844030 Chr.7: 23268017-23268017 on Build GRCh38
rs372548115 Chr.7: 23267924-23267924 on Build GRCh38
rs200582723 Chr.7: 23268001-23268001 on Build GRCh38
rs2307783 Chr.7: 23268016-23268019 on Build GRCh38
rs1011057369 Chr.7: 23268081-23268081 on Build GRCh38
rs770251584 Chr.7: 23267986-23267986 on Build GRCh38
rs376026669 Chr.7: 23267955-23267955 on Build GRCh38
rs1029125143 Chr.7: 23268303-23268303 on Build GRCh38
rs748845826 Chr.7: 23267938-23267938 on Build GRCh38
rs756077015 Chr.7: 23267934-23267934 on Build GRCh38
rs894091102 Chr.7: 23268058-23268058 on Build GRCh38
rs758043102 Chr.7: 23268040-23268040 on Build GRCh38
rs368572840 Chr.7: 23267976-23267976 on Build GRCh38
rs543852109 Chr.7: 23268081-23268096 on Build GRCh38
rs753731225 Chr.7: 23268025-23268025 on Build GRCh38
rs766448354 Chr.7: 23268006-23268006 on Build GRCh38
rs563165181 Chr.7: 23268326-23268326 on Build GRCh38
rs1021518739 Chr.7: 23268098-23268098 on Build GRCh38
rs193013106 Chr.7: 23268005-23268005 on Build GRCh38
rs1049961864 Chr.7: 23268035-23268035 on Build GRCh38
rs185214858 Chr.7: 23268251-23268251 on Build GRCh38
rs560298924 Chr.7: 23268216-23268216 on Build GRCh38
rs996844008 Chr.7: 23268027-23268027 on Build GRCh38
rs146238405 Chr.7: 23267972-23267972 on Build GRCh38
rs991401626 Chr.7: 23267936-23267936 on Build GRCh38
rs1009934937 Chr.7: 23268282-23268282 on Build GRCh38
rs746201106 Chr.7: 23268315-23268315 on Build GRCh38
rs1049741136 Chr.7: 23268291-23268291 on Build GRCh38
rs984067061 Chr.7: 23268166-23268166 on Build GRCh38
rs902677112 Chr.7: 23268391-23268391 on Build GRCh38
rs952711697 Chr.7: 23268165-23268165 on Build GRCh38
rs371531481 Chr.7: 23267992-23267992 on Build GRCh38
rs750383396 Chr.7: 23267930-23267930 on Build GRCh38
rs368057634 Chr.7: 23268054-23268054 on Build GRCh38
rs116622942 Chr.7: 23268127-23268127 on Build GRCh38
rs747408196 Chr.7: 23268112-23268112 on Build GRCh38

View Full Product Details