Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 26190605-26191146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPA2B1
Entrez Gene ID
3181
Gene Name
heterogeneous nuclear ribonucleoprotein A2/B1
Gene Aliases
HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1, HNRPB1, IBMPFD2, RNPA2, SNRPB1
UniGene
Hs.487774
Gene Chromosome Location
Chr.7: 26189927-26200793 on Build GRCh38
Transcripts
NM_002137.3 NM_031243.2 XM_005249729.1 XM_017012109.1 XM_017012110.1

SNP Detail

SNP ID Chromosome Location
rs1048808691 Chr.7: 26190687-26190687 on Build GRCh38
rs74741045 Chr.7: 26190797-26190797 on Build GRCh38
rs542914846 Chr.7: 26190694-26190694 on Build GRCh38
rs1030943110 Chr.7: 26191083-26191084 on Build GRCh38
rs902512333 Chr.7: 26190682-26190682 on Build GRCh38
rs377264305 Chr.7: 26190793-26190793 on Build GRCh38
rs987315000 Chr.7: 26190833-26190833 on Build GRCh38
rs1012834824 Chr.7: 26191009-26191009 on Build GRCh38
rs764745344 Chr.7: 26190778-26190780 on Build GRCh38
rs952291807 Chr.7: 26190775-26190775 on Build GRCh38
rs944550920 Chr.7: 26190865-26190867 on Build GRCh38
rs922353782 Chr.7: 26190826-26190826 on Build GRCh38
rs946666570 Chr.7: 26190881-26190881 on Build GRCh38
rs946614288 Chr.7: 26190676-26190676 on Build GRCh38
rs550062665 Chr.7: 26190779-26190779 on Build GRCh38
rs3210100 Chr.7: 26191075-26191075 on Build GRCh38
rs968265003 Chr.7: 26191066-26191067 on Build GRCh38
rs1056432862 Chr.7: 26190928-26190928 on Build GRCh38
rs3210099 Chr.7: 26191094-26191094 on Build GRCh38
rs1006501283 Chr.7: 26191064-26191064 on Build GRCh38
rs182284582 Chr.7: 26190914-26190914 on Build GRCh38
rs771055933 Chr.7: 26190699-26190701 on Build GRCh38
rs887931982 Chr.7: 26191041-26191041 on Build GRCh38
rs994241964 Chr.7: 26191077-26191077 on Build GRCh38
rs944622391 Chr.7: 26190649-26190649 on Build GRCh38
rs1043607763 Chr.7: 26190889-26190889 on Build GRCh38
rs559040556 Chr.7: 26190798-26190798 on Build GRCh38
rs756529815 Chr.7: 26191067-26191067 on Build GRCh38
rs572449829 Chr.7: 26190823-26190823 on Build GRCh38
rs548242725 Chr.7: 26190698-26190698 on Build GRCh38
rs886127338 Chr.7: 26190898-26190898 on Build GRCh38
rs956734914 Chr.7: 26191095-26191095 on Build GRCh38
rs985718078 Chr.7: 26190873-26190873 on Build GRCh38
rs1023764165 Chr.7: 26190749-26190749 on Build GRCh38
rs911080196 Chr.7: 26191120-26191120 on Build GRCh38
rs1020136363 Chr.7: 26191103-26191103 on Build GRCh38
rs1012308565 Chr.7: 26191006-26191006 on Build GRCh38
rs896564745 Chr.7: 26190975-26190975 on Build GRCh38
rs3210097 Chr.7: 26191100-26191100 on Build GRCh38
rs1048755677 Chr.7: 26190892-26190893 on Build GRCh38
rs555060593 Chr.7: 26190888-26190888 on Build GRCh38
rs3210104 Chr.7: 26191070-26191070 on Build GRCh38
rs896626350 Chr.7: 26190978-26190978 on Build GRCh38
rs751169051 Chr.7: 26190794-26190795 on Build GRCh38
rs1012836456 Chr.7: 26190746-26190746 on Build GRCh38
rs933716504 Chr.7: 26190834-26190834 on Build GRCh38
rs536874477 Chr.7: 26190753-26190753 on Build GRCh38
rs568032043 Chr.7: 26190724-26190724 on Build GRCh38
rs189790171 Chr.7: 26190875-26190875 on Build GRCh38
rs565325391 Chr.7: 26190648-26190648 on Build GRCh38
rs1023857590 Chr.7: 26191055-26191055 on Build GRCh38
rs999641856 Chr.7: 26190683-26190683 on Build GRCh38
rs930580500 Chr.7: 26190892-26190892 on Build GRCh38
rs1029308593 Chr.7: 26191087-26191087 on Build GRCh38
rs911715630 Chr.7: 26190837-26190837 on Build GRCh38
rs933810937 Chr.7: 26190639-26190639 on Build GRCh38
rs756443666 Chr.7: 26190663-26190663 on Build GRCh38
rs3210101 Chr.7: 26191074-26191074 on Build GRCh38
rs949799585 Chr.7: 26190999-26190999 on Build GRCh38
rs751134020 Chr.7: 26190879-26190879 on Build GRCh38
rs962255802 Chr.7: 26191069-26191069 on Build GRCh38
rs3210098 Chr.7: 26191097-26191097 on Build GRCh38
rs955315109 Chr.7: 26191101-26191101 on Build GRCh38
rs1049547906 Chr.7: 26190665-26190665 on Build GRCh38
rs776213579 Chr.7: 26190777-26190779 on Build GRCh38
rs986875040 Chr.7: 26191102-26191102 on Build GRCh38
rs4273 Chr.7: 26190669-26190669 on Build GRCh38
rs185008012 Chr.7: 26191018-26191018 on Build GRCh38
rs967855126 Chr.7: 26191122-26191122 on Build GRCh38
rs574809103 Chr.7: 26190968-26190968 on Build GRCh38
rs961188482 Chr.7: 26190733-26190733 on Build GRCh38
rs780994771 Chr.7: 26191114-26191114 on Build GRCh38
rs902559917 Chr.7: 26190907-26190907 on Build GRCh38
rs1043721552 Chr.7: 26190677-26190677 on Build GRCh38
rs1035174942 Chr.7: 26190714-26190714 on Build GRCh38
rs757798001 Chr.7: 26190769-26190769 on Build GRCh38
rs754618680 Chr.7: 26190904-26190904 on Build GRCh38
rs911754321 Chr.7: 26190629-26190629 on Build GRCh38
rs767946822 Chr.7: 26190883-26190883 on Build GRCh38

View Full Product Details