Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156724056-156724556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISG20L2
Entrez Gene ID
81875
Gene Name
interferon stimulated exonuclease gene 20 like 2
Gene Aliases
HSD38
UniGene
Hs.301904
Gene Chromosome Location
Chr.1: 156721891-156728599 on Build GRCh38
Transcripts
NM_001303095.1 XM_005245516.4 XM_005245518.4 XM_006711560.3 XM_006711561.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
676384 c.847A>T p.T283S Chr.1: 156724249-156724249 on Build GRCh38
1984147 c.858C>T p.I286I Chr.1: 156724238-156724238 on Build GRCh38
1984146 c.871C>T p.R291W Chr.1: 156724225-156724225 on Build GRCh38
676383 c.842G>A p.R281H Chr.1: 156724254-156724254 on Build GRCh38
4657098 c.793C>T p.H265Y Chr.1: 156724303-156724303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371302179 Chr.1: 156724132-156724132 on Build GRCh38
rs749501052 Chr.1: 156724158-156724158 on Build GRCh38
rs780089819 Chr.1: 156724366-156724366 on Build GRCh38
rs530019097 Chr.1: 156724364-156724364 on Build GRCh38
rs58891771 Chr.1: 156724398-156724398 on Build GRCh38
rs576582388 Chr.1: 156724242-156724242 on Build GRCh38
rs754961329 Chr.1: 156724318-156724318 on Build GRCh38
rs776973833 Chr.1: 156724265-156724265 on Build GRCh38
rs1033862623 Chr.1: 156724500-156724500 on Build GRCh38
rs759669515 Chr.1: 156724189-156724189 on Build GRCh38
rs761558950 Chr.1: 156724231-156724231 on Build GRCh38
rs775476547 Chr.1: 156724240-156724240 on Build GRCh38
rs574807006 Chr.1: 156724090-156724090 on Build GRCh38
rs746076196 Chr.1: 156724360-156724360 on Build GRCh38
rs778967770 Chr.1: 156724358-156724358 on Build GRCh38
rs560389558 Chr.1: 156724400-156724400 on Build GRCh38
rs140606389 Chr.1: 156724298-156724298 on Build GRCh38
rs756488990 Chr.1: 156724101-156724101 on Build GRCh38
rs1009738680 Chr.1: 156724481-156724481 on Build GRCh38
rs773078201 Chr.1: 156724485-156724485 on Build GRCh38
rs887199006 Chr.1: 156724144-156724144 on Build GRCh38
rs752941283 Chr.1: 156724098-156724098 on Build GRCh38
rs771338416 Chr.1: 156724159-156724159 on Build GRCh38
rs1019652391 Chr.1: 156724200-156724200 on Build GRCh38
rs776331913 Chr.1: 156724230-156724231 on Build GRCh38
rs199864903 Chr.1: 156724282-156724282 on Build GRCh38
rs758492361 Chr.1: 156724291-156724291 on Build GRCh38
rs981906652 Chr.1: 156724497-156724497 on Build GRCh38
rs778100229 Chr.1: 156724156-156724156 on Build GRCh38
rs368359063 Chr.1: 156724382-156724382 on Build GRCh38
rs774291656 Chr.1: 156724165-156724165 on Build GRCh38
rs764737110 Chr.1: 156724277-156724277 on Build GRCh38
rs958261300 Chr.1: 156724231-156724231 on Build GRCh38
rs751172798 Chr.1: 156724311-156724311 on Build GRCh38
rs116532670 Chr.1: 156724081-156724081 on Build GRCh38
rs200161207 Chr.1: 156724233-156724233 on Build GRCh38
rs535718202 Chr.1: 156724209-156724209 on Build GRCh38
rs112044207 Chr.1: 156724427-156724427 on Build GRCh38
rs768617970 Chr.1: 156724194-156724196 on Build GRCh38
rs756940358 Chr.1: 156724125-156724125 on Build GRCh38
rs776089491 Chr.1: 156724206-156724206 on Build GRCh38
rs756489489 Chr.1: 156724120-156724120 on Build GRCh38
rs778262611 Chr.1: 156724126-156724126 on Build GRCh38
rs899856097 Chr.1: 156724422-156724422 on Build GRCh38
rs753523353 Chr.1: 156724121-156724121 on Build GRCh38
rs746723852 Chr.1: 156724153-156724153 on Build GRCh38
rs1032276850 Chr.1: 156724431-156724431 on Build GRCh38
rs537511202 Chr.1: 156724482-156724482 on Build GRCh38
rs887090134 Chr.1: 156724404-156724404 on Build GRCh38
rs777135294 Chr.1: 156724374-156724374 on Build GRCh38
rs765467869 Chr.1: 156724366-156724367 on Build GRCh38
rs1037346466 Chr.1: 156724255-156724255 on Build GRCh38
rs187406351 Chr.1: 156724414-156724414 on Build GRCh38
rs762166772 Chr.1: 156724271-156724271 on Build GRCh38
rs146831275 Chr.1: 156724234-156724234 on Build GRCh38
rs772405530 Chr.1: 156724191-156724191 on Build GRCh38
rs562035671 Chr.1: 156724355-156724355 on Build GRCh38
rs768592548 Chr.1: 156724154-156724154 on Build GRCh38
rs1027408892 Chr.1: 156724107-156724107 on Build GRCh38
rs61736207 Chr.1: 156724232-156724232 on Build GRCh38
rs920523247 Chr.1: 156724268-156724268 on Build GRCh38
rs1047596835 Chr.1: 156724322-156724322 on Build GRCh38
rs767568584 Chr.1: 156724319-156724319 on Build GRCh38
rs769081758 Chr.1: 156724371-156724371 on Build GRCh38
rs190219758 Chr.1: 156724528-156724528 on Build GRCh38
rs549385361 Chr.1: 156724496-156724496 on Build GRCh38
rs144766964 Chr.1: 156724224-156724224 on Build GRCh38
rs35862203 Chr.1: 156724330-156724331 on Build GRCh38
rs371101829 Chr.1: 156724102-156724102 on Build GRCh38
rs201545907 Chr.1: 156724286-156724286 on Build GRCh38
rs748460849 Chr.1: 156724380-156724380 on Build GRCh38
rs762328954 Chr.1: 156724287-156724287 on Build GRCh38
rs750003427 Chr.1: 156724130-156724130 on Build GRCh38
rs769204504 Chr.1: 156724254-156724254 on Build GRCh38
rs376730745 Chr.1: 156724454-156724454 on Build GRCh38
rs141168468 Chr.1: 156724238-156724238 on Build GRCh38
rs745890632 Chr.1: 156724237-156724237 on Build GRCh38
rs747156270 Chr.1: 156724367-156724367 on Build GRCh38
rs771211670 Chr.1: 156724235-156724235 on Build GRCh38
rs780044263 Chr.1: 156724136-156724136 on Build GRCh38
rs200460667 Chr.1: 156724225-156724225 on Build GRCh38
rs773488563 Chr.1: 156724388-156724388 on Build GRCh38
rs1039529717 Chr.1: 156724263-156724263 on Build GRCh38
rs752486684 Chr.1: 156724345-156724345 on Build GRCh38
rs374960324 Chr.1: 156724241-156724241 on Build GRCh38
rs146104824 Chr.1: 156724208-156724208 on Build GRCh38
rs779212784 Chr.1: 156724236-156724236 on Build GRCh38
rs951447998 Chr.1: 156724124-156724124 on Build GRCh38

View Full Product Details