Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156742789-156743297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDGF
Entrez Gene ID
3068
Gene Name
hepatoma-derived growth factor
Gene Aliases
HMG1L2
UniGene
Hs.743948
Gene Chromosome Location
Chr.1: 156742107-156752448 on Build GRCh38
Transcripts
NM_001126050.1 NM_001126051.1 NM_001319186.1 NM_001319187.1 NM_001319188.1 NM_004494.2 NR_135008.1 XM_011509453.2 XM_011509454.2 XM_011509455.2

SNP Detail

SNP ID Chromosome Location
rs556376794 Chr.1: 156742959-156742959 on Build GRCh38
rs1015139286 Chr.1: 156743091-156743091 on Build GRCh38
rs369723450 Chr.1: 156743081-156743081 on Build GRCh38
rs866153960 Chr.1: 156743079-156743079 on Build GRCh38
rs900153718 Chr.1: 156742993-156742993 on Build GRCh38
rs757395551 Chr.1: 156742829-156742829 on Build GRCh38
rs11556977 Chr.1: 156743265-156743265 on Build GRCh38
rs1040328912 Chr.1: 156743258-156743258 on Build GRCh38
rs995809224 Chr.1: 156743024-156743024 on Build GRCh38
rs979409695 Chr.1: 156743178-156743178 on Build GRCh38
rs1027303633 Chr.1: 156743050-156743050 on Build GRCh38
rs199960326 Chr.1: 156742898-156742904 on Build GRCh38
rs73002713 Chr.1: 156742981-156742981 on Build GRCh38
rs577276057 Chr.1: 156742932-156742932 on Build GRCh38
rs1039957984 Chr.1: 156742991-156742991 on Build GRCh38
rs116207813 Chr.1: 156743117-156743117 on Build GRCh38
rs550698416 Chr.1: 156743211-156743211 on Build GRCh38
rs534268126 Chr.1: 156742954-156742954 on Build GRCh38
rs532200506 Chr.1: 156743209-156743209 on Build GRCh38
rs565052677 Chr.1: 156743156-156743156 on Build GRCh38
rs561181123 Chr.1: 156743048-156743048 on Build GRCh38
rs540058701 Chr.1: 156743120-156743120 on Build GRCh38
rs544732053 Chr.1: 156742934-156742934 on Build GRCh38
rs992464618 Chr.1: 156743172-156743172 on Build GRCh38
rs577504203 Chr.1: 156743078-156743078 on Build GRCh38
rs768366521 Chr.1: 156743198-156743198 on Build GRCh38
rs749563787 Chr.1: 156743175-156743175 on Build GRCh38
rs574912091 Chr.1: 156742968-156742968 on Build GRCh38
rs925345802 Chr.1: 156742833-156742833 on Build GRCh38
rs935406720 Chr.1: 156743206-156743207 on Build GRCh38
rs550444571 Chr.1: 156742931-156742931 on Build GRCh38
rs1004673046 Chr.1: 156743083-156743083 on Build GRCh38
rs781196137 Chr.1: 156743176-156743176 on Build GRCh38
rs781239733 Chr.1: 156742873-156742873 on Build GRCh38
rs568589896 Chr.1: 156743215-156743215 on Build GRCh38
rs539607564 Chr.1: 156743225-156743225 on Build GRCh38

View Full Product Details