Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 35673078-35673574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERPUD2
Entrez Gene ID
64224
Gene Name
HERPUD family member 2
Gene Aliases
-
UniGene
Hs.643051
Gene Chromosome Location
Chr.7: 35632659-35695187 on Build GRCh38
Transcripts
NM_022373.4 XM_006715765.1 XM_011515473.1 XM_017012506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6679528 c.217C>T p.L73F Chr.7: 35673209-35673209 on Build GRCh38
6679527 c.200T>C p.L67P Chr.7: 35673226-35673226 on Build GRCh38
6679526 c.221G>A p.R74K Chr.7: 35673205-35673205 on Build GRCh38
3366918 c.193G>A p.D65N Chr.7: 35673233-35673233 on Build GRCh38
4690161 c.178G>A p.G60S Chr.7: 35673248-35673248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569267917 Chr.7: 35673264-35673264 on Build GRCh38
rs150332990 Chr.7: 35673230-35673230 on Build GRCh38
rs757479703 Chr.7: 35673233-35673233 on Build GRCh38
rs759885848 Chr.7: 35673167-35673167 on Build GRCh38
rs750338415 Chr.7: 35673328-35673328 on Build GRCh38
rs376503739 Chr.7: 35673315-35673315 on Build GRCh38
rs754607758 Chr.7: 35673250-35673250 on Build GRCh38
rs199944743 Chr.7: 35673159-35673159 on Build GRCh38
rs372816333 Chr.7: 35673497-35673497 on Build GRCh38
rs781673336 Chr.7: 35673505-35673508 on Build GRCh38
rs778599545 Chr.7: 35673260-35673260 on Build GRCh38
rs763433248 Chr.7: 35673295-35673295 on Build GRCh38
rs143729755 Chr.7: 35673288-35673288 on Build GRCh38
rs1013926535 Chr.7: 35673277-35673277 on Build GRCh38
rs181625750 Chr.7: 35673437-35673437 on Build GRCh38
rs1043852618 Chr.7: 35673487-35673487 on Build GRCh38
rs537544608 Chr.7: 35673494-35673494 on Build GRCh38
rs1027753912 Chr.7: 35673392-35673392 on Build GRCh38
rs775152017 Chr.7: 35673135-35673135 on Build GRCh38
rs775697443 Chr.7: 35673290-35673290 on Build GRCh38
rs976114853 Chr.7: 35673442-35673442 on Build GRCh38
rs190085600 Chr.7: 35673431-35673431 on Build GRCh38
rs764460760 Chr.7: 35673227-35673227 on Build GRCh38
rs767641455 Chr.7: 35673172-35673172 on Build GRCh38
rs548745246 Chr.7: 35673321-35673321 on Build GRCh38
rs779680647 Chr.7: 35673244-35673244 on Build GRCh38
rs1000499007 Chr.7: 35673163-35673163 on Build GRCh38
rs925748817 Chr.7: 35673512-35673512 on Build GRCh38
rs771583826 Chr.7: 35673274-35673274 on Build GRCh38
rs146809212 Chr.7: 35673394-35673394 on Build GRCh38
rs750343475 Chr.7: 35673235-35673235 on Build GRCh38
rs935858190 Chr.7: 35673514-35673514 on Build GRCh38
rs755891404 Chr.7: 35673243-35673243 on Build GRCh38
rs553188084 Chr.7: 35673402-35673402 on Build GRCh38
rs770375066 Chr.7: 35673289-35673289 on Build GRCh38
rs867887997 Chr.7: 35673334-35673334 on Build GRCh38
rs368898237 Chr.7: 35673234-35673234 on Build GRCh38
rs752923035 Chr.7: 35673192-35673192 on Build GRCh38
rs755981068 Chr.7: 35673329-35673329 on Build GRCh38
rs764407427 Chr.7: 35673304-35673304 on Build GRCh38
rs932077641 Chr.7: 35673482-35673482 on Build GRCh38
rs113175369 Chr.7: 35673403-35673403 on Build GRCh38
rs921997821 Chr.7: 35673457-35673457 on Build GRCh38
rs567455461 Chr.7: 35673383-35673383 on Build GRCh38
rs777127208 Chr.7: 35673161-35673161 on Build GRCh38
rs1001637716 Chr.7: 35673356-35673356 on Build GRCh38
rs376783164 Chr.7: 35673212-35673212 on Build GRCh38
rs574923248 Chr.7: 35673459-35673459 on Build GRCh38
rs984074411 Chr.7: 35673413-35673413 on Build GRCh38
rs373518482 Chr.7: 35673182-35673182 on Build GRCh38
rs749154087 Chr.7: 35673249-35673249 on Build GRCh38
rs904423644 Chr.7: 35673108-35673108 on Build GRCh38
rs545542744 Chr.7: 35673429-35673429 on Build GRCh38

View Full Product Details