Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44826764-44827270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H2AFV
Entrez Gene ID
94239
Gene Name
H2A histone family member V
Gene Aliases
H2A.Z-2, H2AV
UniGene
Hs.488189
Gene Chromosome Location
Chr.7: 44826889-44848126 on Build GRCh38
Transcripts
NM_138635.3

SNP Detail

SNP ID Chromosome Location
rs901240360 Chr.7: 44826862-44826862 on Build GRCh38
rs971623161 Chr.7: 44826863-44826863 on Build GRCh38
rs566008994 Chr.7: 44826955-44826955 on Build GRCh38
rs1006180209 Chr.7: 44826901-44826901 on Build GRCh38
rs1049058146 Chr.7: 44826964-44826964 on Build GRCh38
rs930646173 Chr.7: 44826963-44826964 on Build GRCh38
rs2898 Chr.7: 44827034-44827034 on Build GRCh38
rs548036377 Chr.7: 44827083-44827083 on Build GRCh38
rs958609078 Chr.7: 44826815-44826815 on Build GRCh38
rs1026159403 Chr.7: 44826889-44826889 on Build GRCh38
rs958606407 Chr.7: 44827122-44827122 on Build GRCh38
rs747787398 Chr.7: 44827245-44827245 on Build GRCh38
rs911466702 Chr.7: 44827169-44827169 on Build GRCh38
rs970518299 Chr.7: 44827095-44827095 on Build GRCh38
rs1032677132 Chr.7: 44827107-44827107 on Build GRCh38
rs1053999272 Chr.7: 44827162-44827162 on Build GRCh38
rs972163761 Chr.7: 44826888-44826888 on Build GRCh38
rs1012862018 Chr.7: 44827151-44827151 on Build GRCh38
rs180785630 Chr.7: 44826846-44826846 on Build GRCh38
rs746190455 Chr.7: 44827099-44827099 on Build GRCh38
rs749829275 Chr.7: 44827201-44827201 on Build GRCh38
rs559078518 Chr.7: 44827161-44827161 on Build GRCh38
rs1010132542 Chr.7: 44827081-44827081 on Build GRCh38
rs1041020828 Chr.7: 44827052-44827052 on Build GRCh38
rs910799264 Chr.7: 44827009-44827009 on Build GRCh38
rs1041069626 Chr.7: 44827216-44827216 on Build GRCh38
rs902038953 Chr.7: 44827079-44827079 on Build GRCh38
rs578016020 Chr.7: 44827227-44827227 on Build GRCh38
rs371621361 Chr.7: 44826799-44826799 on Build GRCh38
rs781522414 Chr.7: 44827058-44827058 on Build GRCh38
rs7016 Chr.7: 44827233-44827233 on Build GRCh38
rs994637141 Chr.7: 44826869-44826869 on Build GRCh38
rs971864362 Chr.7: 44827200-44827200 on Build GRCh38
rs994256087 Chr.7: 44827232-44827232 on Build GRCh38
rs2897 Chr.7: 44827105-44827105 on Build GRCh38
rs751762809 Chr.7: 44826933-44826933 on Build GRCh38
rs919320717 Chr.7: 44826926-44826926 on Build GRCh38
rs950491266 Chr.7: 44826896-44826896 on Build GRCh38
rs991810461 Chr.7: 44826837-44826837 on Build GRCh38
rs1041555687 Chr.7: 44826830-44826830 on Build GRCh38
rs917549233 Chr.7: 44827071-44827071 on Build GRCh38
rs556258520 Chr.7: 44827112-44827112 on Build GRCh38
rs901134576 Chr.7: 44827226-44827226 on Build GRCh38
rs959324806 Chr.7: 44827018-44827018 on Build GRCh38
rs564801068 Chr.7: 44826848-44826848 on Build GRCh38
rs766710780 Chr.7: 44827229-44827229 on Build GRCh38
rs532365401 Chr.7: 44826924-44826924 on Build GRCh38
rs567329258 Chr.7: 44827100-44827100 on Build GRCh38
rs764159880 Chr.7: 44826871-44826871 on Build GRCh38
rs768216761 Chr.7: 44827213-44827213 on Build GRCh38
rs368110178 Chr.7: 44826938-44826938 on Build GRCh38

View Full Product Details