Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44829145-44829660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H2AFV
Entrez Gene ID
94239
Gene Name
H2A histone family member V
Gene Aliases
H2A.Z-2, H2AV
UniGene
Hs.488189
Gene Chromosome Location
Chr.7: 44826889-44848126 on Build GRCh38
Transcripts
NM_138635.3

SNP Detail

SNP ID Chromosome Location
rs745618441 Chr.7: 44829599-44829600 on Build GRCh38
rs11297798 Chr.7: 44829620-44829620 on Build GRCh38
rs367904167 Chr.7: 44829601-44829602 on Build GRCh38
rs928772759 Chr.7: 44829525-44829525 on Build GRCh38
rs1044582315 Chr.7: 44829516-44829516 on Build GRCh38
rs956551826 Chr.7: 44829261-44829261 on Build GRCh38
rs968316263 Chr.7: 44829381-44829381 on Build GRCh38
rs908180091 Chr.7: 44829171-44829171 on Build GRCh38
rs974264997 Chr.7: 44829485-44829485 on Build GRCh38
rs11764950 Chr.7: 44829432-44829432 on Build GRCh38
rs756854460 Chr.7: 44829536-44829537 on Build GRCh38
rs779840331 Chr.7: 44829601-44829603 on Build GRCh38
rs552539177 Chr.7: 44829554-44829554 on Build GRCh38
rs537794458 Chr.7: 44829604-44829604 on Build GRCh38
rs986682531 Chr.7: 44829457-44829457 on Build GRCh38
rs766803035 Chr.7: 44829474-44829474 on Build GRCh38
rs67292639 Chr.7: 44829601-44829601 on Build GRCh38
rs537938688 Chr.7: 44829371-44829371 on Build GRCh38
rs777028811 Chr.7: 44829563-44829563 on Build GRCh38
rs574238817 Chr.7: 44829576-44829576 on Build GRCh38
rs1045542297 Chr.7: 44829319-44829319 on Build GRCh38
rs547755369 Chr.7: 44829334-44829334 on Build GRCh38
rs1053053653 Chr.7: 44829367-44829367 on Build GRCh38
rs928010099 Chr.7: 44829336-44829336 on Build GRCh38
rs770097610 Chr.7: 44829604-44829605 on Build GRCh38
rs79024659 Chr.7: 44829603-44829603 on Build GRCh38
rs961030429 Chr.7: 44829478-44829478 on Build GRCh38
rs5883908 Chr.7: 44829599-44829599 on Build GRCh38
rs1005723827 Chr.7: 44829451-44829451 on Build GRCh38
rs765925417 Chr.7: 44829605-44829605 on Build GRCh38
rs923836547 Chr.7: 44829238-44829238 on Build GRCh38
rs869264245 Chr.7: 44829618-44829620 on Build GRCh38
rs184980641 Chr.7: 44829191-44829191 on Build GRCh38
rs530570383 Chr.7: 44829203-44829203 on Build GRCh38
rs1028693681 Chr.7: 44829176-44829176 on Build GRCh38
rs1003051170 Chr.7: 44829360-44829360 on Build GRCh38
rs796950609 Chr.7: 44829601-44829604 on Build GRCh38
rs761790944 Chr.7: 44829294-44829294 on Build GRCh38
rs564377738 Chr.7: 44829265-44829265 on Build GRCh38
rs537620421 Chr.7: 44829427-44829427 on Build GRCh38
rs566220042 Chr.7: 44829338-44829338 on Build GRCh38
rs749709675 Chr.7: 44829600-44829600 on Build GRCh38
rs899992715 Chr.7: 44829481-44829481 on Build GRCh38
rs989295683 Chr.7: 44829263-44829263 on Build GRCh38
rs955289951 Chr.7: 44829449-44829449 on Build GRCh38
rs1038572392 Chr.7: 44829454-44829454 on Build GRCh38
rs186682027 Chr.7: 44829380-44829380 on Build GRCh38
rs747485404 Chr.7: 44829431-44829431 on Build GRCh38
rs377416141 Chr.7: 44829604-44829604 on Build GRCh38
rs532428958 Chr.7: 44829316-44829316 on Build GRCh38
rs948450267 Chr.7: 44829515-44829515 on Build GRCh38
rs571100391 Chr.7: 44829492-44829492 on Build GRCh38
rs989802488 Chr.7: 44829635-44829635 on Build GRCh38
rs996675406 Chr.7: 44829421-44829421 on Build GRCh38
rs545788247 Chr.7: 44829215-44829215 on Build GRCh38
rs908210040 Chr.7: 44829473-44829473 on Build GRCh38
rs192337014 Chr.7: 44829526-44829526 on Build GRCh38
rs939760449 Chr.7: 44829190-44829190 on Build GRCh38
rs66566005 Chr.7: 44829372-44829372 on Build GRCh38
rs1008489268 Chr.7: 44829562-44829562 on Build GRCh38
rs567406450 Chr.7: 44829601-44829601 on Build GRCh38
rs956584363 Chr.7: 44829597-44829597 on Build GRCh38
rs1045066081 Chr.7: 44829281-44829281 on Build GRCh38
rs548656749 Chr.7: 44829375-44829375 on Build GRCh38
rs558458612 Chr.7: 44829534-44829534 on Build GRCh38
rs776790420 Chr.7: 44829603-44829605 on Build GRCh38
rs760421809 Chr.7: 44829603-44829604 on Build GRCh38

View Full Product Details