Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44829694-44830209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H2AFV
Entrez Gene ID
94239
Gene Name
H2A histone family member V
Gene Aliases
H2A.Z-2, H2AV
UniGene
Hs.488189
Gene Chromosome Location
Chr.7: 44826889-44848126 on Build GRCh38
Transcripts
NM_138635.3

SNP Detail

SNP ID Chromosome Location
rs1008051903 Chr.7: 44830105-44830105 on Build GRCh38
rs369560574 Chr.7: 44829843-44829843 on Build GRCh38
rs763128803 Chr.7: 44830133-44830133 on Build GRCh38
rs908540615 Chr.7: 44829788-44829789 on Build GRCh38
rs541113909 Chr.7: 44829925-44829925 on Build GRCh38
rs759878892 Chr.7: 44830182-44830182 on Build GRCh38
rs989050619 Chr.7: 44829936-44829936 on Build GRCh38
rs1043078324 Chr.7: 44829998-44829998 on Build GRCh38
rs1008189012 Chr.7: 44829744-44829744 on Build GRCh38
rs556597713 Chr.7: 44829743-44829743 on Build GRCh38
rs900604595 Chr.7: 44829732-44829732 on Build GRCh38
rs938842896 Chr.7: 44829930-44829930 on Build GRCh38
rs779883916 Chr.7: 44830093-44830093 on Build GRCh38
rs932087769 Chr.7: 44830043-44830043 on Build GRCh38
rs770543813 Chr.7: 44829970-44829970 on Build GRCh38
rs973783509 Chr.7: 44829786-44829786 on Build GRCh38
rs184720090 Chr.7: 44829751-44829751 on Build GRCh38
rs1028165308 Chr.7: 44829742-44829742 on Build GRCh38
rs770987861 Chr.7: 44830159-44830159 on Build GRCh38
rs564257571 Chr.7: 44829767-44829767 on Build GRCh38
rs1050197432 Chr.7: 44829879-44829879 on Build GRCh38
rs371353341 Chr.7: 44830116-44830116 on Build GRCh38
rs900023802 Chr.7: 44829841-44829841 on Build GRCh38
rs548718869 Chr.7: 44830125-44830125 on Build GRCh38
rs1026654463 Chr.7: 44830165-44830165 on Build GRCh38
rs111898421 Chr.7: 44829806-44829806 on Build GRCh38
rs1047055185 Chr.7: 44829777-44829777 on Build GRCh38
rs941628622 Chr.7: 44829793-44829793 on Build GRCh38
rs1043559146 Chr.7: 44829721-44829721 on Build GRCh38
rs375966093 Chr.7: 44830008-44830008 on Build GRCh38
rs969841289 Chr.7: 44829875-44829875 on Build GRCh38
rs766172068 Chr.7: 44830110-44830111 on Build GRCh38
rs189755353 Chr.7: 44829756-44829756 on Build GRCh38
rs367976264 Chr.7: 44830170-44830170 on Build GRCh38
rs746744602 Chr.7: 44830104-44830104 on Build GRCh38
rs371854685 Chr.7: 44830181-44830181 on Build GRCh38
rs890999597 Chr.7: 44830080-44830080 on Build GRCh38
rs1297837 Chr.7: 44829938-44829938 on Build GRCh38
rs890169865 Chr.7: 44829912-44829912 on Build GRCh38
rs775802182 Chr.7: 44829741-44829741 on Build GRCh38
rs767934169 Chr.7: 44830183-44830183 on Build GRCh38
rs184455428 Chr.7: 44830130-44830130 on Build GRCh38
rs1035103440 Chr.7: 44830161-44830164 on Build GRCh38
rs556915152 Chr.7: 44829948-44829952 on Build GRCh38
rs773120293 Chr.7: 44830132-44830132 on Build GRCh38
rs1051847673 Chr.7: 44830055-44830055 on Build GRCh38
rs368913178 Chr.7: 44830115-44830115 on Build GRCh38
rs780859168 Chr.7: 44830007-44830007 on Build GRCh38
rs980218012 Chr.7: 44829920-44829920 on Build GRCh38

View Full Product Details