Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44840821-44841335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H2AFV
Entrez Gene ID
94239
Gene Name
H2A histone family member V
Gene Aliases
H2A.Z-2, H2AV
UniGene
Hs.488189
Gene Chromosome Location
Chr.7: 44826889-44848126 on Build GRCh38
Transcripts
NM_012412.4 NM_138635.3 NM_201436.2 NM_201516.2 NM_201517.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7428963 c.87_88insT p.V30fs*43 Chr.7: 44841006-44841007 on Build GRCh38
7126264 c.134G>A p.G45E Chr.7: 44840960-44840960 on Build GRCh38
4587427 c.189T>C p.T63T Chr.7: 44840905-44840905 on Build GRCh38
1755297 c.171G>A p.A57A Chr.7: 44840923-44840923 on Build GRCh38
5937305 c.100C>T p.H34Y Chr.7: 44840994-44840994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765101006 Chr.7: 44840953-44840953 on Build GRCh38
rs947135621 Chr.7: 44840970-44840970 on Build GRCh38
rs146961116 Chr.7: 44840935-44840935 on Build GRCh38
rs763673952 Chr.7: 44840914-44840914 on Build GRCh38
rs146182342 Chr.7: 44841249-44841249 on Build GRCh38
rs186913857 Chr.7: 44841082-44841082 on Build GRCh38
rs747295881 Chr.7: 44840869-44840869 on Build GRCh38
rs915399278 Chr.7: 44840932-44840932 on Build GRCh38
rs946952557 Chr.7: 44841272-44841272 on Build GRCh38
rs760421407 Chr.7: 44840905-44840905 on Build GRCh38
rs1016526915 Chr.7: 44841117-44841118 on Build GRCh38
rs987507659 Chr.7: 44841185-44841185 on Build GRCh38
rs376117761 Chr.7: 44840848-44840848 on Build GRCh38
rs372942888 Chr.7: 44841110-44841111 on Build GRCh38
rs576905736 Chr.7: 44841147-44841147 on Build GRCh38
rs777006443 Chr.7: 44840880-44840880 on Build GRCh38
rs1029045197 Chr.7: 44841148-44841148 on Build GRCh38
rs6966546 Chr.7: 44841244-44841244 on Build GRCh38
rs138616116 Chr.7: 44841086-44841086 on Build GRCh38
rs780135530 Chr.7: 44840856-44840856 on Build GRCh38
rs79417609 Chr.7: 44841144-44841144 on Build GRCh38
rs748469781 Chr.7: 44841020-44841020 on Build GRCh38
rs138752128 Chr.7: 44840851-44840851 on Build GRCh38
rs758442986 Chr.7: 44840977-44840977 on Build GRCh38
rs982249731 Chr.7: 44841187-44841187 on Build GRCh38
rs894064475 Chr.7: 44841258-44841258 on Build GRCh38
rs1030989589 Chr.7: 44841225-44841225 on Build GRCh38
rs749849564 Chr.7: 44841055-44841055 on Build GRCh38
rs747852308 Chr.7: 44840870-44840870 on Build GRCh38
rs781279463 Chr.7: 44841010-44841010 on Build GRCh38
rs759099787 Chr.7: 44840894-44840894 on Build GRCh38
rs771816821 Chr.7: 44841221-44841221 on Build GRCh38
rs998676138 Chr.7: 44840938-44840938 on Build GRCh38
rs770344684 Chr.7: 44841049-44841049 on Build GRCh38
rs773872384 Chr.7: 44840891-44840891 on Build GRCh38
rs753589092 Chr.7: 44840923-44840923 on Build GRCh38
rs544076727 Chr.7: 44841071-44841071 on Build GRCh38
rs778416883 Chr.7: 44841051-44841051 on Build GRCh38
rs761600116 Chr.7: 44840937-44840937 on Build GRCh38
rs771457854 Chr.7: 44841056-44841056 on Build GRCh38
rs750427588 Chr.7: 44840957-44840957 on Build GRCh38
rs780231707 Chr.7: 44840980-44840980 on Build GRCh38
rs768882229 Chr.7: 44840875-44840875 on Build GRCh38
rs141883069 Chr.7: 44841060-44841060 on Build GRCh38
rs770361196 Chr.7: 44840889-44840889 on Build GRCh38
rs886383296 Chr.7: 44841084-44841084 on Build GRCh38
rs954769396 Chr.7: 44841171-44841171 on Build GRCh38
rs758093468 Chr.7: 44841015-44841015 on Build GRCh38
rs963227600 Chr.7: 44841145-44841145 on Build GRCh38
rs973101044 Chr.7: 44841156-44841156 on Build GRCh38
rs774971540 Chr.7: 44840902-44840902 on Build GRCh38
rs748557616 Chr.7: 44840888-44840888 on Build GRCh38
rs755190543 Chr.7: 44841007-44841007 on Build GRCh38
rs867152303 Chr.7: 44840999-44840999 on Build GRCh38
rs778373104 Chr.7: 44841157-44841157 on Build GRCh38
rs751616035 Chr.7: 44840992-44840992 on Build GRCh38
rs915411252 Chr.7: 44841263-44841263 on Build GRCh38
rs963220237 Chr.7: 44841140-44841140 on Build GRCh38
rs947867758 Chr.7: 44841170-44841170 on Build GRCh38
rs771675197 Chr.7: 44840896-44840896 on Build GRCh38
rs954534498 Chr.7: 44841057-44841057 on Build GRCh38

View Full Product Details