Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107473867-107474388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG5
Entrez Gene ID
10466
Gene Name
component of oligomeric golgi complex 5
Gene Aliases
CDG2I, GOLTC1, GTC90
UniGene
Hs.239631
Gene Chromosome Location
Chr.7: 107201743-107564514 on Build GRCh38
Transcripts
NM_001161520.1 NM_006348.3 NM_181733.2 NM_005295.2 XM_011516056.2

Gene Symbol
GPR22
Entrez Gene ID
2845
Gene Name
G protein-coupled receptor 22
Gene Aliases
-
UniGene
Hs.657277
Gene Chromosome Location
Chr.7: 107470057-107476207 on Build GRCh38
Transcripts
NM_001161520.1 NM_006348.3 NM_181733.2 NM_005295.2 XM_011516056.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
275352 c.74A>G p.D25G Chr.7: 107474134-107474134 on Build GRCh38
4688127 c.228C>T p.V76V Chr.7: 107474288-107474288 on Build GRCh38
6108158 c.98C>A p.P33Q Chr.7: 107474158-107474158 on Build GRCh38
4688126 c.168T>C p.L56L Chr.7: 107474228-107474228 on Build GRCh38
5006849 c.92A>C p.Y31S Chr.7: 107474152-107474152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750018947 Chr.7: 107474018-107474019 on Build GRCh38
rs377116061 Chr.7: 107474049-107474049 on Build GRCh38
rs775834757 Chr.7: 107474118-107474118 on Build GRCh38
rs1014823136 Chr.7: 107473935-107473935 on Build GRCh38
rs544275781 Chr.7: 107473978-107473978 on Build GRCh38
rs372074309 Chr.7: 107474251-107474251 on Build GRCh38
rs386410944 Chr.7: 107473957-107473958 on Build GRCh38
rs766064928 Chr.7: 107474205-107474205 on Build GRCh38
rs149758675 Chr.7: 107474315-107474315 on Build GRCh38
rs754550098 Chr.7: 107474247-107474247 on Build GRCh38
rs934093136 Chr.7: 107474206-107474206 on Build GRCh38
rs746268020 Chr.7: 107474295-107474295 on Build GRCh38
rs762775120 Chr.7: 107474196-107474196 on Build GRCh38
rs1005182852 Chr.7: 107473934-107473934 on Build GRCh38
rs1038617410 Chr.7: 107473926-107473926 on Build GRCh38
rs189832337 Chr.7: 107473984-107473984 on Build GRCh38
rs772463518 Chr.7: 107474116-107474116 on Build GRCh38
rs747163901 Chr.7: 107474308-107474308 on Build GRCh38
rs148017336 Chr.7: 107474119-107474119 on Build GRCh38
rs1049835046 Chr.7: 107474216-107474216 on Build GRCh38
rs575397301 Chr.7: 107473898-107473898 on Build GRCh38
rs751105744 Chr.7: 107474015-107474015 on Build GRCh38
rs373919718 Chr.7: 107474011-107474011 on Build GRCh38
rs375104063 Chr.7: 107473963-107473964 on Build GRCh38
rs921607179 Chr.7: 107474147-107474147 on Build GRCh38
rs913478748 Chr.7: 107474104-107474104 on Build GRCh38
rs899106631 Chr.7: 107473945-107473945 on Build GRCh38
rs373788840 Chr.7: 107474294-107474294 on Build GRCh38
rs368142557 Chr.7: 107474220-107474220 on Build GRCh38
rs772840560 Chr.7: 107474195-107474195 on Build GRCh38
rs754392912 Chr.7: 107474282-107474282 on Build GRCh38
rs141612463 Chr.7: 107474143-107474143 on Build GRCh38
rs765099946 Chr.7: 107474144-107474144 on Build GRCh38
rs778309420 Chr.7: 107474046-107474046 on Build GRCh38
rs184159432 Chr.7: 107474037-107474037 on Build GRCh38
rs755887490 Chr.7: 107474036-107474037 on Build GRCh38
rs746134385 Chr.7: 107474108-107474108 on Build GRCh38
rs188712129 Chr.7: 107474128-107474128 on Build GRCh38
rs780493978 Chr.7: 107474305-107474305 on Build GRCh38
rs1006253041 Chr.7: 107473919-107473919 on Build GRCh38
rs757666894 Chr.7: 107474289-107474289 on Build GRCh38
rs779100586 Chr.7: 107474095-107474095 on Build GRCh38
rs980170234 Chr.7: 107473997-107473997 on Build GRCh38
rs931689670 Chr.7: 107474170-107474170 on Build GRCh38
rs995459221 Chr.7: 107473948-107473948 on Build GRCh38
rs771329646 Chr.7: 107474060-107474060 on Build GRCh38
rs960216716 Chr.7: 107474036-107474036 on Build GRCh38
rs776637482 Chr.7: 107474129-107474129 on Build GRCh38
rs947668702 Chr.7: 107474139-107474139 on Build GRCh38
rs1026981770 Chr.7: 107473977-107473977 on Build GRCh38
rs11453718 Chr.7: 107473955-107473956 on Build GRCh38
rs1023755655 Chr.7: 107473989-107473989 on Build GRCh38
rs923887336 Chr.7: 107474020-107474020 on Build GRCh38
rs890307430 Chr.7: 107473900-107473900 on Build GRCh38
rs776522088 Chr.7: 107474330-107474330 on Build GRCh38
rs372858257 Chr.7: 107473962-107473963 on Build GRCh38

View Full Product Details