Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 113082769-113083282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR85
Entrez Gene ID
54329
Gene Name
G protein-coupled receptor 85
Gene Aliases
SREB, SREB2
UniGene
Hs.152009
Gene Chromosome Location
Chr.7: 113080413-113087778 on Build GRCh38
Transcripts
NM_001146265.1 NM_001146266.1 NM_001146267.1 NM_018970.6 XM_005250451.2 XM_017012352.1

SNP Detail

SNP ID Chromosome Location
rs115954720 Chr.7: 113082795-113082795 on Build GRCh38
rs528915537 Chr.7: 113082881-113082881 on Build GRCh38
rs565853282 Chr.7: 113083239-113083243 on Build GRCh38
rs547081783 Chr.7: 113082888-113082888 on Build GRCh38
rs1037814597 Chr.7: 113083075-113083075 on Build GRCh38
rs1581688 Chr.7: 113083038-113083038 on Build GRCh38
rs1027255142 Chr.7: 113083043-113083043 on Build GRCh38
rs905439551 Chr.7: 113083033-113083033 on Build GRCh38
rs559230372 Chr.7: 113082914-113082914 on Build GRCh38
rs931904621 Chr.7: 113083131-113083131 on Build GRCh38
rs3800571 Chr.7: 113082905-113082905 on Build GRCh38
rs571676735 Chr.7: 113083142-113083142 on Build GRCh38
rs1014931379 Chr.7: 113083206-113083206 on Build GRCh38
rs895464813 Chr.7: 113082826-113082826 on Build GRCh38
rs191219710 Chr.7: 113083066-113083066 on Build GRCh38
rs931931258 Chr.7: 113082876-113082876 on Build GRCh38
rs912671319 Chr.7: 113082798-113082798 on Build GRCh38
rs979288568 Chr.7: 113083137-113083137 on Build GRCh38
rs533550812 Chr.7: 113082930-113082930 on Build GRCh38
rs542978952 Chr.7: 113082800-113082800 on Build GRCh38
rs925220517 Chr.7: 113083171-113083171 on Build GRCh38
rs1040619318 Chr.7: 113082831-113082831 on Build GRCh38
rs934958097 Chr.7: 113083192-113083192 on Build GRCh38
rs922583252 Chr.7: 113082986-113082986 on Build GRCh38
rs1052109542 Chr.7: 113082938-113082938 on Build GRCh38
rs1032786808 Chr.7: 113082946-113082946 on Build GRCh38
rs987455052 Chr.7: 113083070-113083070 on Build GRCh38
rs921864100 Chr.7: 113082853-113082853 on Build GRCh38
rs568247372 Chr.7: 113082979-113082979 on Build GRCh38
rs1024255838 Chr.7: 113082996-113082996 on Build GRCh38
rs553427239 Chr.7: 113083111-113083111 on Build GRCh38
rs944517739 Chr.7: 113082825-113082825 on Build GRCh38
rs535057810 Chr.7: 113083073-113083073 on Build GRCh38
rs896033034 Chr.7: 113083186-113083187 on Build GRCh38
rs78016094 Chr.7: 113082970-113082970 on Build GRCh38
rs149873045 Chr.7: 113083072-113083072 on Build GRCh38
rs982785662 Chr.7: 113083069-113083069 on Build GRCh38
rs368191558 Chr.7: 113082820-113082820 on Build GRCh38
rs561351139 Chr.7: 113082850-113082850 on Build GRCh38
rs975938992 Chr.7: 113083110-113083110 on Build GRCh38
rs34157104 Chr.7: 113083110-113083111 on Build GRCh38
rs937560180 Chr.7: 113083141-113083141 on Build GRCh38
rs1052056177 Chr.7: 113083225-113083225 on Build GRCh38

View Full Product Details