Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 155298473-155298966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSIG1
Entrez Gene ID
3638
Gene Name
insulin induced gene 1
Gene Aliases
CL-6, CL6
UniGene
Hs.520819
Gene Chromosome Location
Chr.7: 155297776-155310235 on Build GRCh38
Transcripts
NM_005542.4 NM_198336.2 NM_198337.2 XM_005249542.1 XM_005249543.1 XM_017012177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6414793 c.371C>A p.S124Y Chr.7: 155298656-155298656 on Build GRCh38
4826872 c.267C>G p.L89L Chr.7: 155298552-155298552 on Build GRCh38
4762974 c.332T>C p.V111A Chr.7: 155298617-155298617 on Build GRCh38
4938960 c.262A>G p.S88G Chr.7: 155298547-155298547 on Build GRCh38
400290 c.355A>T p.I119F Chr.7: 155298640-155298640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896145760 Chr.7: 155298686-155298686 on Build GRCh38
rs531172627 Chr.7: 155298847-155298847 on Build GRCh38
rs925490014 Chr.7: 155298589-155298589 on Build GRCh38
rs912552147 Chr.7: 155298879-155298879 on Build GRCh38
rs771528311 Chr.7: 155298582-155298582 on Build GRCh38
rs765559868 Chr.7: 155298643-155298643 on Build GRCh38
rs746215366 Chr.7: 155298593-155298593 on Build GRCh38
rs1044008758 Chr.7: 155298557-155298557 on Build GRCh38
rs979574974 Chr.7: 155298830-155298830 on Build GRCh38
rs761769847 Chr.7: 155298637-155298637 on Build GRCh38
rs559401493 Chr.7: 155298740-155298740 on Build GRCh38
rs777837279 Chr.7: 155298576-155298576 on Build GRCh38
rs377426409 Chr.7: 155298717-155298717 on Build GRCh38
rs371087646 Chr.7: 155298521-155298521 on Build GRCh38
rs371131835 Chr.7: 155298724-155298724 on Build GRCh38
rs1048690617 Chr.7: 155298940-155298940 on Build GRCh38
rs753458387 Chr.7: 155298764-155298764 on Build GRCh38
rs904926401 Chr.7: 155298926-155298926 on Build GRCh38
rs373426066 Chr.7: 155298708-155298708 on Build GRCh38
rs1001631094 Chr.7: 155298722-155298722 on Build GRCh38
rs1129826 Chr.7: 155298580-155298580 on Build GRCh38
rs949069807 Chr.7: 155298890-155298890 on Build GRCh38
rs1020735659 Chr.7: 155298761-155298761 on Build GRCh38
rs764160755 Chr.7: 155298512-155298512 on Build GRCh38
rs755231844 Chr.7: 155298533-155298533 on Build GRCh38
rs779083874 Chr.7: 155298535-155298535 on Build GRCh38
rs1026876707 Chr.7: 155298763-155298763 on Build GRCh38
rs760447154 Chr.7: 155298642-155298642 on Build GRCh38
rs942522263 Chr.7: 155298843-155298843 on Build GRCh38
rs147110873 Chr.7: 155298675-155298675 on Build GRCh38
rs781762054 Chr.7: 155298710-155298710 on Build GRCh38
rs758401252 Chr.7: 155298552-155298552 on Build GRCh38
rs529612635 Chr.7: 155298777-155298777 on Build GRCh38
rs754766497 Chr.7: 155298768-155298768 on Build GRCh38
rs915629114 Chr.7: 155298813-155298813 on Build GRCh38
rs1035743213 Chr.7: 155298770-155298770 on Build GRCh38
rs773774481 Chr.7: 155298748-155298748 on Build GRCh38
rs199932909 Chr.7: 155298529-155298529 on Build GRCh38
rs767408850 Chr.7: 155298639-155298639 on Build GRCh38
rs948317491 Chr.7: 155298540-155298540 on Build GRCh38
rs78105975 Chr.7: 155298852-155298852 on Build GRCh38
rs772056331 Chr.7: 155298853-155298853 on Build GRCh38
rs957814679 Chr.7: 155298726-155298726 on Build GRCh38
rs202184874 Chr.7: 155298607-155298607 on Build GRCh38
rs911295816 Chr.7: 155298822-155298822 on Build GRCh38
rs762601697 Chr.7: 155298620-155298620 on Build GRCh38
rs752876373 Chr.7: 155298544-155298544 on Build GRCh38
rs57346078 Chr.7: 155298932-155298932 on Build GRCh38
rs899903475 Chr.7: 155298902-155298902 on Build GRCh38
rs373597330 Chr.7: 155298538-155298538 on Build GRCh38
rs1039772250 Chr.7: 155298901-155298901 on Build GRCh38
rs898393358 Chr.7: 155298636-155298636 on Build GRCh38
rs1046975019 Chr.7: 155298864-155298864 on Build GRCh38
rs143931518 Chr.7: 155298649-155298649 on Build GRCh38
rs376499291 Chr.7: 155298504-155298504 on Build GRCh38
rs751663546 Chr.7: 155298690-155298690 on Build GRCh38
rs147727463 Chr.7: 155298791-155298791 on Build GRCh38
rs762357555 Chr.7: 155298509-155298509 on Build GRCh38
rs780408890 Chr.7: 155298733-155298733 on Build GRCh38
rs758584360 Chr.7: 155298655-155298655 on Build GRCh38
rs781246212 Chr.7: 155298886-155298886 on Build GRCh38
rs751456328 Chr.7: 155298513-155298513 on Build GRCh38
rs934032847 Chr.7: 155298522-155298522 on Build GRCh38
rs747480310 Chr.7: 155298579-155298579 on Build GRCh38
rs959853983 Chr.7: 155298788-155298788 on Build GRCh38
rs748056088 Chr.7: 155298823-155298823 on Build GRCh38
rs750266191 Chr.7: 155298640-155298640 on Build GRCh38
rs115472901 Chr.7: 155298896-155298896 on Build GRCh38
rs1023325021 Chr.7: 155298711-155298711 on Build GRCh38
rs775118067 Chr.7: 155298506-155298506 on Build GRCh38
rs773861240 Chr.7: 155298633-155298633 on Build GRCh38
rs759297214 Chr.7: 155298496-155298496 on Build GRCh38
rs761681762 Chr.7: 155298517-155298517 on Build GRCh38
rs138010301 Chr.7: 155298585-155298585 on Build GRCh38
rs570246769 Chr.7: 155298820-155298820 on Build GRCh38
rs779702577 Chr.7: 155298718-155298718 on Build GRCh38
rs911097384 Chr.7: 155298592-155298592 on Build GRCh38
rs987313390 Chr.7: 155298816-155298816 on Build GRCh38
rs769631428 Chr.7: 155298612-155298612 on Build GRCh38
rs200641765 Chr.7: 155298617-155298617 on Build GRCh38
rs748916911 Chr.7: 155298736-155298736 on Build GRCh38
rs149516023 Chr.7: 155298621-155298621 on Build GRCh38
rs896165801 Chr.7: 155298866-155298866 on Build GRCh38

View Full Product Details