Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 155302162-155302661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSIG1
Entrez Gene ID
3638
Gene Name
insulin induced gene 1
Gene Aliases
CL-6, CL6
UniGene
Hs.520819
Gene Chromosome Location
Chr.7: 155297776-155310235 on Build GRCh38
Transcripts
NM_005542.4 NM_198336.2 NM_198337.2 XM_005249542.1 XM_005249543.1 XM_017012177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5031693 c.588A>G p.L196L Chr.7: 155302301-155302301 on Build GRCh38
4926681 c.596G>A p.G199D Chr.7: 155302309-155302309 on Build GRCh38
5824303 c.561C>A p.V187V Chr.7: 155302274-155302274 on Build GRCh38
6846620 c.661G>C p.A221P Chr.7: 155302374-155302374 on Build GRCh38
3879729 c.665C>T p.T222M Chr.7: 155302378-155302378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750612055 Chr.7: 155302226-155302227 on Build GRCh38
rs116500782 Chr.7: 155302414-155302414 on Build GRCh38
rs192867646 Chr.7: 155302555-155302555 on Build GRCh38
rs76899936 Chr.7: 155302196-155302196 on Build GRCh38
rs1031041811 Chr.7: 155302568-155302568 on Build GRCh38
rs567627468 Chr.7: 155302391-155302391 on Build GRCh38
rs757992959 Chr.7: 155302321-155302321 on Build GRCh38
rs539217399 Chr.7: 155302591-155302591 on Build GRCh38
rs752944998 Chr.7: 155302518-155302518 on Build GRCh38
rs777393215 Chr.7: 155302435-155302435 on Build GRCh38
rs746510175 Chr.7: 155302331-155302331 on Build GRCh38
rs562345271 Chr.7: 155302379-155302379 on Build GRCh38
rs1038040224 Chr.7: 155302264-155302264 on Build GRCh38
rs999941395 Chr.7: 155302563-155302563 on Build GRCh38
rs527701821 Chr.7: 155302329-155302329 on Build GRCh38
rs776953189 Chr.7: 155302524-155302525 on Build GRCh38
rs554334089 Chr.7: 155302629-155302629 on Build GRCh38
rs765610318 Chr.7: 155302411-155302411 on Build GRCh38
rs897710551 Chr.7: 155302308-155302308 on Build GRCh38
rs953352859 Chr.7: 155302614-155302617 on Build GRCh38
rs139463395 Chr.7: 155302397-155302397 on Build GRCh38
rs73485859 Chr.7: 155302215-155302215 on Build GRCh38
rs745728958 Chr.7: 155302361-155302361 on Build GRCh38
rs542985237 Chr.7: 155302195-155302195 on Build GRCh38
rs567118208 Chr.7: 155302536-155302536 on Build GRCh38
rs760796100 Chr.7: 155302394-155302394 on Build GRCh38
rs758099145 Chr.7: 155302426-155302426 on Build GRCh38
rs147365058 Chr.7: 155302388-155302388 on Build GRCh38
rs770836270 Chr.7: 155302332-155302332 on Build GRCh38
rs776137976 Chr.7: 155302582-155302582 on Build GRCh38
rs967644581 Chr.7: 155302526-155302526 on Build GRCh38
rs184422722 Chr.7: 155302581-155302581 on Build GRCh38
rs566604081 Chr.7: 155302310-155302310 on Build GRCh38
rs1022308664 Chr.7: 155302521-155302521 on Build GRCh38
rs550559593 Chr.7: 155302243-155302243 on Build GRCh38
rs986179610 Chr.7: 155302624-155302624 on Build GRCh38
rs901842084 Chr.7: 155302571-155302571 on Build GRCh38
rs371349471 Chr.7: 155302223-155302223 on Build GRCh38
rs750409882 Chr.7: 155302203-155302203 on Build GRCh38
rs370285334 Chr.7: 155302632-155302632 on Build GRCh38
rs779178640 Chr.7: 155302305-155302305 on Build GRCh38
rs142687202 Chr.7: 155302387-155302387 on Build GRCh38
rs776629161 Chr.7: 155302349-155302349 on Build GRCh38
rs756262816 Chr.7: 155302453-155302456 on Build GRCh38
rs9718441 Chr.7: 155302462-155302462 on Build GRCh38
rs767727993 Chr.7: 155302207-155302207 on Build GRCh38
rs971954481 Chr.7: 155302639-155302639 on Build GRCh38
rs200756620 Chr.7: 155302398-155302398 on Build GRCh38
rs201303039 Chr.7: 155302241-155302241 on Build GRCh38
rs754211515 Chr.7: 155302257-155302257 on Build GRCh38
rs769646258 Chr.7: 155302378-155302378 on Build GRCh38
rs538543655 Chr.7: 155302549-155302549 on Build GRCh38
rs997623607 Chr.7: 155302577-155302577 on Build GRCh38
rs759655828 Chr.7: 155302598-155302598 on Build GRCh38
rs747810129 Chr.7: 155302311-155302311 on Build GRCh38
rs774720517 Chr.7: 155302201-155302201 on Build GRCh38
rs780510811 Chr.7: 155302469-155302469 on Build GRCh38
rs548930206 Chr.7: 155302330-155302330 on Build GRCh38
rs542720116 Chr.7: 155302633-155302633 on Build GRCh38
rs372903186 Chr.7: 155302485-155302485 on Build GRCh38
rs566216322 Chr.7: 155302438-155302438 on Build GRCh38
rs755364368 Chr.7: 155302295-155302295 on Build GRCh38

View Full Product Details