Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 155302659-155303164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSIG1
Entrez Gene ID
3638
Gene Name
insulin induced gene 1
Gene Aliases
CL-6, CL6
UniGene
Hs.520819
Gene Chromosome Location
Chr.7: 155297776-155310235 on Build GRCh38
Transcripts
NM_005542.4 NM_198336.2 NM_198337.2 XM_005249542.1 XM_005249543.1 XM_017012177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6877258 c.775G>A p.V259M Chr.7: 155302817-155302817 on Build GRCh38
300701 c.746C>T p.P249L Chr.7: 155302788-155302788 on Build GRCh38
3950263 c.746C>G p.P249R Chr.7: 155302788-155302788 on Build GRCh38
1622621 c.734G>T p.R245L Chr.7: 155302776-155302776 on Build GRCh38
5788695 c.705-7G>C p.? Chr.7: 155302740-155302740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947190735 Chr.7: 155303058-155303058 on Build GRCh38
rs553933996 Chr.7: 155303062-155303062 on Build GRCh38
rs9768652 Chr.7: 155302882-155302882 on Build GRCh38
rs202139336 Chr.7: 155302739-155302739 on Build GRCh38
rs571945056 Chr.7: 155302995-155302995 on Build GRCh38
rs962169337 Chr.7: 155302911-155302911 on Build GRCh38
rs551141308 Chr.7: 155302711-155302711 on Build GRCh38
rs893572184 Chr.7: 155303017-155303017 on Build GRCh38
rs542655462 Chr.7: 155303073-155303073 on Build GRCh38
rs184003228 Chr.7: 155303041-155303041 on Build GRCh38
rs770355507 Chr.7: 155302895-155302895 on Build GRCh38
rs181066493 Chr.7: 155302742-155302742 on Build GRCh38
rs11772189 Chr.7: 155302883-155302883 on Build GRCh38
rs927843017 Chr.7: 155302955-155302955 on Build GRCh38
rs1052705713 Chr.7: 155303103-155303103 on Build GRCh38
rs780037209 Chr.7: 155302705-155302705 on Build GRCh38
rs986063608 Chr.7: 155302865-155302865 on Build GRCh38
rs749068682 Chr.7: 155302707-155302707 on Build GRCh38
rs1129830 Chr.7: 155302753-155302753 on Build GRCh38
rs763305791 Chr.7: 155302756-155302756 on Build GRCh38
rs1055303346 Chr.7: 155302727-155302727 on Build GRCh38
rs1035540304 Chr.7: 155302992-155302992 on Build GRCh38
rs764380311 Chr.7: 155302768-155302768 on Build GRCh38
rs935473373 Chr.7: 155303100-155303100 on Build GRCh38
rs954944187 Chr.7: 155303084-155303084 on Build GRCh38
rs967645802 Chr.7: 155302816-155302816 on Build GRCh38
rs935634979 Chr.7: 155302822-155302822 on Build GRCh38
rs376491438 Chr.7: 155302716-155302716 on Build GRCh38
rs566098260 Chr.7: 155303080-155303080 on Build GRCh38
rs572139059 Chr.7: 155303070-155303070 on Build GRCh38
rs756170771 Chr.7: 155302844-155302844 on Build GRCh38
rs112779865 Chr.7: 155302745-155302745 on Build GRCh38
rs536753604 Chr.7: 155303128-155303128 on Build GRCh38
rs752597940 Chr.7: 155302898-155302899 on Build GRCh38
rs768576009 Chr.7: 155302867-155302867 on Build GRCh38
rs760116811 Chr.7: 155302740-155302740 on Build GRCh38
rs780785188 Chr.7: 155302811-155302811 on Build GRCh38
rs771053389 Chr.7: 155302726-155302726 on Build GRCh38
rs576061560 Chr.7: 155302810-155302810 on Build GRCh38
rs112769976 Chr.7: 155302746-155302746 on Build GRCh38
rs997494265 Chr.7: 155302881-155302881 on Build GRCh38
rs551521799 Chr.7: 155302922-155302922 on Build GRCh38
rs769816473 Chr.7: 155302703-155302703 on Build GRCh38
rs1030551815 Chr.7: 155303076-155303076 on Build GRCh38
rs766442701 Chr.7: 155302854-155302874 on Build GRCh38
rs780125640 Chr.7: 155302856-155302856 on Build GRCh38
rs973681590 Chr.7: 155302887-155302887 on Build GRCh38
rs149813005 Chr.7: 155303136-155303136 on Build GRCh38
rs937674421 Chr.7: 155302972-155302972 on Build GRCh38
rs142549077 Chr.7: 155302931-155302931 on Build GRCh38
rs915651832 Chr.7: 155303038-155303038 on Build GRCh38
rs999057752 Chr.7: 155302818-155302818 on Build GRCh38
rs1030499235 Chr.7: 155302827-155302827 on Build GRCh38
rs1046349952 Chr.7: 155302937-155302937 on Build GRCh38
rs745938764 Chr.7: 155302890-155302890 on Build GRCh38
rs372798445 Chr.7: 155302709-155302709 on Build GRCh38
rs376290152 Chr.7: 155302775-155302775 on Build GRCh38
rs368706880 Chr.7: 155302696-155302696 on Build GRCh38
rs369310435 Chr.7: 155302863-155302863 on Build GRCh38
rs61740679 Chr.7: 155302843-155302843 on Build GRCh38
rs919089234 Chr.7: 155302901-155302901 on Build GRCh38

View Full Product Details