Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 155309627-155310138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSIG1
Entrez Gene ID
3638
Gene Name
insulin induced gene 1
Gene Aliases
CL-6, CL6
UniGene
Hs.520819
Gene Chromosome Location
Chr.7: 155297776-155310235 on Build GRCh38
Transcripts
NM_005542.4 NM_198336.2 NM_198337.2 XM_005249542.1 XM_005249543.1 XM_017012177.1

SNP Detail

SNP ID Chromosome Location
rs943380401 Chr.7: 155309813-155309813 on Build GRCh38
rs933783384 Chr.7: 155310059-155310059 on Build GRCh38
rs538562956 Chr.7: 155309801-155309801 on Build GRCh38
rs369450229 Chr.7: 155309957-155309957 on Build GRCh38
rs184818901 Chr.7: 155309692-155309692 on Build GRCh38
rs370300725 Chr.7: 155309849-155309849 on Build GRCh38
rs925626832 Chr.7: 155309845-155309845 on Build GRCh38
rs1003877180 Chr.7: 155309661-155309661 on Build GRCh38
rs925823254 Chr.7: 155310044-155310045 on Build GRCh38
rs373737802 Chr.7: 155309903-155309903 on Build GRCh38
rs988533795 Chr.7: 155310045-155310045 on Build GRCh38
rs1030727739 Chr.7: 155310002-155310002 on Build GRCh38
rs556833223 Chr.7: 155309956-155309956 on Build GRCh38
rs558995161 Chr.7: 155309982-155309982 on Build GRCh38
rs746086648 Chr.7: 155309960-155309960 on Build GRCh38
rs1047772980 Chr.7: 155309877-155309877 on Build GRCh38
rs575395550 Chr.7: 155309958-155309958 on Build GRCh38
rs567982788 Chr.7: 155309690-155309690 on Build GRCh38
rs541407485 Chr.7: 155310111-155310111 on Build GRCh38
rs184872948 Chr.7: 155310041-155310041 on Build GRCh38
rs1051259549 Chr.7: 155310099-155310099 on Build GRCh38
rs978260691 Chr.7: 155309987-155309987 on Build GRCh38
rs1017589324 Chr.7: 155309698-155309698 on Build GRCh38
rs955022205 Chr.7: 155309693-155309693 on Build GRCh38
rs924783929 Chr.7: 155309757-155309757 on Build GRCh38
rs536437290 Chr.7: 155309870-155309870 on Build GRCh38
rs1022027981 Chr.7: 155309733-155309733 on Build GRCh38
rs778260451 Chr.7: 155309790-155309791 on Build GRCh38
rs76948004 Chr.7: 155309823-155309823 on Build GRCh38
rs1034913518 Chr.7: 155309687-155309687 on Build GRCh38
rs988148140 Chr.7: 155309773-155309773 on Build GRCh38
rs191603973 Chr.7: 155309976-155309976 on Build GRCh38
rs867548336 Chr.7: 155310103-155310103 on Build GRCh38
rs912870595 Chr.7: 155310057-155310061 on Build GRCh38
rs537112609 Chr.7: 155309753-155309753 on Build GRCh38
rs935613973 Chr.7: 155309869-155309869 on Build GRCh38
rs3188220 Chr.7: 155309681-155309681 on Build GRCh38
rs891575049 Chr.7: 155309888-155309888 on Build GRCh38
rs935894906 Chr.7: 155310045-155310045 on Build GRCh38
rs948643180 Chr.7: 155309812-155309813 on Build GRCh38
rs775855362 Chr.7: 155309981-155309981 on Build GRCh38
rs1007745161 Chr.7: 155309696-155309696 on Build GRCh38
rs543497156 Chr.7: 155309978-155309978 on Build GRCh38
rs933730463 Chr.7: 155309896-155309896 on Build GRCh38
rs1052849138 Chr.7: 155309660-155309660 on Build GRCh38
rs76635730 Chr.7: 155309863-155309863 on Build GRCh38
rs934819370 Chr.7: 155309772-155309772 on Build GRCh38
rs558277656 Chr.7: 155310013-155310013 on Build GRCh38
rs1017789228 Chr.7: 155309966-155309966 on Build GRCh38
rs990426378 Chr.7: 155309714-155309714 on Build GRCh38
rs993304497 Chr.7: 155309988-155309988 on Build GRCh38
rs1009700915 Chr.7: 155309679-155309679 on Build GRCh38
rs140281130 Chr.7: 155309756-155309756 on Build GRCh38
rs188107182 Chr.7: 155309791-155309791 on Build GRCh38
rs963322055 Chr.7: 155309719-155309719 on Build GRCh38
rs554772328 Chr.7: 155309927-155309927 on Build GRCh38
rs899285706 Chr.7: 155309824-155309824 on Build GRCh38
rs749603261 Chr.7: 155310016-155310016 on Build GRCh38

View Full Product Details