Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 166917214-166917730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ILDR2
Entrez Gene ID
387597
Gene Name
immunoglobulin like domain containing receptor 2
Gene Aliases
C1orf32, dJ782G3.1
UniGene
Hs.444835
Gene Chromosome Location
Chr.1: 166913204-166975324 on Build GRCh38
Transcripts
NM_199351.2 XM_006711314.3 XM_017001252.1 XM_017001253.1 XM_017001254.1 XM_017001255.1 XM_017001256.1

SNP Detail

SNP ID Chromosome Location
rs374197564 Chr.1: 166917433-166917433 on Build GRCh38
rs1006646853 Chr.1: 166917658-166917658 on Build GRCh38
rs529661278 Chr.1: 166917405-166917405 on Build GRCh38
rs534877465 Chr.1: 166917564-166917564 on Build GRCh38
rs1042612195 Chr.1: 166917497-166917497 on Build GRCh38
rs756086679 Chr.1: 166917383-166917383 on Build GRCh38
rs1029725691 Chr.1: 166917285-166917285 on Build GRCh38
rs771985007 Chr.1: 166917406-166917406 on Build GRCh38
rs558440409 Chr.1: 166917262-166917262 on Build GRCh38
rs568437974 Chr.1: 166917671-166917671 on Build GRCh38
rs1023572237 Chr.1: 166917605-166917605 on Build GRCh38
rs899227685 Chr.1: 166917593-166917593 on Build GRCh38
rs982764159 Chr.1: 166917640-166917640 on Build GRCh38
rs552342580 Chr.1: 166917508-166917508 on Build GRCh38
rs1006615315 Chr.1: 166917305-166917305 on Build GRCh38
rs371340331 Chr.1: 166917340-166917340 on Build GRCh38
rs997901587 Chr.1: 166917279-166917279 on Build GRCh38
rs1016657064 Chr.1: 166917661-166917661 on Build GRCh38
rs147004327 Chr.1: 166917301-166917301 on Build GRCh38
rs889430409 Chr.1: 166917653-166917653 on Build GRCh38
rs748480407 Chr.1: 166917665-166917665 on Build GRCh38
rs938117352 Chr.1: 166917472-166917472 on Build GRCh38
rs971031257 Chr.1: 166917634-166917634 on Build GRCh38
rs115487676 Chr.1: 166917323-166917323 on Build GRCh38
rs777811026 Chr.1: 166917385-166917385 on Build GRCh38
rs1004677929 Chr.1: 166917548-166917548 on Build GRCh38
rs997373727 Chr.1: 166917242-166917242 on Build GRCh38
rs537728734 Chr.1: 166917384-166917384 on Build GRCh38
rs72707810 Chr.1: 166917707-166917707 on Build GRCh38
rs372032818 Chr.1: 166917606-166917606 on Build GRCh38
rs75005035 Chr.1: 166917541-166917541 on Build GRCh38
rs150119462 Chr.1: 166917487-166917487 on Build GRCh38
rs749267999 Chr.1: 166917392-166917392 on Build GRCh38
rs150940042 Chr.1: 166917479-166917479 on Build GRCh38
rs1051636821 Chr.1: 166917540-166917540 on Build GRCh38
rs879903525 Chr.1: 166917441-166917441 on Build GRCh38
rs182473356 Chr.1: 166917691-166917691 on Build GRCh38
rs141353502 Chr.1: 166917684-166917684 on Build GRCh38
rs1016308481 Chr.1: 166917567-166917567 on Build GRCh38
rs571579087 Chr.1: 166917646-166917646 on Build GRCh38
rs776953505 Chr.1: 166917698-166917698 on Build GRCh38
rs747188821 Chr.1: 166917655-166917655 on Build GRCh38
rs974069031 Chr.1: 166917332-166917332 on Build GRCh38
rs1017804330 Chr.1: 166917315-166917315 on Build GRCh38
rs989614265 Chr.1: 166917667-166917667 on Build GRCh38
rs367848112 Chr.1: 166917601-166917601 on Build GRCh38
rs902452792 Chr.1: 166917509-166917509 on Build GRCh38
rs140669690 Chr.1: 166917480-166917480 on Build GRCh38
rs924092323 Chr.1: 166917402-166917402 on Build GRCh38
rs145129301 Chr.1: 166917702-166917702 on Build GRCh38
rs914284906 Chr.1: 166917439-166917439 on Build GRCh38
rs1026599150 Chr.1: 166917348-166917348 on Build GRCh38
rs902421986 Chr.1: 166917278-166917278 on Build GRCh38
rs1012221338 Chr.1: 166917597-166917597 on Build GRCh38
rs553308454 Chr.1: 166917616-166917616 on Build GRCh38
rs1005367785 Chr.1: 166917295-166917295 on Build GRCh38

View Full Product Details