Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167053568-167054064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPA33
Entrez Gene ID
10223
Gene Name
glycoprotein A33
Gene Aliases
A33
UniGene
Hs.651244
Gene Chromosome Location
Chr.1: 167052836-167090631 on Build GRCh38
Transcripts
NM_005814.2 XM_017000005.1

SNP Detail

SNP ID Chromosome Location
rs35471899 Chr.1: 167053845-167053846 on Build GRCh38
rs1017701725 Chr.1: 167053732-167053732 on Build GRCh38
rs909441368 Chr.1: 167053771-167053771 on Build GRCh38
rs12076250 Chr.1: 167053683-167053683 on Build GRCh38
rs1033582521 Chr.1: 167054020-167054020 on Build GRCh38
rs192132959 Chr.1: 167053614-167053614 on Build GRCh38
rs931853971 Chr.1: 167053872-167053872 on Build GRCh38
rs149110409 Chr.1: 167053765-167053765 on Build GRCh38
rs55881654 Chr.1: 167053912-167053912 on Build GRCh38
rs976156140 Chr.1: 167053851-167053851 on Build GRCh38
rs975809752 Chr.1: 167053749-167053749 on Build GRCh38
rs1011916396 Chr.1: 167053942-167053942 on Build GRCh38
rs988839416 Chr.1: 167053801-167053801 on Build GRCh38
rs1030068837 Chr.1: 167053965-167053965 on Build GRCh38
rs138642494 Chr.1: 167054005-167054005 on Build GRCh38
rs989809468 Chr.1: 167053725-167053725 on Build GRCh38
rs575655173 Chr.1: 167053880-167053880 on Build GRCh38
rs546752990 Chr.1: 167053978-167053978 on Build GRCh38
rs545411681 Chr.1: 167053816-167053816 on Build GRCh38
rs189874689 Chr.1: 167054037-167054037 on Build GRCh38
rs1046550887 Chr.1: 167053815-167053815 on Build GRCh38
rs950487574 Chr.1: 167053976-167053976 on Build GRCh38
rs957666750 Chr.1: 167054021-167054021 on Build GRCh38
rs145794171 Chr.1: 167053866-167053866 on Build GRCh38
rs1055097595 Chr.1: 167053794-167053794 on Build GRCh38
rs528566247 Chr.1: 167053946-167053946 on Build GRCh38
rs546171516 Chr.1: 167053892-167053892 on Build GRCh38
rs1031620493 Chr.1: 167053698-167053698 on Build GRCh38
rs542052539 Chr.1: 167053793-167053793 on Build GRCh38
rs997215361 Chr.1: 167053956-167053956 on Build GRCh38
rs1014533433 Chr.1: 167053602-167053602 on Build GRCh38
rs944809932 Chr.1: 167053844-167053844 on Build GRCh38
rs964459275 Chr.1: 167053735-167053735 on Build GRCh38
rs940596440 Chr.1: 167053897-167053897 on Build GRCh38
rs896265833 Chr.1: 167053675-167053675 on Build GRCh38
rs907074794 Chr.1: 167053637-167053637 on Build GRCh38
rs574648434 Chr.1: 167053783-167053783 on Build GRCh38
rs893025006 Chr.1: 167053927-167053927 on Build GRCh38
rs1017234155 Chr.1: 167053943-167053943 on Build GRCh38
rs887732321 Chr.1: 167053884-167053884 on Build GRCh38
rs1024462434 Chr.1: 167053719-167053719 on Build GRCh38
rs200911711 Chr.1: 167053845-167053845 on Build GRCh38
rs998713313 Chr.1: 167053911-167053911 on Build GRCh38
rs536604410 Chr.1: 167053625-167053625 on Build GRCh38
rs916576927 Chr.1: 167054034-167054034 on Build GRCh38
rs376733018 Chr.1: 167053977-167053977 on Build GRCh38
rs923440761 Chr.1: 167053754-167053754 on Build GRCh38
rs1001722410 Chr.1: 167054018-167054018 on Build GRCh38
rs41270692 Chr.1: 167053770-167053770 on Build GRCh38
rs576855919 Chr.1: 167053705-167053705 on Build GRCh38
rs747496437 Chr.1: 167054019-167054019 on Build GRCh38
rs772675633 Chr.1: 167053994-167053994 on Build GRCh38
rs949445599 Chr.1: 167053804-167053804 on Build GRCh38

View Full Product Details