Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167054279-167054783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPA33
Entrez Gene ID
10223
Gene Name
glycoprotein A33
Gene Aliases
A33
UniGene
Hs.651244
Gene Chromosome Location
Chr.1: 167052836-167090631 on Build GRCh38
Transcripts
NM_005814.2 XM_017000005.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
899012 c.934G>T p.E312* Chr.1: 167054360-167054360 on Build GRCh38
1733535 c.931C>T p.R311C Chr.1: 167054363-167054363 on Build GRCh38
5719393 c.930G>A p.G310G Chr.1: 167054364-167054364 on Build GRCh38
3980019 c.940C>G p.P314A Chr.1: 167054354-167054354 on Build GRCh38
6122381 c.905G>T p.R302M Chr.1: 167054389-167054389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776512239 Chr.1: 167054457-167054457 on Build GRCh38
rs974945753 Chr.1: 167054595-167054595 on Build GRCh38
rs373122976 Chr.1: 167054400-167054400 on Build GRCh38
rs775602869 Chr.1: 167054389-167054389 on Build GRCh38
rs1029392225 Chr.1: 167054328-167054328 on Build GRCh38
rs898882629 Chr.1: 167054716-167054716 on Build GRCh38
rs751492026 Chr.1: 167054622-167054622 on Build GRCh38
rs1016781258 Chr.1: 167054659-167054659 on Build GRCh38
rs552781090 Chr.1: 167054745-167054745 on Build GRCh38
rs188063662 Chr.1: 167054758-167054758 on Build GRCh38
rs146949681 Chr.1: 167054319-167054319 on Build GRCh38
rs746964648 Chr.1: 167054398-167054400 on Build GRCh38
rs1023769744 Chr.1: 167054736-167054736 on Build GRCh38
rs953529951 Chr.1: 167054333-167054333 on Build GRCh38
rs771418121 Chr.1: 167054365-167054365 on Build GRCh38
rs998177095 Chr.1: 167054637-167054637 on Build GRCh38
rs111372554 Chr.1: 167054441-167054441 on Build GRCh38
rs770285244 Chr.1: 167054362-167054362 on Build GRCh38
rs936334976 Chr.1: 167054722-167054722 on Build GRCh38
rs778599635 Chr.1: 167054346-167054346 on Build GRCh38
rs1012007600 Chr.1: 167054733-167054733 on Build GRCh38
rs534189955 Chr.1: 167054699-167054699 on Build GRCh38
rs750060636 Chr.1: 167054352-167054352 on Build GRCh38
rs368887906 Chr.1: 167054744-167054744 on Build GRCh38
rs779396234 Chr.1: 167054473-167054473 on Build GRCh38
rs766281753 Chr.1: 167054342-167054342 on Build GRCh38
rs546662754 Chr.1: 167054666-167054666 on Build GRCh38
rs533595357 Chr.1: 167054512-167054512 on Build GRCh38
rs148351388 Chr.1: 167054353-167054353 on Build GRCh38
rs767047993 Chr.1: 167054707-167054707 on Build GRCh38
rs971900557 Chr.1: 167054664-167054664 on Build GRCh38
rs779451400 Chr.1: 167054412-167054412 on Build GRCh38
rs548776013 Chr.1: 167054757-167054757 on Build GRCh38
rs146470813 Chr.1: 167054374-167054374 on Build GRCh38
rs771460866 Chr.1: 167054462-167054462 on Build GRCh38
rs1053951999 Chr.1: 167054561-167054561 on Build GRCh38
rs1010425298 Chr.1: 167054575-167054575 on Build GRCh38
rs753924459 Chr.1: 167054629-167054629 on Build GRCh38
rs186168290 Chr.1: 167054500-167054500 on Build GRCh38
rs762131091 Chr.1: 167054511-167054511 on Build GRCh38
rs1041897596 Chr.1: 167054582-167054582 on Build GRCh38
rs371904851 Chr.1: 167054655-167054655 on Build GRCh38
rs373226332 Chr.1: 167054474-167054474 on Build GRCh38
rs551752831 Chr.1: 167054523-167054523 on Build GRCh38
rs372035414 Chr.1: 167054315-167054315 on Build GRCh38
rs921768829 Chr.1: 167054646-167054646 on Build GRCh38
rs868248660 Chr.1: 167054643-167054643 on Build GRCh38
rs561800466 Chr.1: 167054398-167054398 on Build GRCh38
rs962074435 Chr.1: 167054660-167054660 on Build GRCh38
rs767693291 Chr.1: 167054324-167054324 on Build GRCh38
rs540631759 Chr.1: 167054367-167054367 on Build GRCh38
rs746112648 Chr.1: 167054477-167054477 on Build GRCh38
rs970657034 Chr.1: 167054706-167054706 on Build GRCh38
rs768618489 Chr.1: 167054503-167054503 on Build GRCh38
rs753711114 Chr.1: 167054399-167054399 on Build GRCh38
rs886142733 Chr.1: 167054406-167054406 on Build GRCh38
rs902121952 Chr.1: 167054627-167054627 on Build GRCh38
rs989569104 Chr.1: 167054563-167054563 on Build GRCh38
rs760555302 Chr.1: 167054393-167054393 on Build GRCh38
rs889124409 Chr.1: 167054645-167054645 on Build GRCh38
rs777178171 Chr.1: 167054302-167054302 on Build GRCh38
rs758004986 Chr.1: 167054409-167054409 on Build GRCh38
rs764166750 Chr.1: 167054489-167054489 on Build GRCh38
rs759757858 Chr.1: 167054323-167054323 on Build GRCh38
rs565884581 Chr.1: 167054318-167054318 on Build GRCh38
rs768631567 Chr.1: 167054438-167054451 on Build GRCh38
rs990013431 Chr.1: 167054339-167054339 on Build GRCh38
rs748632848 Chr.1: 167054360-167054360 on Build GRCh38
rs377508532 Chr.1: 167054566-167054566 on Build GRCh38
rs751023332 Chr.1: 167054433-167054433 on Build GRCh38
rs956727575 Chr.1: 167054535-167054535 on Build GRCh38
rs3738211 Chr.1: 167054487-167054487 on Build GRCh38
rs772321239 Chr.1: 167054375-167054375 on Build GRCh38
rs375309413 Chr.1: 167054408-167054408 on Build GRCh38
rs748664530 Chr.1: 167054310-167054310 on Build GRCh38
rs545808958 Chr.1: 167054334-167054334 on Build GRCh38
rs778412255 Chr.1: 167054461-167054461 on Build GRCh38
rs375619086 Chr.1: 167054459-167054459 on Build GRCh38
rs369408073 Chr.1: 167054451-167054451 on Build GRCh38
rs35152579 Chr.1: 167054343-167054343 on Build GRCh38
rs144624914 Chr.1: 167054443-167054443 on Build GRCh38
rs763105903 Chr.1: 167054314-167054314 on Build GRCh38
rs773584975 Chr.1: 167054312-167054312 on Build GRCh38
rs1051666495 Chr.1: 167054698-167054698 on Build GRCh38
rs926523356 Chr.1: 167054713-167054713 on Build GRCh38
rs906739623 Chr.1: 167054737-167054737 on Build GRCh38
rs774773587 Chr.1: 167054368-167054368 on Build GRCh38
rs139067139 Chr.1: 167054363-167054363 on Build GRCh38
rs752628346 Chr.1: 167054331-167054331 on Build GRCh38
rs772376155 Chr.1: 167054481-167054481 on Build GRCh38
rs770094266 Chr.1: 167054311-167054311 on Build GRCh38

View Full Product Details