Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167090542-167091033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPA33
Entrez Gene ID
10223
Gene Name
glycoprotein A33
Gene Aliases
A33
UniGene
Hs.651244
Gene Chromosome Location
Chr.1: 167052836-167090631 on Build GRCh38
Transcripts
NM_005814.2 XM_017003063.1

Gene Symbol
LOC105371598
Entrez Gene ID
105371598
Gene Name
uncharacterized LOC105371598
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.1: 167088000-167091553 on Build GRCh38
Transcripts
NM_005814.2 XM_017003063.1

SNP Detail

SNP ID Chromosome Location
rs373109894 Chr.1: 167090868-167090868 on Build GRCh38
rs546547339 Chr.1: 167090736-167090736 on Build GRCh38
rs35777372 Chr.1: 167090843-167090843 on Build GRCh38
rs149274372 Chr.1: 167090759-167090759 on Build GRCh38
rs889319414 Chr.1: 167090728-167090728 on Build GRCh38
rs901831429 Chr.1: 167090800-167090800 on Build GRCh38
rs544691409 Chr.1: 167090855-167090855 on Build GRCh38
rs904644625 Chr.1: 167090919-167090919 on Build GRCh38
rs948765208 Chr.1: 167090899-167090899 on Build GRCh38
rs112148498 Chr.1: 167090839-167090839 on Build GRCh38
rs974378629 Chr.1: 167090744-167090744 on Build GRCh38
rs540155824 Chr.1: 167090831-167090831 on Build GRCh38
rs924286836 Chr.1: 167091008-167091008 on Build GRCh38
rs1019239618 Chr.1: 167090632-167090632 on Build GRCh38
rs1016710198 Chr.1: 167090854-167090854 on Build GRCh38
rs533552467 Chr.1: 167090997-167090997 on Build GRCh38
rs2281960 Chr.1: 167090596-167090596 on Build GRCh38
rs1044354269 Chr.1: 167090901-167090901 on Build GRCh38
rs984319225 Chr.1: 167090818-167090818 on Build GRCh38
rs550803537 Chr.1: 167090565-167090565 on Build GRCh38
rs902893863 Chr.1: 167090625-167090625 on Build GRCh38
rs1026733786 Chr.1: 167090838-167090838 on Build GRCh38
rs201486979 Chr.1: 167090838-167090839 on Build GRCh38
rs575073161 Chr.1: 167090784-167090784 on Build GRCh38
rs917139840 Chr.1: 167090977-167090977 on Build GRCh38
rs2281961 Chr.1: 167090593-167090593 on Build GRCh38
rs142567853 Chr.1: 167090856-167090856 on Build GRCh38
rs971724627 Chr.1: 167090988-167090988 on Build GRCh38
rs909735616 Chr.1: 167090578-167090578 on Build GRCh38
rs1000008862 Chr.1: 167090573-167090573 on Build GRCh38
rs563061539 Chr.1: 167090566-167090566 on Build GRCh38
rs949780775 Chr.1: 167090599-167090599 on Build GRCh38
rs190462448 Chr.1: 167090782-167090782 on Build GRCh38
rs937230490 Chr.1: 167090582-167090582 on Build GRCh38
rs765884595 Chr.1: 167090622-167090622 on Build GRCh38
rs776889855 Chr.1: 167090842-167090842 on Build GRCh38
rs977091711 Chr.1: 167090992-167090992 on Build GRCh38
rs1054601285 Chr.1: 167090687-167090687 on Build GRCh38
rs567206621 Chr.1: 167090606-167090606 on Build GRCh38
rs991358379 Chr.1: 167090951-167090951 on Build GRCh38
rs539280777 Chr.1: 167090795-167090795 on Build GRCh38
rs1000228067 Chr.1: 167090944-167090944 on Build GRCh38
rs534241675 Chr.1: 167090647-167090647 on Build GRCh38
rs535524947 Chr.1: 167090774-167090774 on Build GRCh38
rs1019245493 Chr.1: 167090758-167090758 on Build GRCh38
rs935756504 Chr.1: 167090686-167090686 on Build GRCh38
rs1031504882 Chr.1: 167090986-167090986 on Build GRCh38
rs113555225 Chr.1: 167090807-167090807 on Build GRCh38
rs765297317 Chr.1: 167090870-167090870 on Build GRCh38
rs557545462 Chr.1: 167090803-167090803 on Build GRCh38
rs113286931 Chr.1: 167090739-167090739 on Build GRCh38

View Full Product Details