Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 81658904-81659411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPA1
Entrez Gene ID
3612
Gene Name
inositol monophosphatase 1
Gene Aliases
IMP, IMPA
UniGene
Hs.656694
Gene Chromosome Location
Chr.8: 81656916-81686354 on Build GRCh38
Transcripts
NM_001144878.1 NM_001144879.1 NM_005536.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3651325 c.782G>A p.R261K Chr.8: 81659403-81659403 on Build GRCh38
6181933 c.970G>C p.E324Q Chr.8: 81659392-81659392 on Build GRCh38
2876991 c.798T>C p.I266I Chr.8: 81659387-81659387 on Build GRCh38
3651324 c.959G>A p.R320K Chr.8: 81659403-81659403 on Build GRCh38
751641 c.825C>T p.D275D Chr.8: 81659360-81659360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773393432 Chr.8: 81659027-81659027 on Build GRCh38
rs575339306 Chr.8: 81659169-81659169 on Build GRCh38
rs553908024 Chr.8: 81659073-81659073 on Build GRCh38
rs969792290 Chr.8: 81659274-81659274 on Build GRCh38
rs764082454 Chr.8: 81659369-81659369 on Build GRCh38
rs372313423 Chr.8: 81659363-81659363 on Build GRCh38
rs557151871 Chr.8: 81659049-81659049 on Build GRCh38
rs745483441 Chr.8: 81659327-81659327 on Build GRCh38
rs572464432 Chr.8: 81659132-81659132 on Build GRCh38
rs552863713 Chr.8: 81659095-81659095 on Build GRCh38
rs758492327 Chr.8: 81659218-81659218 on Build GRCh38
rs773742914 Chr.8: 81659355-81659355 on Build GRCh38
rs770239191 Chr.8: 81659352-81659352 on Build GRCh38
rs778603291 Chr.8: 81659307-81659307 on Build GRCh38
rs569379428 Chr.8: 81658980-81658980 on Build GRCh38
rs1011238446 Chr.8: 81659230-81659230 on Build GRCh38
rs767783535 Chr.8: 81659362-81659362 on Build GRCh38
rs544608670 Chr.8: 81659148-81659148 on Build GRCh38
rs530342052 Chr.8: 81659202-81659202 on Build GRCh38
rs112960654 Chr.8: 81659248-81659249 on Build GRCh38
rs1012146271 Chr.8: 81659270-81659270 on Build GRCh38
rs10105165 Chr.8: 81659344-81659344 on Build GRCh38
rs922199431 Chr.8: 81659128-81659128 on Build GRCh38
rs929213633 Chr.8: 81658957-81658957 on Build GRCh38
rs771811787 Chr.8: 81659328-81659328 on Build GRCh38
rs895079303 Chr.8: 81659268-81659268 on Build GRCh38
rs970820296 Chr.8: 81658981-81658981 on Build GRCh38
rs748984652 Chr.8: 81659302-81659303 on Build GRCh38
rs981263373 Chr.8: 81658929-81658929 on Build GRCh38
rs370041028 Chr.8: 81659076-81659076 on Build GRCh38
rs755994131 Chr.8: 81659367-81659367 on Build GRCh38
rs1001109417 Chr.8: 81659206-81659206 on Build GRCh38
rs773830760 Chr.8: 81659295-81659295 on Build GRCh38
rs908029335 Chr.8: 81659071-81659071 on Build GRCh38
rs973614465 Chr.8: 81659088-81659088 on Build GRCh38
rs891307121 Chr.8: 81659144-81659144 on Build GRCh38
rs186229166 Chr.8: 81659182-81659182 on Build GRCh38
rs1047691063 Chr.8: 81658965-81658965 on Build GRCh38
rs999036558 Chr.8: 81659183-81659183 on Build GRCh38
rs768524208 Chr.8: 81659355-81659356 on Build GRCh38
rs776798105 Chr.8: 81659330-81659330 on Build GRCh38
rs1038460056 Chr.8: 81659045-81659045 on Build GRCh38
rs536113859 Chr.8: 81659094-81659094 on Build GRCh38
rs775410099 Chr.8: 81659276-81659276 on Build GRCh38
rs928458715 Chr.8: 81658952-81658952 on Build GRCh38
rs766424862 Chr.8: 81659357-81659357 on Build GRCh38
rs532545900 Chr.8: 81659293-81659293 on Build GRCh38
rs756855526 Chr.8: 81659300-81659300 on Build GRCh38
rs560778521 Chr.8: 81659285-81659285 on Build GRCh38
rs190693412 Chr.8: 81659261-81659261 on Build GRCh38
rs573831926 Chr.8: 81659089-81659089 on Build GRCh38
rs183172092 Chr.8: 81659164-81659164 on Build GRCh38
rs2955014 Chr.8: 81659199-81659199 on Build GRCh38
rs536842609 Chr.8: 81659007-81659007 on Build GRCh38
rs943798573 Chr.8: 81659193-81659193 on Build GRCh38
rs1049412508 Chr.8: 81659140-81659140 on Build GRCh38
rs908530900 Chr.8: 81658976-81658976 on Build GRCh38
rs1053606270 Chr.8: 81659254-81659254 on Build GRCh38
rs144509656 Chr.8: 81659359-81659359 on Build GRCh38
rs928457327 Chr.8: 81659332-81659332 on Build GRCh38
rs368377674 Chr.8: 81659331-81659331 on Build GRCh38
rs912236525 Chr.8: 81659165-81659165 on Build GRCh38
rs1004935117 Chr.8: 81659376-81659376 on Build GRCh38

View Full Product Details