Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 81673752-81674255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPA1
Entrez Gene ID
3612
Gene Name
inositol monophosphatase 1
Gene Aliases
IMP, IMPA
UniGene
Hs.656694
Gene Chromosome Location
Chr.8: 81656916-81686354 on Build GRCh38
Transcripts
NM_001144878.1 NM_001144879.1 NM_005536.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5948326 c.526-8delC p.? Chr.8: 81673957-81673957 on Build GRCh38
3901771 c.424A>G p.N142D Chr.8: 81673874-81673874 on Build GRCh38
461403 c.438A>G p.L146L Chr.8: 81673860-81673860 on Build GRCh38
1458374 c.435A>G p.K145K Chr.8: 81673863-81673863 on Build GRCh38
3901768 c.606T>C p.G202G Chr.8: 81673869-81673869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868132323 Chr.8: 81673785-81673785 on Build GRCh38
rs536222560 Chr.8: 81673790-81673790 on Build GRCh38
rs149891590 Chr.8: 81673967-81673967 on Build GRCh38
rs753984254 Chr.8: 81674000-81674000 on Build GRCh38
rs770680532 Chr.8: 81673864-81673864 on Build GRCh38
rs191162314 Chr.8: 81674022-81674022 on Build GRCh38
rs370768848 Chr.8: 81673988-81673988 on Build GRCh38
rs558424491 Chr.8: 81674155-81674155 on Build GRCh38
rs745791755 Chr.8: 81673875-81673875 on Build GRCh38
rs377431444 Chr.8: 81673818-81673818 on Build GRCh38
rs759615303 Chr.8: 81674000-81674001 on Build GRCh38
rs3214541 Chr.8: 81674056-81674057 on Build GRCh38
rs200950319 Chr.8: 81673872-81673872 on Build GRCh38
rs768151800 Chr.8: 81673984-81673984 on Build GRCh38
rs776118679 Chr.8: 81673953-81673953 on Build GRCh38
rs1035999053 Chr.8: 81673894-81673894 on Build GRCh38
rs752041231 Chr.8: 81673968-81673968 on Build GRCh38
rs1020318207 Chr.8: 81674079-81674079 on Build GRCh38
rs761609809 Chr.8: 81673954-81673954 on Build GRCh38
rs992073760 Chr.8: 81674195-81674195 on Build GRCh38
rs566621311 Chr.8: 81674226-81674226 on Build GRCh38
rs929260657 Chr.8: 81674233-81674233 on Build GRCh38
rs757460207 Chr.8: 81674199-81674199 on Build GRCh38
rs967851157 Chr.8: 81674096-81674096 on Build GRCh38
rs760113609 Chr.8: 81673937-81673937 on Build GRCh38
rs574265720 Chr.8: 81673964-81673965 on Build GRCh38
rs1031903625 Chr.8: 81674003-81674003 on Build GRCh38
rs537263906 Chr.8: 81674095-81674095 on Build GRCh38
rs766892204 Chr.8: 81673794-81673794 on Build GRCh38
rs376742471 Chr.8: 81673919-81673919 on Build GRCh38
rs756303906 Chr.8: 81673848-81673848 on Build GRCh38
rs777881800 Chr.8: 81673849-81673849 on Build GRCh38
rs752774247 Chr.8: 81673987-81673987 on Build GRCh38
rs897766403 Chr.8: 81673838-81673838 on Build GRCh38
rs967750198 Chr.8: 81674106-81674106 on Build GRCh38
rs939187148 Chr.8: 81674157-81674157 on Build GRCh38
rs17850313 Chr.8: 81673854-81673854 on Build GRCh38
rs373743142 Chr.8: 81673974-81673974 on Build GRCh38
rs753195422 Chr.8: 81673985-81673985 on Build GRCh38
rs776687363 Chr.8: 81673978-81673978 on Build GRCh38
rs926110851 Chr.8: 81674148-81674148 on Build GRCh38
rs79636176 Chr.8: 81674136-81674136 on Build GRCh38
rs372426719 Chr.8: 81674133-81674133 on Build GRCh38
rs781238704 Chr.8: 81674232-81674232 on Build GRCh38
rs1056725556 Chr.8: 81674175-81674175 on Build GRCh38
rs917471134 Chr.8: 81674196-81674196 on Build GRCh38
rs369641233 Chr.8: 81673881-81673881 on Build GRCh38
rs939242403 Chr.8: 81673784-81673784 on Build GRCh38
rs749069269 Chr.8: 81673860-81673860 on Build GRCh38
rs768303327 Chr.8: 81673944-81673944 on Build GRCh38
rs772060191 Chr.8: 81673905-81673905 on Build GRCh38
rs914919028 Chr.8: 81674064-81674064 on Build GRCh38
rs956314084 Chr.8: 81674008-81674008 on Build GRCh38
rs373708876 Chr.8: 81673989-81673989 on Build GRCh38
rs755604618 Chr.8: 81673815-81673815 on Build GRCh38
rs953277361 Chr.8: 81673972-81673972 on Build GRCh38
rs752094409 Chr.8: 81673798-81673798 on Build GRCh38

View Full Product Details