Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94827222-94827732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS8
Entrez Gene ID
55656
Gene Name
integrator complex subunit 8
Gene Aliases
C8orf52, INT8
UniGene
Hs.727669
Gene Chromosome Location
Chr.8: 94823290-94894664 on Build GRCh38
Transcripts
NM_017864.3 NR_073444.1 NR_073445.1 XM_017013616.1 XM_017013617.1 XM_017013618.1 XM_017013619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7122960 c.409C>T p.P137S Chr.8: 94827366-94827366 on Build GRCh38
3726882 c.397G>C p.A133P Chr.8: 94827354-94827354 on Build GRCh38
5757267 c.441T>G p.N147K Chr.8: 94827398-94827398 on Build GRCh38
405112 c.328_329insT p.M110fs*5 Chr.8: 94827285-94827286 on Build GRCh38
4925170 c.449C>A p.A150E Chr.8: 94827724-94827724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774736396 Chr.8: 94827707-94827707 on Build GRCh38
rs557882923 Chr.8: 94827524-94827524 on Build GRCh38
rs1014115739 Chr.8: 94827584-94827584 on Build GRCh38
rs779690391 Chr.8: 94827445-94827445 on Build GRCh38
rs370688322 Chr.8: 94827255-94827255 on Build GRCh38
rs763536818 Chr.8: 94827344-94827344 on Build GRCh38
rs375467035 Chr.8: 94827455-94827455 on Build GRCh38
rs1024652410 Chr.8: 94827607-94827607 on Build GRCh38
rs764147184 Chr.8: 94827393-94827393 on Build GRCh38
rs771425594 Chr.8: 94827695-94827695 on Build GRCh38
rs937040446 Chr.8: 94827541-94827541 on Build GRCh38
rs185845898 Chr.8: 94827431-94827431 on Build GRCh38
rs1037442609 Chr.8: 94827496-94827496 on Build GRCh38
rs375805430 Chr.8: 94827290-94827290 on Build GRCh38
rs940458878 Chr.8: 94827481-94827481 on Build GRCh38
rs753774112 Chr.8: 94827281-94827281 on Build GRCh38
rs778747564 Chr.8: 94827286-94827286 on Build GRCh38
rs978258948 Chr.8: 94827662-94827662 on Build GRCh38
rs752871566 Chr.8: 94827366-94827366 on Build GRCh38
rs778749075 Chr.8: 94827417-94827417 on Build GRCh38
rs933527397 Chr.8: 94827454-94827454 on Build GRCh38
rs191142007 Chr.8: 94827572-94827572 on Build GRCh38
rs760958145 Chr.8: 94827371-94827371 on Build GRCh38
rs149807637 Chr.8: 94827354-94827354 on Build GRCh38
rs763668989 Chr.8: 94827302-94827302 on Build GRCh38
rs761205918 Chr.8: 94827679-94827682 on Build GRCh38
rs776344019 Chr.8: 94827342-94827342 on Build GRCh38
rs757244580 Chr.8: 94827285-94827285 on Build GRCh38
rs753968659 Chr.8: 94827398-94827398 on Build GRCh38
rs188064876 Chr.8: 94827597-94827597 on Build GRCh38
rs902995258 Chr.8: 94827594-94827594 on Build GRCh38
rs759843814 Chr.8: 94827357-94827357 on Build GRCh38
rs183807003 Chr.8: 94827586-94827586 on Build GRCh38
rs771388344 Chr.8: 94827345-94827345 on Build GRCh38
rs779594257 Chr.8: 94827324-94827324 on Build GRCh38
rs112990773 Chr.8: 94827628-94827628 on Build GRCh38
rs577767812 Chr.8: 94827525-94827525 on Build GRCh38
rs923690253 Chr.8: 94827664-94827664 on Build GRCh38
rs758875840 Chr.8: 94827348-94827348 on Build GRCh38
rs753485075 Chr.8: 94827483-94827483 on Build GRCh38
rs567627377 Chr.8: 94827305-94827305 on Build GRCh38
rs183337632 Chr.8: 94827259-94827259 on Build GRCh38
rs746654759 Chr.8: 94827449-94827449 on Build GRCh38
rs1032448290 Chr.8: 94827600-94827600 on Build GRCh38
rs747698703 Chr.8: 94827689-94827689 on Build GRCh38
rs115942323 Chr.8: 94827503-94827503 on Build GRCh38
rs888623272 Chr.8: 94827548-94827548 on Build GRCh38
rs545265955 Chr.8: 94827511-94827511 on Build GRCh38
rs750183143 Chr.8: 94827430-94827430 on Build GRCh38
rs768284706 Chr.8: 94827341-94827341 on Build GRCh38
rs368018334 Chr.8: 94827667-94827667 on Build GRCh38
rs760223689 Chr.8: 94827253-94827253 on Build GRCh38
rs536512408 Chr.8: 94827326-94827326 on Build GRCh38
rs780822843 Chr.8: 94827673-94827673 on Build GRCh38
rs746664328 Chr.8: 94827617-94827620 on Build GRCh38
rs542936494 Chr.8: 94827641-94827641 on Build GRCh38
rs908039889 Chr.8: 94827480-94827480 on Build GRCh38
rs767724233 Chr.8: 94827358-94827358 on Build GRCh38
rs767669653 Chr.8: 94827247-94827247 on Build GRCh38
rs1034496578 Chr.8: 94827573-94827573 on Build GRCh38
rs757157062 Chr.8: 94827410-94827410 on Build GRCh38
rs761516772 Chr.8: 94827640-94827640 on Build GRCh38

View Full Product Details