Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94831851-94832395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS8
Entrez Gene ID
55656
Gene Name
integrator complex subunit 8
Gene Aliases
C8orf52, INT8
UniGene
Hs.727669
Gene Chromosome Location
Chr.8: 94823290-94894664 on Build GRCh38
Transcripts
NM_017864.3 NR_073444.1 NR_073445.1 XM_017013616.1 XM_017013617.1 XM_017013618.1 XM_017013619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3652014 c.751C>A p.Q251K Chr.8: 94832172-94832172 on Build GRCh38
6853015 c.647C>G p.T216R Chr.8: 94832068-94832068 on Build GRCh38
6315153 c.640G>T p.V214F Chr.8: 94832061-94832061 on Build GRCh38
5798676 c.741A>T p.E247D Chr.8: 94832162-94832162 on Build GRCh38
4381596 c.575A>G p.K192R Chr.8: 94831996-94831996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200279468 Chr.8: 94832226-94832226 on Build GRCh38
rs772664425 Chr.8: 94832225-94832225 on Build GRCh38
rs201252065 Chr.8: 94832133-94832133 on Build GRCh38
rs551888205 Chr.8: 94831906-94831906 on Build GRCh38
rs376751042 Chr.8: 94832187-94832187 on Build GRCh38
rs748582735 Chr.8: 94832024-94832024 on Build GRCh38
rs745977096 Chr.8: 94832190-94832190 on Build GRCh38
rs986234354 Chr.8: 94832270-94832270 on Build GRCh38
rs749516355 Chr.8: 94832171-94832171 on Build GRCh38
rs767411130 Chr.8: 94831946-94831946 on Build GRCh38
rs944462589 Chr.8: 94832276-94832276 on Build GRCh38
rs760213962 Chr.8: 94831978-94831978 on Build GRCh38
rs564677351 Chr.8: 94831884-94831884 on Build GRCh38
rs775108211 Chr.8: 94832100-94832100 on Build GRCh38
rs535481635 Chr.8: 94832255-94832255 on Build GRCh38
rs757155998 Chr.8: 94832207-94832208 on Build GRCh38
rs559016415 Chr.8: 94831928-94831928 on Build GRCh38
rs577858605 Chr.8: 94832358-94832358 on Build GRCh38
rs537317808 Chr.8: 94832157-94832157 on Build GRCh38
rs190757765 Chr.8: 94832198-94832198 on Build GRCh38
rs771133747 Chr.8: 94832068-94832068 on Build GRCh38
rs564611067 Chr.8: 94832343-94832343 on Build GRCh38
rs547782053 Chr.8: 94832199-94832199 on Build GRCh38
rs555702010 Chr.8: 94832260-94832260 on Build GRCh38
rs753239742 Chr.8: 94831988-94831988 on Build GRCh38
rs764854837 Chr.8: 94831943-94831945 on Build GRCh38
rs769864028 Chr.8: 94832037-94832037 on Build GRCh38
rs752055122 Chr.8: 94832014-94832014 on Build GRCh38
rs905355201 Chr.8: 94831942-94831942 on Build GRCh38
rs772086443 Chr.8: 94832194-94832194 on Build GRCh38
rs753534590 Chr.8: 94832127-94832127 on Build GRCh38
rs1005755864 Chr.8: 94831930-94831931 on Build GRCh38
rs758156365 Chr.8: 94831978-94831981 on Build GRCh38
rs866996340 Chr.8: 94832119-94832119 on Build GRCh38
rs778068612 Chr.8: 94832166-94832166 on Build GRCh38
rs774632834 Chr.8: 94832069-94832069 on Build GRCh38
rs780470909 Chr.8: 94831996-94831996 on Build GRCh38
rs979672233 Chr.8: 94832236-94832236 on Build GRCh38
rs544919905 Chr.8: 94832287-94832287 on Build GRCh38
rs770840763 Chr.8: 94831920-94831920 on Build GRCh38
rs780829660 Chr.8: 94831889-94831889 on Build GRCh38
rs570686474 Chr.8: 94832189-94832189 on Build GRCh38
rs1033678962 Chr.8: 94832152-94832152 on Build GRCh38
rs1008646023 Chr.8: 94831999-94831999 on Build GRCh38
rs760541827 Chr.8: 94832111-94832111 on Build GRCh38
rs910441386 Chr.8: 94832180-94832180 on Build GRCh38
rs145640930 Chr.8: 94832072-94832072 on Build GRCh38
rs140588875 Chr.8: 94832048-94832048 on Build GRCh38
rs749385633 Chr.8: 94832222-94832222 on Build GRCh38
rs775512897 Chr.8: 94832197-94832197 on Build GRCh38
rs988246379 Chr.8: 94832137-94832137 on Build GRCh38
rs951927080 Chr.8: 94832104-94832104 on Build GRCh38
rs763597932 Chr.8: 94832117-94832117 on Build GRCh38
rs752590044 Chr.8: 94831948-94831948 on Build GRCh38
rs547031391 Chr.8: 94832156-94832156 on Build GRCh38
rs748176236 Chr.8: 94832004-94832004 on Build GRCh38
rs905415447 Chr.8: 94832321-94832321 on Build GRCh38
rs774685051 Chr.8: 94831938-94831940 on Build GRCh38
rs749983516 Chr.8: 94832135-94832135 on Build GRCh38
rs757606871 Chr.8: 94832177-94832177 on Build GRCh38
rs374779459 Chr.8: 94832207-94832207 on Build GRCh38
rs751329641 Chr.8: 94832170-94832181 on Build GRCh38
rs17719449 Chr.8: 94832210-94832210 on Build GRCh38
rs777500986 Chr.8: 94832131-94832133 on Build GRCh38
rs752394386 Chr.8: 94831971-94831971 on Build GRCh38
rs377738957 Chr.8: 94832016-94832016 on Build GRCh38
rs763421394 Chr.8: 94831914-94831917 on Build GRCh38
rs575873434 Chr.8: 94832273-94832273 on Build GRCh38
rs528122547 Chr.8: 94832088-94832088 on Build GRCh38
rs767733255 Chr.8: 94832105-94832105 on Build GRCh38
rs781000970 Chr.8: 94832208-94832208 on Build GRCh38
rs759527863 Chr.8: 94832076-94832076 on Build GRCh38
rs777270426 Chr.8: 94832206-94832206 on Build GRCh38
rs377644142 Chr.8: 94832184-94832184 on Build GRCh38
rs533329284 Chr.8: 94831905-94831905 on Build GRCh38
rs764909803 Chr.8: 94832216-94832216 on Build GRCh38
rs754846138 Chr.8: 94831883-94831883 on Build GRCh38
rs138094711 Chr.8: 94832284-94832284 on Build GRCh38
rs925561757 Chr.8: 94832238-94832238 on Build GRCh38
rs370079184 Chr.8: 94832367-94832367 on Build GRCh38
rs186708108 Chr.8: 94832142-94832142 on Build GRCh38
rs539554395 Chr.8: 94832140-94832140 on Build GRCh38
rs756805905 Chr.8: 94831995-94831995 on Build GRCh38

View Full Product Details