Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94836439-94836915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS8
Entrez Gene ID
55656
Gene Name
integrator complex subunit 8
Gene Aliases
C8orf52, INT8
UniGene
Hs.727669
Gene Chromosome Location
Chr.8: 94823290-94894664 on Build GRCh38
Transcripts
NM_017864.3 NR_073444.1 NR_073445.1 XM_017013616.1 XM_017013617.1 XM_017013618.1 XM_017013619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3652015 c.803C>T p.S268L Chr.8: 94836573-94836573 on Build GRCh38
6472760 c.787C>T p.Q263* Chr.8: 94836557-94836557 on Build GRCh38
1643664 c.851T>A p.L284Q Chr.8: 94836621-94836621 on Build GRCh38
752459 c.777A>T p.A259A Chr.8: 94836547-94836547 on Build GRCh38
1102672 c.827A>G p.E276G Chr.8: 94836597-94836597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895554937 Chr.8: 94836890-94836890 on Build GRCh38
rs6471497 Chr.8: 94836805-94836805 on Build GRCh38
rs1056831269 Chr.8: 94836858-94836858 on Build GRCh38
rs762282399 Chr.8: 94836681-94836681 on Build GRCh38
rs753250205 Chr.8: 94836480-94836480 on Build GRCh38
rs764500058 Chr.8: 94836506-94836506 on Build GRCh38
rs754206097 Chr.8: 94836674-94836674 on Build GRCh38
rs754297953 Chr.8: 94836510-94836510 on Build GRCh38
rs984023488 Chr.8: 94836748-94836748 on Build GRCh38
rs72486224 Chr.8: 94836850-94836851 on Build GRCh38
rs149765435 Chr.8: 94836845-94836846 on Build GRCh38
rs1007510187 Chr.8: 94836886-94836886 on Build GRCh38
rs549449671 Chr.8: 94836712-94836712 on Build GRCh38
rs370466637 Chr.8: 94836542-94836542 on Build GRCh38
rs778250436 Chr.8: 94836511-94836511 on Build GRCh38
rs1022303591 Chr.8: 94836845-94836848 on Build GRCh38
rs780066941 Chr.8: 94836578-94836578 on Build GRCh38
rs115309393 Chr.8: 94836794-94836794 on Build GRCh38
rs200460975 Chr.8: 94836659-94836659 on Build GRCh38
rs921913564 Chr.8: 94836648-94836648 on Build GRCh38
rs938729698 Chr.8: 94836799-94836799 on Build GRCh38
rs754675969 Chr.8: 94836644-94836644 on Build GRCh38
rs1010004860 Chr.8: 94836515-94836515 on Build GRCh38
rs746949799 Chr.8: 94836580-94836580 on Build GRCh38
rs940864784 Chr.8: 94836719-94836719 on Build GRCh38
rs920881004 Chr.8: 94836728-94836728 on Build GRCh38
rs772862347 Chr.8: 94836623-94836623 on Build GRCh38
rs781696043 Chr.8: 94836693-94836693 on Build GRCh38
rs778103259 Chr.8: 94836819-94836819 on Build GRCh38
rs537579792 Chr.8: 94836835-94836835 on Build GRCh38
rs756639874 Chr.8: 94836503-94836503 on Build GRCh38
rs747968129 Chr.8: 94836610-94836610 on Build GRCh38
rs969294229 Chr.8: 94836668-94836668 on Build GRCh38
rs985613110 Chr.8: 94836678-94836678 on Build GRCh38
rs764543634 Chr.8: 94836661-94836661 on Build GRCh38
rs370048441 Chr.8: 94836606-94836606 on Build GRCh38
rs767850277 Chr.8: 94836479-94836479 on Build GRCh38
rs1035500776 Chr.8: 94836488-94836488 on Build GRCh38
rs1044895645 Chr.8: 94836788-94836788 on Build GRCh38
rs773785096 Chr.8: 94836651-94836651 on Build GRCh38
rs538230484 Chr.8: 94836720-94836720 on Build GRCh38
rs768917684 Chr.8: 94836571-94836572 on Build GRCh38
rs765901853 Chr.8: 94836628-94836628 on Build GRCh38
rs146535460 Chr.8: 94836541-94836541 on Build GRCh38
rs149581605 Chr.8: 94836768-94836768 on Build GRCh38
rs1002906887 Chr.8: 94836469-94836469 on Build GRCh38
rs552102086 Chr.8: 94836743-94836743 on Build GRCh38
rs762381138 Chr.8: 94836625-94836625 on Build GRCh38
rs758511855 Chr.8: 94836543-94836543 on Build GRCh38
rs965155408 Chr.8: 94836685-94836685 on Build GRCh38
rs565922073 Chr.8: 94836752-94836752 on Build GRCh38
rs768522808 Chr.8: 94836582-94836582 on Build GRCh38
rs34273729 Chr.8: 94836840-94836840 on Build GRCh38
rs557475564 Chr.8: 94836851-94836851 on Build GRCh38
rs769384020 Chr.8: 94836620-94836620 on Build GRCh38

View Full Product Details