Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 125024567-125025068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0196
Entrez Gene ID
9897
Gene Name
KIAA0196
Gene Aliases
RTSC, RTSC1, SPG8
UniGene
Hs.270043
Gene Chromosome Location
Chr.8: 125024260-125091819 on Build GRCh38
Transcripts
NM_014846.3 XM_005251120.3 XM_017014113.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5745020 c.3446A>T p.N1149I Chr.8: 125024651-125024651 on Build GRCh38
3367193 c.3432A>C p.E1144D Chr.8: 125024665-125024665 on Build GRCh38
6693022 c.3456delT p.F1152fs*>8 Chr.8: 125024641-125024641 on Build GRCh38
3778894 c.3457G>A p.D1153N Chr.8: 125024640-125024640 on Build GRCh38
5774885 c.3474G>C p.V1158V Chr.8: 125024623-125024623 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919803081 Chr.8: 125024915-125024915 on Build GRCh38
rs764539889 Chr.8: 125024659-125024659 on Build GRCh38
rs1000212557 Chr.8: 125024832-125024832 on Build GRCh38
rs776659440 Chr.8: 125024630-125024630 on Build GRCh38
rs771999043 Chr.8: 125024593-125024593 on Build GRCh38
rs775825862 Chr.8: 125024675-125024675 on Build GRCh38
rs752142468 Chr.8: 125024685-125024685 on Build GRCh38
rs746336406 Chr.8: 125024620-125024620 on Build GRCh38
rs140825931 Chr.8: 125024776-125024776 on Build GRCh38
rs1032585637 Chr.8: 125024843-125024843 on Build GRCh38
rs903866046 Chr.8: 125024722-125024722 on Build GRCh38
rs920977650 Chr.8: 125024683-125024683 on Build GRCh38
rs1028090746 Chr.8: 125024855-125024855 on Build GRCh38
rs984656396 Chr.8: 125024836-125024836 on Build GRCh38
rs747329114 Chr.8: 125024704-125024704 on Build GRCh38
rs953754648 Chr.8: 125024872-125024872 on Build GRCh38
rs1050550623 Chr.8: 125024947-125024947 on Build GRCh38
rs923133327 Chr.8: 125024913-125024913 on Build GRCh38
rs774068159 Chr.8: 125024594-125024594 on Build GRCh38
rs912414685 Chr.8: 125024900-125024900 on Build GRCh38
rs774785909 Chr.8: 125024937-125024938 on Build GRCh38
rs868296854 Chr.8: 125025000-125025000 on Build GRCh38
rs187590959 Chr.8: 125024797-125024797 on Build GRCh38
rs769385731 Chr.8: 125024827-125024827 on Build GRCh38
rs907826814 Chr.8: 125024837-125024837 on Build GRCh38
rs949493416 Chr.8: 125024992-125024992 on Build GRCh38
rs531879765 Chr.8: 125024737-125024737 on Build GRCh38
rs748065422 Chr.8: 125024605-125024605 on Build GRCh38
rs1055137518 Chr.8: 125024795-125024795 on Build GRCh38
rs780398820 Chr.8: 125024701-125024701 on Build GRCh38
rs17398450 Chr.8: 125024791-125024791 on Build GRCh38
rs780929650 Chr.8: 125025023-125025026 on Build GRCh38
rs931321125 Chr.8: 125024918-125024918 on Build GRCh38
rs772334826 Chr.8: 125024631-125024631 on Build GRCh38
rs748221523 Chr.8: 125024779-125024781 on Build GRCh38
rs766096156 Chr.8: 125024908-125024908 on Build GRCh38
rs986551293 Chr.8: 125024885-125024885 on Build GRCh38
rs989931355 Chr.8: 125024656-125024656 on Build GRCh38
rs1013793439 Chr.8: 125024771-125024774 on Build GRCh38
rs772184173 Chr.8: 125024606-125024606 on Build GRCh38
rs769870389 Chr.8: 125024619-125024623 on Build GRCh38
rs781660589 Chr.8: 125024693-125024693 on Build GRCh38
rs773048670 Chr.8: 125024844-125024844 on Build GRCh38
rs961228927 Chr.8: 125024901-125024901 on Build GRCh38
rs35499702 Chr.8: 125024641-125024641 on Build GRCh38
rs562865800 Chr.8: 125024706-125024706 on Build GRCh38
rs536950013 Chr.8: 125024822-125024822 on Build GRCh38
rs769201837 Chr.8: 125024717-125024722 on Build GRCh38
rs902713035 Chr.8: 125025002-125025002 on Build GRCh38
rs759863027 Chr.8: 125024635-125024635 on Build GRCh38
rs953868695 Chr.8: 125024734-125024734 on Build GRCh38
rs781072677 Chr.8: 125024598-125024600 on Build GRCh38
rs757502744 Chr.8: 125024691-125024691 on Build GRCh38
rs569426560 Chr.8: 125024614-125024614 on Build GRCh38
rs776396649 Chr.8: 125024800-125024800 on Build GRCh38
rs1031281519 Chr.8: 125024753-125024753 on Build GRCh38
rs916682548 Chr.8: 125024976-125024976 on Build GRCh38
rs1055020328 Chr.8: 125024962-125024962 on Build GRCh38
rs377725102 Chr.8: 125024845-125024845 on Build GRCh38
rs779162197 Chr.8: 125024740-125024744 on Build GRCh38
rs192545899 Chr.8: 125024738-125024738 on Build GRCh38
rs182909922 Chr.8: 125024768-125024768 on Build GRCh38
rs752817483 Chr.8: 125024648-125024648 on Build GRCh38
rs902660169 Chr.8: 125024819-125024819 on Build GRCh38
rs549407575 Chr.8: 125024963-125024963 on Build GRCh38
rs758883409 Chr.8: 125024657-125024657 on Build GRCh38
rs768266028 Chr.8: 125024593-125024593 on Build GRCh38
rs557346189 Chr.8: 125024709-125024709 on Build GRCh38
rs762892593 Chr.8: 125024899-125024899 on Build GRCh38
rs778541992 Chr.8: 125024710-125024710 on Build GRCh38
rs766201677 Chr.8: 125024627-125024627 on Build GRCh38
rs761446740 Chr.8: 125024801-125024801 on Build GRCh38
rs1011215196 Chr.8: 125024848-125024848 on Build GRCh38
rs1036938448 Chr.8: 125024775-125024775 on Build GRCh38
rs763391243 Chr.8: 125024688-125024692 on Build GRCh38
rs765297353 Chr.8: 125024637-125024637 on Build GRCh38

View Full Product Details