Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 112660362-112660871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1958
Entrez Gene ID
158405
Gene Name
KIAA1958
Gene Aliases
-
UniGene
Hs.656619
Gene Chromosome Location
Chr.9: 112486968-112665311 on Build GRCh38
Transcripts
NM_001287036.1 NM_001287038.1 NM_133465.3

SNP Detail

SNP ID Chromosome Location
rs978939922 Chr.9: 112660410-112660410 on Build GRCh38
rs967241530 Chr.9: 112660790-112660790 on Build GRCh38
rs991515938 Chr.9: 112660636-112660636 on Build GRCh38
rs950179832 Chr.9: 112660674-112660674 on Build GRCh38
rs935894610 Chr.9: 112660635-112660635 on Build GRCh38
rs1008660840 Chr.9: 112660746-112660746 on Build GRCh38
rs1056292539 Chr.9: 112660767-112660767 on Build GRCh38
rs186382107 Chr.9: 112660462-112660462 on Build GRCh38
rs1045920710 Chr.9: 112660485-112660485 on Build GRCh38
rs531360378 Chr.9: 112660688-112660688 on Build GRCh38
rs1018797806 Chr.9: 112660591-112660591 on Build GRCh38
rs1044940798 Chr.9: 112660475-112660475 on Build GRCh38
rs375726575 Chr.9: 112660641-112660641 on Build GRCh38
rs888755979 Chr.9: 112660743-112660743 on Build GRCh38
rs7028901 Chr.9: 112660574-112660574 on Build GRCh38
rs1017105348 Chr.9: 112660527-112660527 on Build GRCh38
rs368433898 Chr.9: 112660713-112660713 on Build GRCh38
rs977345459 Chr.9: 112660618-112660618 on Build GRCh38
rs548492716 Chr.9: 112660844-112660844 on Build GRCh38
rs976909780 Chr.9: 112660657-112660657 on Build GRCh38
rs191135045 Chr.9: 112660501-112660501 on Build GRCh38
rs776932311 Chr.9: 112660718-112660718 on Build GRCh38
rs939169634 Chr.9: 112660753-112660753 on Build GRCh38
rs925898520 Chr.9: 112660619-112660619 on Build GRCh38
rs1032832235 Chr.9: 112660830-112660830 on Build GRCh38
rs865981930 Chr.9: 112660796-112660796 on Build GRCh38
rs766690810 Chr.9: 112660714-112660714 on Build GRCh38
rs1030166331 Chr.9: 112660668-112660668 on Build GRCh38
rs778506131 Chr.9: 112660427-112660427 on Build GRCh38
rs555688904 Chr.9: 112660447-112660447 on Build GRCh38
rs1007157832 Chr.9: 112660824-112660824 on Build GRCh38
rs769075951 Chr.9: 112660759-112660760 on Build GRCh38
rs987609781 Chr.9: 112660583-112660583 on Build GRCh38
rs887248534 Chr.9: 112660492-112660492 on Build GRCh38
rs941285528 Chr.9: 112660687-112660687 on Build GRCh38
rs771383106 Chr.9: 112660411-112660414 on Build GRCh38
rs909865397 Chr.9: 112660682-112660682 on Build GRCh38
rs571629826 Chr.9: 112660387-112660390 on Build GRCh38
rs772659571 Chr.9: 112660551-112660551 on Build GRCh38
rs764898703 Chr.9: 112660797-112660797 on Build GRCh38
rs552526135 Chr.9: 112660811-112660811 on Build GRCh38
rs1021960139 Chr.9: 112660573-112660573 on Build GRCh38
rs905013449 Chr.9: 112660479-112660479 on Build GRCh38
rs964126475 Chr.9: 112660425-112660425 on Build GRCh38
rs146413413 Chr.9: 112660675-112660675 on Build GRCh38
rs976820712 Chr.9: 112660426-112660427 on Build GRCh38
rs1038501453 Chr.9: 112660707-112660707 on Build GRCh38
rs762189645 Chr.9: 112660504-112660504 on Build GRCh38
rs951486913 Chr.9: 112660669-112660669 on Build GRCh38
rs898839459 Chr.9: 112660540-112660540 on Build GRCh38
rs62575228 Chr.9: 112660782-112660782 on Build GRCh38
rs935954409 Chr.9: 112660439-112660439 on Build GRCh38
rs925957320 Chr.9: 112660427-112660427 on Build GRCh38
rs542159909 Chr.9: 112660507-112660507 on Build GRCh38
rs941254204 Chr.9: 112660457-112660457 on Build GRCh38
rs904808692 Chr.9: 112660574-112660574 on Build GRCh38
rs183410438 Chr.9: 112660562-112660562 on Build GRCh38
rs953186942 Chr.9: 112660582-112660582 on Build GRCh38
rs574937745 Chr.9: 112660482-112660482 on Build GRCh38
rs1043813453 Chr.9: 112660793-112660793 on Build GRCh38
rs760881650 Chr.9: 112660593-112660593 on Build GRCh38
rs940143426 Chr.9: 112660695-112660695 on Build GRCh38

View Full Product Details