Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114094216-114094719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF12
Entrez Gene ID
113220
Gene Name
kinesin family member 12
Gene Aliases
-
UniGene
Hs.28149
Gene Chromosome Location
Chr.9: 114091623-114100099 on Build GRCh38
Transcripts
NM_138424.1 XM_005251683.4 XM_006716947.2 XM_011518190.2 XM_011518191.2 XM_011518192.2 XM_011518193.2 XM_017014240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6874134 c.808G>A p.A270T Chr.9: 114094353-114094353 on Build GRCh38
3653310 c.1183C>T p.Q395* Chr.9: 114094377-114094377 on Build GRCh38
5770167 c.765G>C p.E255D Chr.9: 114094396-114094396 on Build GRCh38
6694654 c.1140A>C p.T380T Chr.9: 114094420-114094420 on Build GRCh38
3653309 c.784C>T p.Q262* Chr.9: 114094377-114094377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762657264 Chr.9: 114094288-114094288 on Build GRCh38
rs755066105 Chr.9: 114094347-114094347 on Build GRCh38
rs981532200 Chr.9: 114094568-114094568 on Build GRCh38
rs772255782 Chr.9: 114094461-114094461 on Build GRCh38
rs755822198 Chr.9: 114094314-114094314 on Build GRCh38
rs545570151 Chr.9: 114094283-114094283 on Build GRCh38
rs78900227 Chr.9: 114094521-114094521 on Build GRCh38
rs759045422 Chr.9: 114094310-114094310 on Build GRCh38
rs185740710 Chr.9: 114094500-114094500 on Build GRCh38
rs754456860 Chr.9: 114094240-114094240 on Build GRCh38
rs768651392 Chr.9: 114094265-114094265 on Build GRCh38
rs1465660 Chr.9: 114094609-114094609 on Build GRCh38
rs200777995 Chr.9: 114094437-114094437 on Build GRCh38
rs778098370 Chr.9: 114094469-114094469 on Build GRCh38
rs746361323 Chr.9: 114094252-114094252 on Build GRCh38
rs758713491 Chr.9: 114094326-114094326 on Build GRCh38
rs371195261 Chr.9: 114094277-114094277 on Build GRCh38
rs773967379 Chr.9: 114094651-114094651 on Build GRCh38
rs770660521 Chr.9: 114094384-114094384 on Build GRCh38
rs202004050 Chr.9: 114094284-114094284 on Build GRCh38
rs766279222 Chr.9: 114094289-114094289 on Build GRCh38
rs201028865 Chr.9: 114094264-114094264 on Build GRCh38
rs1021273702 Chr.9: 114094254-114094254 on Build GRCh38
rs370280229 Chr.9: 114094391-114094391 on Build GRCh38
rs780572486 Chr.9: 114094345-114094345 on Build GRCh38
rs944475537 Chr.9: 114094673-114094673 on Build GRCh38
rs559933389 Chr.9: 114094267-114094267 on Build GRCh38
rs140268348 Chr.9: 114094554-114094554 on Build GRCh38
rs367842026 Chr.9: 114094524-114094524 on Build GRCh38
rs752375951 Chr.9: 114094248-114094248 on Build GRCh38
rs772563682 Chr.9: 114094258-114094258 on Build GRCh38
rs755506925 Chr.9: 114094450-114094450 on Build GRCh38
rs374214880 Chr.9: 114094495-114094495 on Build GRCh38
rs769595544 Chr.9: 114094532-114094532 on Build GRCh38
rs750729178 Chr.9: 114094321-114094321 on Build GRCh38
rs745813229 Chr.9: 114094476-114094476 on Build GRCh38
rs547922910 Chr.9: 114094392-114094392 on Build GRCh38
rs774175398 Chr.9: 114094308-114094308 on Build GRCh38
rs535919981 Chr.9: 114094644-114094644 on Build GRCh38
rs776326702 Chr.9: 114094494-114094494 on Build GRCh38
rs964547824 Chr.9: 114094304-114094304 on Build GRCh38
rs376905379 Chr.9: 114094388-114094388 on Build GRCh38
rs963024513 Chr.9: 114094614-114094614 on Build GRCh38
rs138603268 Chr.9: 114094364-114094364 on Build GRCh38
rs1017567142 Chr.9: 114094528-114094528 on Build GRCh38
rs746866762 Chr.9: 114094319-114094319 on Build GRCh38
rs749510835 Chr.9: 114094471-114094471 on Build GRCh38
rs377283601 Chr.9: 114094498-114094498 on Build GRCh38
rs927590494 Chr.9: 114094630-114094630 on Build GRCh38
rs754157551 Chr.9: 114094489-114094490 on Build GRCh38
rs149645876 Chr.9: 114094260-114094260 on Build GRCh38
rs374557934 Chr.9: 114094430-114094430 on Build GRCh38
rs182844693 Chr.9: 114094346-114094346 on Build GRCh38
rs570211051 Chr.9: 114094589-114094589 on Build GRCh38
rs554157552 Chr.9: 114094478-114094478 on Build GRCh38
rs145112002 Chr.9: 114094566-114094566 on Build GRCh38
rs766705828 Chr.9: 114094435-114094435 on Build GRCh38
rs778047024 Chr.9: 114094501-114094501 on Build GRCh38
rs771939585 Chr.9: 114094477-114094477 on Build GRCh38
rs745707764 Chr.9: 114094488-114094488 on Build GRCh38
rs758371889 Chr.9: 114094251-114094251 on Build GRCh38
rs774071073 Chr.9: 114094475-114094475 on Build GRCh38
rs375694046 Chr.9: 114094482-114094482 on Build GRCh38
rs930530389 Chr.9: 114094616-114094616 on Build GRCh38
rs760191734 Chr.9: 114094400-114094400 on Build GRCh38
rs1050179159 Chr.9: 114094413-114094413 on Build GRCh38
rs781574221 Chr.9: 114094355-114094355 on Build GRCh38
rs752095690 Chr.9: 114094440-114094440 on Build GRCh38
rs561028103 Chr.9: 114094250-114094250 on Build GRCh38
rs753521320 Chr.9: 114094431-114094431 on Build GRCh38
rs368096781 Chr.9: 114094474-114094474 on Build GRCh38
rs768196263 Chr.9: 114094487-114094487 on Build GRCh38
rs908882914 Chr.9: 114094408-114094408 on Build GRCh38
rs571139806 Chr.9: 114094522-114094522 on Build GRCh38
rs751808338 Chr.9: 114094496-114094496 on Build GRCh38
rs148793055 Chr.9: 114094375-114094375 on Build GRCh38
rs372259818 Chr.9: 114094479-114094479 on Build GRCh38
rs766974860 Chr.9: 114094312-114094312 on Build GRCh38
rs775047921 Chr.9: 114094396-114094396 on Build GRCh38
rs201033769 Chr.9: 114094279-114094279 on Build GRCh38
rs35634077 Chr.9: 114094521-114094522 on Build GRCh38
rs780439251 Chr.9: 114094259-114094259 on Build GRCh38
rs377685500 Chr.9: 114094245-114094245 on Build GRCh38
rs763741985 Chr.9: 114094317-114094317 on Build GRCh38
rs940030565 Chr.9: 114094599-114094599 on Build GRCh38
rs754076236 Chr.9: 114094502-114094502 on Build GRCh38
rs753284275 Chr.9: 114094499-114094499 on Build GRCh38
rs371123919 Chr.9: 114094452-114094452 on Build GRCh38
rs748381825 Chr.9: 114094373-114094373 on Build GRCh38
rs113114344 Chr.9: 114094602-114094602 on Build GRCh38

View Full Product Details