Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121302953-121303448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSN
Entrez Gene ID
2934
Gene Name
gelsolin
Gene Aliases
ADF, AGEL
UniGene
Hs.522373
Gene Chromosome Location
Chr.9: 121201082-121332844 on Build GRCh38
Transcripts
NM_000177.4 NM_001127662.1 NM_001127663.1 NM_001127664.1 NM_001127665.1 NM_001127666.1 NM_001127667.1 NM_001258029.1 NM_001258030.1 NM_198252.2 XM_005251943.1 XM_005251944.1 XM_005251945.3 XM_006717079.1 XM_011518584.1 XM_011518585.1 XM_011518586.1 XM_011518587.2 XM_011518588.2 XM_011518589.2 XM_011518590.2 XM_011518591.2 XM_011518592.1 XM_011518593.1 XM_017014643.1 XM_017014644.1 XM_017014645.1 XM_017014646.1 XM_017014647.1 XM_017014648.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6676517 c.365A>G p.K122R Chr.9: 121303046-121303046 on Build GRCh38
3847549 c.351+2T>G p.? Chr.9: 121303067-121303067 on Build GRCh38
6676518 c.332A>G p.K111R Chr.9: 121303046-121303046 on Build GRCh38
1173440 c.308C>A p.S103* Chr.9: 121303022-121303022 on Build GRCh38
3653800 c.243C>T p.T81T Chr.9: 121302957-121302957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895304313 Chr.9: 121303018-121303018 on Build GRCh38
rs919764789 Chr.9: 121303201-121303201 on Build GRCh38
rs1047053570 Chr.9: 121303217-121303217 on Build GRCh38
rs938357762 Chr.9: 121303273-121303273 on Build GRCh38
rs777396943 Chr.9: 121302980-121302980 on Build GRCh38
rs759334030 Chr.9: 121303019-121303019 on Build GRCh38
rs936534111 Chr.9: 121303389-121303390 on Build GRCh38
rs929873453 Chr.9: 121303203-121303203 on Build GRCh38
rs750877079 Chr.9: 121302982-121302982 on Build GRCh38
rs763868927 Chr.9: 121303169-121303169 on Build GRCh38
rs770917949 Chr.9: 121303097-121303097 on Build GRCh38
rs904022394 Chr.9: 121303388-121303388 on Build GRCh38
rs745854588 Chr.9: 121303109-121303109 on Build GRCh38
rs781594640 Chr.9: 121303080-121303080 on Build GRCh38
rs367933536 Chr.9: 121303036-121303036 on Build GRCh38
rs142155964 Chr.9: 121302999-121302999 on Build GRCh38
rs763484567 Chr.9: 121303052-121303052 on Build GRCh38
rs879733222 Chr.9: 121303281-121303281 on Build GRCh38
rs373779982 Chr.9: 121303017-121303017 on Build GRCh38
rs577093505 Chr.9: 121303130-121303130 on Build GRCh38
rs752067269 Chr.9: 121303070-121303070 on Build GRCh38
rs918362584 Chr.9: 121303179-121303179 on Build GRCh38
rs12685309 Chr.9: 121303238-121303238 on Build GRCh38
rs201920706 Chr.9: 121303001-121303001 on Build GRCh38
rs766817635 Chr.9: 121303068-121303068 on Build GRCh38
rs138153246 Chr.9: 121302986-121302986 on Build GRCh38
rs758988443 Chr.9: 121302983-121302983 on Build GRCh38
rs561296617 Chr.9: 121303207-121303207 on Build GRCh38
rs147841943 Chr.9: 121303138-121303138 on Build GRCh38
rs569552248 Chr.9: 121303092-121303092 on Build GRCh38
rs1052209391 Chr.9: 121303426-121303426 on Build GRCh38
rs753352592 Chr.9: 121303054-121303054 on Build GRCh38
rs139793292 Chr.9: 121303156-121303156 on Build GRCh38
rs752380492 Chr.9: 121303023-121303023 on Build GRCh38
rs146956976 Chr.9: 121302985-121302985 on Build GRCh38
rs866179533 Chr.9: 121303000-121303000 on Build GRCh38
rs368858384 Chr.9: 121303411-121303411 on Build GRCh38
rs965919012 Chr.9: 121302981-121302981 on Build GRCh38
rs962881810 Chr.9: 121303139-121303139 on Build GRCh38
rs548138013 Chr.9: 121302990-121302990 on Build GRCh38
rs537136378 Chr.9: 121303093-121303093 on Build GRCh38
rs1011755922 Chr.9: 121303356-121303356 on Build GRCh38
rs760357425 Chr.9: 121302991-121302991 on Build GRCh38
rs541762629 Chr.9: 121303187-121303187 on Build GRCh38
rs942508061 Chr.9: 121303144-121303144 on Build GRCh38
rs1000625882 Chr.9: 121303063-121303063 on Build GRCh38
rs767484600 Chr.9: 121303022-121303022 on Build GRCh38
rs749329655 Chr.9: 121303095-121303095 on Build GRCh38
rs756743284 Chr.9: 121303065-121303065 on Build GRCh38
rs774424025 Chr.9: 121303107-121303107 on Build GRCh38
rs558734499 Chr.9: 121303113-121303113 on Build GRCh38
rs770851113 Chr.9: 121303005-121303005 on Build GRCh38
rs865879074 Chr.9: 121303024-121303024 on Build GRCh38
rs903705821 Chr.9: 121303044-121303044 on Build GRCh38
rs565065403 Chr.9: 121303254-121303254 on Build GRCh38
rs777879692 Chr.9: 121303094-121303094 on Build GRCh38
rs1055535153 Chr.9: 121303316-121303316 on Build GRCh38
rs894201378 Chr.9: 121303340-121303340 on Build GRCh38
rs763755977 Chr.9: 121303115-121303115 on Build GRCh38
rs903756850 Chr.9: 121303385-121303385 on Build GRCh38
rs576398568 Chr.9: 121303230-121303230 on Build GRCh38
rs755334053 Chr.9: 121303078-121303078 on Build GRCh38
rs781506692 Chr.9: 121302982-121302982 on Build GRCh38
rs776163774 Chr.9: 121303116-121303116 on Build GRCh38
rs149721923 Chr.9: 121303132-121303132 on Build GRCh38
rs866707411 Chr.9: 121303163-121303163 on Build GRCh38
rs532551035 Chr.9: 121303360-121303360 on Build GRCh38

View Full Product Details