Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15786166-15786662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA5B
Entrez Gene ID
11238
Gene Name
carbonic anhydrase 5B
Gene Aliases
CA-VB
UniGene
Hs.653287
Gene Chromosome Location
Chr.X: 15738281-15787625 on Build GRCh38
Transcripts
NM_007220.3 NR_002725.2

Gene Symbol
INE2
Entrez Gene ID
8551
Gene Name
inactivation escape 2 (non-protein coding)
Gene Aliases
NCRNA00011
UniGene
Hs.653287
Gene Chromosome Location
Chr.X: 15785716-15787589 on Build GRCh38
Transcripts
NM_007220.3 NR_002725.2

SNP Detail

SNP ID Chromosome Location
rs746207669 Chr.X: 15786326-15786326 on Build GRCh38
rs768930282 Chr.X: 15786444-15786444 on Build GRCh38
rs769944208 Chr.X: 15786614-15786614 on Build GRCh38
rs768769324 Chr.X: 15786408-15786408 on Build GRCh38
rs868453336 Chr.X: 15786275-15786275 on Build GRCh38
rs954848553 Chr.X: 15786624-15786624 on Build GRCh38
rs930087284 Chr.X: 15786520-15786520 on Build GRCh38
rs983271674 Chr.X: 15786530-15786530 on Build GRCh38
rs180710329 Chr.X: 15786385-15786385 on Build GRCh38
rs200213506 Chr.X: 15786261-15786261 on Build GRCh38
rs917097958 Chr.X: 15786584-15786584 on Build GRCh38
rs949940689 Chr.X: 15786603-15786603 on Build GRCh38
rs143459956 Chr.X: 15786237-15786237 on Build GRCh38
rs867984688 Chr.X: 15786263-15786263 on Build GRCh38
rs996401836 Chr.X: 15786594-15786594 on Build GRCh38
rs35824487 Chr.X: 15786263-15786264 on Build GRCh38
rs900756998 Chr.X: 15786585-15786585 on Build GRCh38
rs774382129 Chr.X: 15786463-15786463 on Build GRCh38
rs770055368 Chr.X: 15786344-15786344 on Build GRCh38
rs879195287 Chr.X: 15786617-15786617 on Build GRCh38
rs949973140 Chr.X: 15786222-15786222 on Build GRCh38
rs1032741813 Chr.X: 15786307-15786307 on Build GRCh38
rs757821576 Chr.X: 15786278-15786278 on Build GRCh38
rs1006631505 Chr.X: 15786500-15786500 on Build GRCh38
rs918676736 Chr.X: 15786482-15786482 on Build GRCh38
rs773025077 Chr.X: 15786617-15786618 on Build GRCh38
rs955675404 Chr.X: 15786220-15786220 on Build GRCh38
rs999013647 Chr.X: 15786424-15786424 on Build GRCh38
rs190910008 Chr.X: 15786453-15786453 on Build GRCh38
rs1128070 Chr.X: 15786392-15786392 on Build GRCh38
rs1030520759 Chr.X: 15786617-15786617 on Build GRCh38
rs974504084 Chr.X: 15786481-15786481 on Build GRCh38
rs185958020 Chr.X: 15786399-15786399 on Build GRCh38
rs183419468 Chr.X: 15786469-15786469 on Build GRCh38
rs189749353 Chr.X: 15786316-15786316 on Build GRCh38

View Full Product Details