Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15786395-15786890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA5B
Entrez Gene ID
11238
Gene Name
carbonic anhydrase 5B
Gene Aliases
CA-VB
UniGene
Hs.653287
Gene Chromosome Location
Chr.X: 15738281-15787625 on Build GRCh38
Transcripts
NM_007220.3 NR_002725.2

Gene Symbol
INE2
Entrez Gene ID
8551
Gene Name
inactivation escape 2 (non-protein coding)
Gene Aliases
NCRNA00011
UniGene
Hs.653287
Gene Chromosome Location
Chr.X: 15785716-15787589 on Build GRCh38
Transcripts
NM_007220.3 NR_002725.2

SNP Detail

SNP ID Chromosome Location
rs1031424159 Chr.X: 15786662-15786662 on Build GRCh38
rs1054561228 Chr.X: 15786815-15786815 on Build GRCh38
rs769944208 Chr.X: 15786614-15786614 on Build GRCh38
rs768930282 Chr.X: 15786444-15786444 on Build GRCh38
rs66690979 Chr.X: 15786655-15786658 on Build GRCh38
rs954848553 Chr.X: 15786624-15786624 on Build GRCh38
rs983271674 Chr.X: 15786530-15786530 on Build GRCh38
rs930087284 Chr.X: 15786520-15786520 on Build GRCh38
rs917097958 Chr.X: 15786584-15786584 on Build GRCh38
rs369120363 Chr.X: 15786801-15786801 on Build GRCh38
rs949940689 Chr.X: 15786603-15786603 on Build GRCh38
rs867165030 Chr.X: 15786655-15786655 on Build GRCh38
rs867426947 Chr.X: 15786656-15786656 on Build GRCh38
rs1008006002 Chr.X: 15786644-15786644 on Build GRCh38
rs970398105 Chr.X: 15786735-15786735 on Build GRCh38
rs980014030 Chr.X: 15786780-15786780 on Build GRCh38
rs996401836 Chr.X: 15786594-15786594 on Build GRCh38
rs900756998 Chr.X: 15786585-15786585 on Build GRCh38
rs955767248 Chr.X: 15786675-15786675 on Build GRCh38
rs774382129 Chr.X: 15786463-15786463 on Build GRCh38
rs879195287 Chr.X: 15786617-15786617 on Build GRCh38
rs57459975 Chr.X: 15786860-15786860 on Build GRCh38
rs35851144 Chr.X: 15786649-15786652 on Build GRCh38
rs773052791 Chr.X: 15786703-15786703 on Build GRCh38
rs1006631505 Chr.X: 15786500-15786500 on Build GRCh38
rs760461936 Chr.X: 15786709-15786709 on Build GRCh38
rs773025077 Chr.X: 15786617-15786618 on Build GRCh38
rs918676736 Chr.X: 15786482-15786482 on Build GRCh38
rs999013647 Chr.X: 15786424-15786424 on Build GRCh38
rs10521650 Chr.X: 15786861-15786861 on Build GRCh38
rs190910008 Chr.X: 15786453-15786453 on Build GRCh38
rs1030520759 Chr.X: 15786617-15786617 on Build GRCh38
rs754647407 Chr.X: 15786863-15786863 on Build GRCh38
rs974504084 Chr.X: 15786481-15786481 on Build GRCh38
rs914203939 Chr.X: 15786718-15786718 on Build GRCh38
rs183419468 Chr.X: 15786469-15786469 on Build GRCh38

View Full Product Details