Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 29917386-29917841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1RAPL1
Entrez Gene ID
11141
Gene Name
interleukin 1 receptor accessory protein like 1
Gene Aliases
IL1R8, IL1RAPL, MRX10, MRX21, MRX34, OPHN4, TIGIRR-2
UniGene
Hs.658912
Gene Chromosome Location
Chr.X: 28587564-29956350 on Build GRCh38
Transcripts
NM_014271.3 XM_017029240.1 XM_017029241.1

SNP Detail

SNP ID Chromosome Location
rs182138680 Chr.X: 29917531-29917531 on Build GRCh38
rs753934234 Chr.X: 29917433-29917433 on Build GRCh38
rs980119982 Chr.X: 29917767-29917767 on Build GRCh38
rs935357332 Chr.X: 29917816-29917816 on Build GRCh38
rs765317365 Chr.X: 29917601-29917601 on Build GRCh38
rs751803618 Chr.X: 29917727-29917730 on Build GRCh38
rs925549434 Chr.X: 29917786-29917786 on Build GRCh38
rs111626723 Chr.X: 29917734-29917734 on Build GRCh38
rs200700252 Chr.X: 29917456-29917456 on Build GRCh38
rs375172685 Chr.X: 29917420-29917420 on Build GRCh38
rs749331045 Chr.X: 29917510-29917510 on Build GRCh38
rs760355758 Chr.X: 29917602-29917602 on Build GRCh38
rs772466086 Chr.X: 29917589-29917589 on Build GRCh38
rs1016840091 Chr.X: 29917501-29917501 on Build GRCh38
rs763691271 Chr.X: 29917617-29917617 on Build GRCh38
rs962728005 Chr.X: 29917507-29917507 on Build GRCh38
rs1025225413 Chr.X: 29917541-29917541 on Build GRCh38
rs781329082 Chr.X: 29917440-29917440 on Build GRCh38
rs201842225 Chr.X: 29917553-29917553 on Build GRCh38
rs942864168 Chr.X: 29917758-29917758 on Build GRCh38
rs905665357 Chr.X: 29917432-29917432 on Build GRCh38
rs758156048 Chr.X: 29917434-29917434 on Build GRCh38
rs878853146 Chr.X: 29917579-29917588 on Build GRCh38
rs775048888 Chr.X: 29917592-29917592 on Build GRCh38
rs748396756 Chr.X: 29917453-29917453 on Build GRCh38
rs747371577 Chr.X: 29917678-29917678 on Build GRCh38
rs1032419964 Chr.X: 29917655-29917655 on Build GRCh38
rs778785634 Chr.X: 29917422-29917422 on Build GRCh38
rs753825085 Chr.X: 29917418-29917418 on Build GRCh38
rs955134038 Chr.X: 29917544-29917544 on Build GRCh38
rs774927191 Chr.X: 29917542-29917542 on Build GRCh38
rs988295710 Chr.X: 29917809-29917809 on Build GRCh38
rs150439009 Chr.X: 29917469-29917469 on Build GRCh38
rs761246673 Chr.X: 29917646-29917646 on Build GRCh38
rs753380346 Chr.X: 29917642-29917642 on Build GRCh38
rs758709777 Chr.X: 29917600-29917601 on Build GRCh38
rs1000964966 Chr.X: 29917603-29917603 on Build GRCh38
rs778228325 Chr.X: 29917674-29917675 on Build GRCh38
rs777978404 Chr.X: 29917508-29917508 on Build GRCh38

View Full Product Details