Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 29919748-29920247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1RAPL1
Entrez Gene ID
11141
Gene Name
interleukin 1 receptor accessory protein like 1
Gene Aliases
IL1R8, IL1RAPL, MRX10, MRX21, MRX34, OPHN4, TIGIRR-2
UniGene
Hs.658912
Gene Chromosome Location
Chr.X: 28587564-29956350 on Build GRCh38
Transcripts
NM_014271.3 XM_017029240.1 XM_017029241.1

SNP Detail

SNP ID Chromosome Location
rs761773742 Chr.X: 29919902-29919903 on Build GRCh38
rs183925588 Chr.X: 29920084-29920084 on Build GRCh38
rs375016829 Chr.X: 29919905-29919905 on Build GRCh38
rs899519363 Chr.X: 29919779-29919779 on Build GRCh38
rs371275154 Chr.X: 29920072-29920072 on Build GRCh38
rs1018206304 Chr.X: 29919877-29919877 on Build GRCh38
rs749380359 Chr.X: 29920070-29920070 on Build GRCh38
rs770839799 Chr.X: 29920074-29920074 on Build GRCh38
rs778100377 Chr.X: 29920067-29920067 on Build GRCh38
rs1026232210 Chr.X: 29920217-29920217 on Build GRCh38
rs758196137 Chr.X: 29919957-29919957 on Build GRCh38
rs970231054 Chr.X: 29920153-29920153 on Build GRCh38
rs768677816 Chr.X: 29920215-29920216 on Build GRCh38
rs750272193 Chr.X: 29920144-29920145 on Build GRCh38
rs767414860 Chr.X: 29919910-29919913 on Build GRCh38
rs200419221 Chr.X: 29920076-29920076 on Build GRCh38
rs995057356 Chr.X: 29920061-29920061 on Build GRCh38
rs899290884 Chr.X: 29919880-29919880 on Build GRCh38
rs765172636 Chr.X: 29919903-29919903 on Build GRCh38
rs199971765 Chr.X: 29920063-29920063 on Build GRCh38
rs779866333 Chr.X: 29920062-29920062 on Build GRCh38
rs1009288428 Chr.X: 29919929-29919929 on Build GRCh38
rs111532996 Chr.X: 29920154-29920154 on Build GRCh38
rs746852113 Chr.X: 29919872-29919872 on Build GRCh38
rs756297239 Chr.X: 29920188-29920188 on Build GRCh38
rs372256169 Chr.X: 29920118-29920118 on Build GRCh38
rs979839151 Chr.X: 29920194-29920194 on Build GRCh38
rs756426427 Chr.X: 29920065-29920065 on Build GRCh38
rs369557817 Chr.X: 29920075-29920075 on Build GRCh38
rs747300297 Chr.X: 29920092-29920092 on Build GRCh38
rs749383705 Chr.X: 29920117-29920117 on Build GRCh38
rs374844839 Chr.X: 29920135-29920135 on Build GRCh38
rs936318591 Chr.X: 29919809-29919809 on Build GRCh38
rs777509099 Chr.X: 29919854-29919854 on Build GRCh38
rs772374800 Chr.X: 29920079-29920079 on Build GRCh38
rs963444717 Chr.X: 29920133-29920133 on Build GRCh38
rs764725197 Chr.X: 29920142-29920142 on Build GRCh38

View Full Product Details