Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 180935682-180936185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1614
Entrez Gene ID
57710
Gene Name
KIAA1614
Gene Aliases
-
UniGene
Hs.734816
Gene Chromosome Location
Chr.1: 180913177-180946103 on Build GRCh38
Transcripts
NM_020950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6122952 c.2179C>A p.P727T Chr.1: 180936088-180936088 on Build GRCh38
4475609 c.2002C>T p.P668S Chr.1: 180935911-180935911 on Build GRCh38
7169824 c.2266G>A p.E756K Chr.1: 180936175-180936175 on Build GRCh38
7169823 c.1937G>A p.R646Q Chr.1: 180935846-180935846 on Build GRCh38
5632989 c.2101C>G p.L701V Chr.1: 180936010-180936010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751648938 Chr.1: 180935792-180935792 on Build GRCh38
rs368068411 Chr.1: 180936105-180936105 on Build GRCh38
rs751777199 Chr.1: 180935866-180935866 on Build GRCh38
rs1028544697 Chr.1: 180936026-180936026 on Build GRCh38
rs563066623 Chr.1: 180935806-180935806 on Build GRCh38
rs757002043 Chr.1: 180936069-180936069 on Build GRCh38
rs750892476 Chr.1: 180935801-180935801 on Build GRCh38
rs759442139 Chr.1: 180935777-180935777 on Build GRCh38
rs757771254 Chr.1: 180935723-180935723 on Build GRCh38
rs780530111 Chr.1: 180935914-180935914 on Build GRCh38
rs970501082 Chr.1: 180936110-180936110 on Build GRCh38
rs761873697 Chr.1: 180935721-180935722 on Build GRCh38
rs202196321 Chr.1: 180935788-180935788 on Build GRCh38
rs764273570 Chr.1: 180935863-180935863 on Build GRCh38
rs750064062 Chr.1: 180936165-180936165 on Build GRCh38
rs546298540 Chr.1: 180935886-180935886 on Build GRCh38
rs762591781 Chr.1: 180935843-180935844 on Build GRCh38
rs778570803 Chr.1: 180935730-180935730 on Build GRCh38
rs777025822 Chr.1: 180936139-180936139 on Build GRCh38
rs766692333 Chr.1: 180935904-180935904 on Build GRCh38
rs780174951 Chr.1: 180936085-180936085 on Build GRCh38
rs369125012 Chr.1: 180935925-180935925 on Build GRCh38
rs760727456 Chr.1: 180936022-180936022 on Build GRCh38
rs774463895 Chr.1: 180935990-180935990 on Build GRCh38
rs776463337 Chr.1: 180935843-180935843 on Build GRCh38
rs746632843 Chr.1: 180935931-180935931 on Build GRCh38
rs765082260 Chr.1: 180935779-180935779 on Build GRCh38
rs769912495 Chr.1: 180935952-180935952 on Build GRCh38
rs758670383 Chr.1: 180936160-180936160 on Build GRCh38
rs999362518 Chr.1: 180935969-180935969 on Build GRCh38
rs754310661 Chr.1: 180936042-180936042 on Build GRCh38
rs966574947 Chr.1: 180935757-180935757 on Build GRCh38
rs56104489 Chr.1: 180935976-180935976 on Build GRCh38
rs747558624 Chr.1: 180935719-180935719 on Build GRCh38
rs775548600 Chr.1: 180935970-180935970 on Build GRCh38
rs760224801 Chr.1: 180935746-180935746 on Build GRCh38
rs200170063 Chr.1: 180935732-180935732 on Build GRCh38
rs376947197 Chr.1: 180936128-180936128 on Build GRCh38
rs751184262 Chr.1: 180936067-180936067 on Build GRCh38
rs747888190 Chr.1: 180936116-180936116 on Build GRCh38
rs376112299 Chr.1: 180935997-180935997 on Build GRCh38
rs998764593 Chr.1: 180936008-180936008 on Build GRCh38
rs745368430 Chr.1: 180935947-180935947 on Build GRCh38
rs766197445 Chr.1: 180935706-180935706 on Build GRCh38
rs746548304 Chr.1: 180935822-180935822 on Build GRCh38
rs773156409 Chr.1: 180935762-180935763 on Build GRCh38
rs942249479 Chr.1: 180935893-180935893 on Build GRCh38
rs752981786 Chr.1: 180935782-180935782 on Build GRCh38
rs767589413 Chr.1: 180935874-180935874 on Build GRCh38
rs752934576 Chr.1: 180935715-180935715 on Build GRCh38
rs768353072 Chr.1: 180936090-180936090 on Build GRCh38
rs17302207 Chr.1: 180936127-180936127 on Build GRCh38
rs772334197 Chr.1: 180936126-180936126 on Build GRCh38
rs745421836 Chr.1: 180936082-180936082 on Build GRCh38
rs769806195 Chr.1: 180935754-180935755 on Build GRCh38
rs113108579 Chr.1: 180935816-180935816 on Build GRCh38
rs781716381 Chr.1: 180935726-180935726 on Build GRCh38
rs757341832 Chr.1: 180935796-180935796 on Build GRCh38
rs772527816 Chr.1: 180935744-180935744 on Build GRCh38
rs762828590 Chr.1: 180935858-180935858 on Build GRCh38
rs754257262 Chr.1: 180935916-180935916 on Build GRCh38
rs909402780 Chr.1: 180936159-180936159 on Build GRCh38
rs188274594 Chr.1: 180935815-180935815 on Build GRCh38
rs577489860 Chr.1: 180936084-180936084 on Build GRCh38
rs776608636 Chr.1: 180935770-180935770 on Build GRCh38
rs749079647 Chr.1: 180935980-180935980 on Build GRCh38
rs958155391 Chr.1: 180935775-180935775 on Build GRCh38
rs777658675 Chr.1: 180935716-180935716 on Build GRCh38
rs747801791 Chr.1: 180935813-180935813 on Build GRCh38
rs781594767 Chr.1: 180935798-180935798 on Build GRCh38
rs765588296 Chr.1: 180936064-180936064 on Build GRCh38
rs372096628 Chr.1: 180935903-180935903 on Build GRCh38
rs773342548 Chr.1: 180936016-180936016 on Build GRCh38
rs368738392 Chr.1: 180935733-180935733 on Build GRCh38
rs149910072 Chr.1: 180935920-180935920 on Build GRCh38
rs777522200 Chr.1: 180936161-180936161 on Build GRCh38
rs373192353 Chr.1: 180935951-180935951 on Build GRCh38
rs1047849189 Chr.1: 180935870-180935870 on Build GRCh38
rs566865809 Chr.1: 180935996-180935996 on Build GRCh38
rs965614040 Chr.1: 180936007-180936007 on Build GRCh38
rs371122535 Chr.1: 180936056-180936056 on Build GRCh38
rs766917985 Chr.1: 180936036-180936036 on Build GRCh38
rs528128697 Chr.1: 180935876-180935876 on Build GRCh38
rs769718089 Chr.1: 180935854-180935854 on Build GRCh38
rs79791475 Chr.1: 180936158-180936158 on Build GRCh38
rs548264420 Chr.1: 180935989-180935989 on Build GRCh38
rs865999112 Chr.1: 180935896-180935896 on Build GRCh38
rs182446414 Chr.1: 180935785-180935785 on Build GRCh38
rs375009931 Chr.1: 180935919-180935919 on Build GRCh38
rs909466136 Chr.1: 180935872-180935872 on Build GRCh38
rs761832211 Chr.1: 180935873-180935873 on Build GRCh38
rs372602361 Chr.1: 180935904-180935904 on Build GRCh38
rs139066333 Chr.1: 180935731-180935731 on Build GRCh38
rs774132007 Chr.1: 180936115-180936115 on Build GRCh38
rs377007881 Chr.1: 180935727-180935727 on Build GRCh38
rs950760800 Chr.1: 180936131-180936131 on Build GRCh38
rs760030556 Chr.1: 180936149-180936149 on Build GRCh38
rs953039218 Chr.1: 180935761-180935761 on Build GRCh38
rs192741655 Chr.1: 180935769-180935769 on Build GRCh38
rs1039276178 Chr.1: 180935907-180935907 on Build GRCh38
rs749202852 Chr.1: 180936086-180936086 on Build GRCh38
rs754806755 Chr.1: 180935707-180935707 on Build GRCh38
rs929507879 Chr.1: 180935846-180935846 on Build GRCh38
rs554099630 Chr.1: 180936153-180936153 on Build GRCh38
rs536565441 Chr.1: 180935940-180935940 on Build GRCh38
rs368421393 Chr.1: 180935832-180935832 on Build GRCh38
rs559138596 Chr.1: 180936072-180936072 on Build GRCh38
rs985741506 Chr.1: 180936040-180936040 on Build GRCh38
rs765605749 Chr.1: 180935714-180935714 on Build GRCh38
rs775495512 Chr.1: 180935856-180935856 on Build GRCh38
rs544916854 Chr.1: 180935802-180935802 on Build GRCh38

View Full Product Details