Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134473319-134473867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPRT1
Entrez Gene ID
3251
Gene Name
hypoxanthine phosphoribosyltransferase 1
Gene Aliases
HGPRT, HPRT
UniGene
Hs.412707
Gene Chromosome Location
Chr.X: 134460145-134500668 on Build GRCh38
Transcripts
NM_000194.2

SNP Detail

SNP ID Chromosome Location
rs374646638 Chr.X: 134473385-134473385 on Build GRCh38
rs941357288 Chr.X: 134473753-134473753 on Build GRCh38
rs765096233 Chr.X: 134473776-134473776 on Build GRCh38
rs751314237 Chr.X: 134473507-134473507 on Build GRCh38
rs759587825 Chr.X: 134473717-134473717 on Build GRCh38
rs905777674 Chr.X: 134473820-134473820 on Build GRCh38
rs762739119 Chr.X: 134473408-134473408 on Build GRCh38
rs763953406 Chr.X: 134473483-134473483 on Build GRCh38
rs940105641 Chr.X: 134473692-134473692 on Build GRCh38
rs191993127 Chr.X: 134473816-134473816 on Build GRCh38
rs754813142 Chr.X: 134473345-134473345 on Build GRCh38
rs888579006 Chr.X: 134473613-134473613 on Build GRCh38
rs556008472 Chr.X: 134473810-134473810 on Build GRCh38
rs1049031592 Chr.X: 134473592-134473592 on Build GRCh38
rs747906552 Chr.X: 134473488-134473489 on Build GRCh38
rs752560279 Chr.X: 134473804-134473804 on Build GRCh38
rs137852480 Chr.X: 134473453-134473453 on Build GRCh38
rs1014187553 Chr.X: 134473798-134473798 on Build GRCh38
rs137852499 Chr.X: 134473377-134473377 on Build GRCh38
rs987346132 Chr.X: 134473653-134473653 on Build GRCh38
rs137852491 Chr.X: 134473465-134473465 on Build GRCh38
rs753709252 Chr.X: 134473608-134473608 on Build GRCh38
rs955544279 Chr.X: 134473546-134473546 on Build GRCh38
rs17882387 Chr.X: 134473540-134473540 on Build GRCh38
rs756925981 Chr.X: 134473510-134473510 on Build GRCh38
rs932836425 Chr.X: 134473563-134473563 on Build GRCh38
rs896549609 Chr.X: 134473823-134473823 on Build GRCh38
rs1037087476 Chr.X: 134473715-134473715 on Build GRCh38
rs944325300 Chr.X: 134473359-134473359 on Build GRCh38
rs921498963 Chr.X: 134473474-134473474 on Build GRCh38
rs794727238 Chr.X: 134473471-134473474 on Build GRCh38
rs940228019 Chr.X: 134473751-134473751 on Build GRCh38
rs867788976 Chr.X: 134473344-134473344 on Build GRCh38
rs908801873 Chr.X: 134473696-134473696 on Build GRCh38

View Full Product Details