Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200971248-200971746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF21B
Entrez Gene ID
23046
Gene Name
kinesin family member 21B
Gene Aliases
-
UniGene
Hs.169182
Gene Chromosome Location
Chr.1: 200969386-201023700 on Build GRCh38
Transcripts
NM_001252100.1 NM_001252102.1 NM_001252103.1 NM_017596.3

SNP Detail

SNP ID Chromosome Location
rs990844794 Chr.1: 200971288-200971288 on Build GRCh38
rs911395912 Chr.1: 200971506-200971506 on Build GRCh38
rs897175111 Chr.1: 200971427-200971427 on Build GRCh38
rs760383090 Chr.1: 200971609-200971609 on Build GRCh38
rs1035173964 Chr.1: 200971464-200971465 on Build GRCh38
rs535929945 Chr.1: 200971485-200971485 on Build GRCh38
rs886358384 Chr.1: 200971382-200971382 on Build GRCh38
rs1022768123 Chr.1: 200971619-200971619 on Build GRCh38
rs892840177 Chr.1: 200971409-200971409 on Build GRCh38
rs528791081 Chr.1: 200971401-200971401 on Build GRCh38
rs58664533 Chr.1: 200971599-200971599 on Build GRCh38
rs969966765 Chr.1: 200971442-200971442 on Build GRCh38
rs914729743 Chr.1: 200971339-200971339 on Build GRCh38
rs960847018 Chr.1: 200971473-200971473 on Build GRCh38
rs762412279 Chr.1: 200971359-200971359 on Build GRCh38
rs1041716075 Chr.1: 200971553-200971553 on Build GRCh38
rs988544107 Chr.1: 200971319-200971319 on Build GRCh38
rs967376835 Chr.1: 200971441-200971441 on Build GRCh38
rs1006092164 Chr.1: 200971437-200971437 on Build GRCh38
rs940608762 Chr.1: 200971556-200971556 on Build GRCh38
rs990898823 Chr.1: 200971491-200971491 on Build GRCh38
rs985206296 Chr.1: 200971508-200971508 on Build GRCh38
rs887600678 Chr.1: 200971436-200971436 on Build GRCh38
rs750120496 Chr.1: 200971570-200971570 on Build GRCh38
rs1011296122 Chr.1: 200971651-200971651 on Build GRCh38
rs981777998 Chr.1: 200971444-200971444 on Build GRCh38
rs1012752086 Chr.1: 200971618-200971618 on Build GRCh38
rs766891985 Chr.1: 200971687-200971687 on Build GRCh38
rs955734584 Chr.1: 200971482-200971482 on Build GRCh38
rs1041023273 Chr.1: 200971423-200971423 on Build GRCh38
rs1048506139 Chr.1: 200971598-200971598 on Build GRCh38
rs567297032 Chr.1: 200971474-200971474 on Build GRCh38
rs994231803 Chr.1: 200971431-200971431 on Build GRCh38
rs569725516 Chr.1: 200971507-200971507 on Build GRCh38
rs569217817 Chr.1: 200971433-200971433 on Build GRCh38
rs1040246739 Chr.1: 200971385-200971385 on Build GRCh38
rs930632237 Chr.1: 200971302-200971302 on Build GRCh38
rs890006826 Chr.1: 200971394-200971394 on Build GRCh38
rs1058000 Chr.1: 200971294-200971294 on Build GRCh38
rs867927310 Chr.1: 200971647-200971647 on Build GRCh38
rs761643987 Chr.1: 200971503-200971503 on Build GRCh38
rs149081045 Chr.1: 200971439-200971439 on Build GRCh38
rs1040298553 Chr.1: 200971566-200971566 on Build GRCh38
rs192056886 Chr.1: 200971502-200971502 on Build GRCh38
rs902837549 Chr.1: 200971694-200971694 on Build GRCh38
rs888492192 Chr.1: 200971615-200971615 on Build GRCh38
rs538843556 Chr.1: 200971555-200971555 on Build GRCh38
rs1035300596 Chr.1: 200971472-200971472 on Build GRCh38
rs535208336 Chr.1: 200971484-200971484 on Build GRCh38
rs1050429370 Chr.1: 200971383-200971383 on Build GRCh38

View Full Product Details