Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206869150-206869651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL20
Entrez Gene ID
50604
Gene Name
interleukin 20
Gene Aliases
IL-20, IL10D, ZCYTO10
UniGene
Hs.272373
Gene Chromosome Location
Chr.1: 206865247-206869223 on Build GRCh38
Transcripts
NM_018724.3 XM_005273152.3

SNP Detail

SNP ID Chromosome Location
rs569475825 Chr.1: 206869469-206869469 on Build GRCh38
rs1026971487 Chr.1: 206869313-206869313 on Build GRCh38
rs892842251 Chr.1: 206869180-206869180 on Build GRCh38
rs958877995 Chr.1: 206869277-206869277 on Build GRCh38
rs372321490 Chr.1: 206869317-206869317 on Build GRCh38
rs3024523 Chr.1: 206869339-206869339 on Build GRCh38
rs888426510 Chr.1: 206869571-206869571 on Build GRCh38
rs527421722 Chr.1: 206869391-206869391 on Build GRCh38
rs960574054 Chr.1: 206869362-206869362 on Build GRCh38
rs1003135304 Chr.1: 206869267-206869267 on Build GRCh38
rs1057365361 Chr.1: 206869487-206869487 on Build GRCh38
rs984856981 Chr.1: 206869622-206869622 on Build GRCh38
rs965837659 Chr.1: 206869621-206869621 on Build GRCh38
rs1022780757 Chr.1: 206869306-206869306 on Build GRCh38
rs1052651456 Chr.1: 206869466-206869466 on Build GRCh38
rs1002291252 Chr.1: 206869558-206869558 on Build GRCh38
rs1004219365 Chr.1: 206869593-206869593 on Build GRCh38
rs984497421 Chr.1: 206869400-206869400 on Build GRCh38
rs892739930 Chr.1: 206869517-206869517 on Build GRCh38
rs3024528 Chr.1: 206869402-206869402 on Build GRCh38
rs960542929 Chr.1: 206869592-206869592 on Build GRCh38
rs952720771 Chr.1: 206869314-206869314 on Build GRCh38
rs567063427 Chr.1: 206869417-206869417 on Build GRCh38
rs918897721 Chr.1: 206869398-206869398 on Build GRCh38
rs965535421 Chr.1: 206869363-206869363 on Build GRCh38
rs571278095 Chr.1: 206869624-206869624 on Build GRCh38
rs562553754 Chr.1: 206869272-206869272 on Build GRCh38
rs952025747 Chr.1: 206869600-206869600 on Build GRCh38
rs892473456 Chr.1: 206869250-206869250 on Build GRCh38
rs3024524 Chr.1: 206869463-206869463 on Build GRCh38
rs775633711 Chr.1: 206869352-206869352 on Build GRCh38
rs899668572 Chr.1: 206869496-206869496 on Build GRCh38
rs1026917768 Chr.1: 206869554-206869554 on Build GRCh38
rs930324499 Chr.1: 206869399-206869399 on Build GRCh38
rs3024525 Chr.1: 206869553-206869553 on Build GRCh38
rs547281669 Chr.1: 206869403-206869403 on Build GRCh38
rs560036258 Chr.1: 206869181-206869181 on Build GRCh38
rs969525697 Chr.1: 206869324-206869324 on Build GRCh38
rs1012881013 Chr.1: 206869308-206869308 on Build GRCh38
rs971981951 Chr.1: 206869385-206869385 on Build GRCh38
rs1034816242 Chr.1: 206869560-206869560 on Build GRCh38
rs1053988596 Chr.1: 206869418-206869418 on Build GRCh38
rs1039847692 Chr.1: 206869414-206869414 on Build GRCh38
rs114133732 Chr.1: 206869189-206869189 on Build GRCh38
rs763153315 Chr.1: 206869551-206869551 on Build GRCh38
rs879494120 Chr.1: 206869625-206869625 on Build GRCh38
rs1041475904 Chr.1: 206869204-206869204 on Build GRCh38
rs3024522 Chr.1: 206869218-206869218 on Build GRCh38
rs914224628 Chr.1: 206869429-206869429 on Build GRCh38
rs867557126 Chr.1: 206869598-206869598 on Build GRCh38
rs1034888432 Chr.1: 206869349-206869349 on Build GRCh38
rs373746705 Chr.1: 206869176-206869176 on Build GRCh38
rs148564546 Chr.1: 206869599-206869599 on Build GRCh38
rs902932193 Chr.1: 206869465-206869465 on Build GRCh38
rs536000694 Chr.1: 206869462-206869462 on Build GRCh38
rs773096955 Chr.1: 206869337-206869337 on Build GRCh38
rs564856820 Chr.1: 206869397-206869397 on Build GRCh38

View Full Product Details