Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220102399-220102948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IARS2
Entrez Gene ID
55699
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
Gene Aliases
CAGSSS, ILERS
UniGene
Hs.262823
Gene Chromosome Location
Chr.1: 220094113-220148041 on Build GRCh38
Transcripts
NM_018060.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4927929 c.900A>G p.Q300Q Chr.1: 220102727-220102727 on Build GRCh38
1959100 c.749+2T>C p.? Chr.1: 220102414-220102414 on Build GRCh38
1338968 c.808T>C p.Y270H Chr.1: 220102553-220102553 on Build GRCh38
1210208 c.785C>A p.P262H Chr.1: 220102530-220102530 on Build GRCh38
1959102 c.887T>C p.V296A Chr.1: 220102714-220102714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201916678 Chr.1: 220102540-220102540 on Build GRCh38
rs777635643 Chr.1: 220102690-220102690 on Build GRCh38
rs746455426 Chr.1: 220102533-220102533 on Build GRCh38
rs377432411 Chr.1: 220102545-220102545 on Build GRCh38
rs757171258 Chr.1: 220102809-220102809 on Build GRCh38
rs763844430 Chr.1: 220102790-220102790 on Build GRCh38
rs984688280 Chr.1: 220102694-220102694 on Build GRCh38
rs924595832 Chr.1: 220102641-220102641 on Build GRCh38
rs774960759 Chr.1: 220102463-220102463 on Build GRCh38
rs751547113 Chr.1: 220102804-220102804 on Build GRCh38
rs762202072 Chr.1: 220102465-220102465 on Build GRCh38
rs769112693 Chr.1: 220102553-220102553 on Build GRCh38
rs755004683 Chr.1: 220102762-220102762 on Build GRCh38
rs773819050 Chr.1: 220102748-220102748 on Build GRCh38
rs148564492 Chr.1: 220102769-220102769 on Build GRCh38
rs780543464 Chr.1: 220102716-220102716 on Build GRCh38
rs917298497 Chr.1: 220102544-220102544 on Build GRCh38
rs751095095 Chr.1: 220102512-220102512 on Build GRCh38
rs372618006 Chr.1: 220102429-220102429 on Build GRCh38
rs771509480 Chr.1: 220102686-220102688 on Build GRCh38
rs112842443 Chr.1: 220102727-220102727 on Build GRCh38
rs904984929 Chr.1: 220102455-220102455 on Build GRCh38
rs752366568 Chr.1: 220102669-220102669 on Build GRCh38
rs752555234 Chr.1: 220102782-220102782 on Build GRCh38
rs756801026 Chr.1: 220102708-220102708 on Build GRCh38
rs776586188 Chr.1: 220102820-220102820 on Build GRCh38
rs1032292776 Chr.1: 220102518-220102518 on Build GRCh38
rs201539020 Chr.1: 220102436-220102436 on Build GRCh38
rs777266427 Chr.1: 220102531-220102531 on Build GRCh38
rs766741951 Chr.1: 220102626-220102626 on Build GRCh38
rs754766431 Chr.1: 220102498-220102498 on Build GRCh38
rs769328073 Chr.1: 220102730-220102730 on Build GRCh38
rs539127735 Chr.1: 220102629-220102629 on Build GRCh38
rs1046217829 Chr.1: 220102918-220102918 on Build GRCh38
rs764539319 Chr.1: 220102449-220102449 on Build GRCh38
rs767766570 Chr.1: 220102467-220102467 on Build GRCh38
rs1017161423 Chr.1: 220102910-220102910 on Build GRCh38
rs190919285 Chr.1: 220102869-220102869 on Build GRCh38
rs758325882 Chr.1: 220102529-220102529 on Build GRCh38
rs373433981 Chr.1: 220102520-220102520 on Build GRCh38
rs887437954 Chr.1: 220102861-220102861 on Build GRCh38
rs976770107 Chr.1: 220102591-220102591 on Build GRCh38
rs770588599 Chr.1: 220102534-220102534 on Build GRCh38
rs775841640 Chr.1: 220102431-220102431 on Build GRCh38
rs773439017 Chr.1: 220102594-220102594 on Build GRCh38
rs762693000 Chr.1: 220102786-220102786 on Build GRCh38
rs772728778 Chr.1: 220102743-220102743 on Build GRCh38
rs370591047 Chr.1: 220102721-220102721 on Build GRCh38
rs780819893 Chr.1: 220102810-220102810 on Build GRCh38
rs958850547 Chr.1: 220102443-220102443 on Build GRCh38
rs910165696 Chr.1: 220102695-220102695 on Build GRCh38
rs373895742 Chr.1: 220102800-220102800 on Build GRCh38
rs949125150 Chr.1: 220102907-220102907 on Build GRCh38
rs766651661 Chr.1: 220102493-220102493 on Build GRCh38
rs759657956 Chr.1: 220102637-220102637 on Build GRCh38
rs762450476 Chr.1: 220102592-220102592 on Build GRCh38
rs756410015 Chr.1: 220102490-220102490 on Build GRCh38
rs774564592 Chr.1: 220102566-220102566 on Build GRCh38
rs370306595 Chr.1: 220102452-220102452 on Build GRCh38
rs1047413901 Chr.1: 220102856-220102856 on Build GRCh38
rs760791870 Chr.1: 220102602-220102602 on Build GRCh38
rs761237590 Chr.1: 220102758-220102758 on Build GRCh38
rs776306373 Chr.1: 220102537-220102537 on Build GRCh38
rs778544749 Chr.1: 220102507-220102507 on Build GRCh38
rs772640835 Chr.1: 220102593-220102593 on Build GRCh38
rs758342230 Chr.1: 220102682-220102682 on Build GRCh38
rs750528193 Chr.1: 220102478-220102478 on Build GRCh38
rs779341602 Chr.1: 220102735-220102735 on Build GRCh38
rs751099655 Chr.1: 220102691-220102691 on Build GRCh38
rs1028913458 Chr.1: 220102609-220102609 on Build GRCh38
rs1009545961 Chr.1: 220102536-220102536 on Build GRCh38
rs374904870 Chr.1: 220102430-220102430 on Build GRCh38
rs370222815 Chr.1: 220102438-220102438 on Build GRCh38
rs533806944 Chr.1: 220102657-220102657 on Build GRCh38
rs753601921 Chr.1: 220102497-220102497 on Build GRCh38
rs1040103665 Chr.1: 220102819-220102819 on Build GRCh38
rs745590557 Chr.1: 220102723-220102723 on Build GRCh38
rs993639355 Chr.1: 220102850-220102850 on Build GRCh38
rs748465920 Chr.1: 220102739-220102739 on Build GRCh38

View Full Product Details