Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220105539-220106034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IARS2
Entrez Gene ID
55699
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
Gene Aliases
CAGSSS, ILERS
UniGene
Hs.262823
Gene Chromosome Location
Chr.1: 220094113-220148041 on Build GRCh38
Transcripts
NM_018060.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4028378 c.1091G>A p.C364Y Chr.1: 220105915-220105915 on Build GRCh38
334253 c.1153G>T p.A385S Chr.1: 220105977-220105977 on Build GRCh38
4028379 c.1172T>C p.V391A Chr.1: 220105996-220105996 on Build GRCh38
6485440 c.1100C>A p.P367Q Chr.1: 220105924-220105924 on Build GRCh38
4028380 c.1207G>A p.G403S Chr.1: 220106031-220106031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757599257 Chr.1: 220105853-220105853 on Build GRCh38
rs267598383 Chr.1: 220105932-220105932 on Build GRCh38
rs764931000 Chr.1: 220105840-220105840 on Build GRCh38
rs756637382 Chr.1: 220105868-220105868 on Build GRCh38
rs529898953 Chr.1: 220105698-220105698 on Build GRCh38
rs762652814 Chr.1: 220105947-220105947 on Build GRCh38
rs773051028 Chr.1: 220105918-220105918 on Build GRCh38
rs376352707 Chr.1: 220105699-220105699 on Build GRCh38
rs1009546323 Chr.1: 220105676-220105676 on Build GRCh38
rs6681516 Chr.1: 220105911-220105911 on Build GRCh38
rs781179718 Chr.1: 220105657-220105657 on Build GRCh38
rs749309935 Chr.1: 220105893-220105893 on Build GRCh38
rs563322955 Chr.1: 220105851-220105851 on Build GRCh38
rs745973991 Chr.1: 220105914-220105915 on Build GRCh38
rs1046940346 Chr.1: 220105817-220105817 on Build GRCh38
rs373657129 Chr.1: 220105917-220105917 on Build GRCh38
rs760343115 Chr.1: 220105752-220105752 on Build GRCh38
rs148305375 Chr.1: 220105628-220105628 on Build GRCh38
rs980824585 Chr.1: 220105732-220105732 on Build GRCh38
rs565659901 Chr.1: 220105960-220105960 on Build GRCh38
rs920652142 Chr.1: 220105708-220105708 on Build GRCh38
rs753952424 Chr.1: 220105971-220105971 on Build GRCh38
rs1003118784 Chr.1: 220105849-220105849 on Build GRCh38
rs748372193 Chr.1: 220105912-220105912 on Build GRCh38
rs752338172 Chr.1: 220105945-220105945 on Build GRCh38
rs367794415 Chr.1: 220105931-220105931 on Build GRCh38
rs528155025 Chr.1: 220105974-220105974 on Build GRCh38
rs996501724 Chr.1: 220105642-220105642 on Build GRCh38
rs533095444 Chr.1: 220105631-220105631 on Build GRCh38
rs199869294 Chr.1: 220105948-220105948 on Build GRCh38
rs781597874 Chr.1: 220105858-220105858 on Build GRCh38
rs780110239 Chr.1: 220105869-220105869 on Build GRCh38
rs770929208 Chr.1: 220105927-220105927 on Build GRCh38
rs182541053 Chr.1: 220105975-220105975 on Build GRCh38
rs927936022 Chr.1: 220105770-220105770 on Build GRCh38
rs79922039 Chr.1: 220105979-220105979 on Build GRCh38
rs779067575 Chr.1: 220105907-220105907 on Build GRCh38
rs768889727 Chr.1: 220105902-220105902 on Build GRCh38
rs373578830 Chr.1: 220105987-220105987 on Build GRCh38
rs898267420 Chr.1: 220105792-220105792 on Build GRCh38
rs756590905 Chr.1: 220105963-220105963 on Build GRCh38
rs1039758154 Chr.1: 220105753-220105753 on Build GRCh38
rs1029675021 Chr.1: 220105645-220105645 on Build GRCh38
rs748379506 Chr.1: 220105984-220105984 on Build GRCh38
rs371548673 Chr.1: 220105966-220105966 on Build GRCh38
rs760599825 Chr.1: 220105923-220105923 on Build GRCh38
rs538008787 Chr.1: 220105655-220105655 on Build GRCh38
rs933575623 Chr.1: 220105582-220105582 on Build GRCh38
rs931022499 Chr.1: 220105804-220105804 on Build GRCh38
rs750949355 Chr.1: 220105860-220105860 on Build GRCh38
rs764804286 Chr.1: 220105942-220105942 on Build GRCh38
rs143213917 Chr.1: 220105596-220105596 on Build GRCh38
rs143373097 Chr.1: 220105988-220105988 on Build GRCh38
rs149196564 Chr.1: 220105901-220105901 on Build GRCh38
rs969388493 Chr.1: 220105725-220105725 on Build GRCh38
rs886902286 Chr.1: 220105824-220105824 on Build GRCh38
rs973485174 Chr.1: 220105699-220105699 on Build GRCh38
rs746765893 Chr.1: 220106002-220106002 on Build GRCh38
rs141451740 Chr.1: 220105684-220105684 on Build GRCh38
rs939321424 Chr.1: 220105816-220105816 on Build GRCh38

View Full Product Details