Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 245156334-245156828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF26B
Entrez Gene ID
55083
Gene Name
kinesin family member 26B
Gene Aliases
-
UniGene
Hs.143134
Gene Chromosome Location
Chr.1: 245154985-245703126 on Build GRCh38
Transcripts
NM_018012.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5448492 c.343A>G p.S115G Chr.1: 245156561-245156561 on Build GRCh38
6306061 c.367T>A p.S123T Chr.1: 245156585-245156585 on Build GRCh38
4764952 c.118T>G p.W40G Chr.1: 245156336-245156336 on Build GRCh38
6386217 c.155G>A p.S52N Chr.1: 245156373-245156373 on Build GRCh38
5088690 c.185C>T p.A62V Chr.1: 245156403-245156403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765071022 Chr.1: 245156440-245156440 on Build GRCh38
rs780380375 Chr.1: 245156563-245156563 on Build GRCh38
rs554058547 Chr.1: 245156413-245156413 on Build GRCh38
rs757794492 Chr.1: 245156558-245156558 on Build GRCh38
rs772909497 Chr.1: 245156582-245156582 on Build GRCh38
rs545986853 Chr.1: 245156570-245156570 on Build GRCh38
rs898399865 Chr.1: 245156449-245156449 on Build GRCh38
rs1029051686 Chr.1: 245156714-245156715 on Build GRCh38
rs886250607 Chr.1: 245156650-245156650 on Build GRCh38
rs997789586 Chr.1: 245156522-245156542 on Build GRCh38
rs987657478 Chr.1: 245156711-245156711 on Build GRCh38
rs957590336 Chr.1: 245156547-245156547 on Build GRCh38
rs774527027 Chr.1: 245156367-245156367 on Build GRCh38
rs544676853 Chr.1: 245156673-245156673 on Build GRCh38
rs940029296 Chr.1: 245156569-245156569 on Build GRCh38
rs907710783 Chr.1: 245156755-245156755 on Build GRCh38
rs562693918 Chr.1: 245156587-245156587 on Build GRCh38
rs759187749 Chr.1: 245156617-245156617 on Build GRCh38
rs906602396 Chr.1: 245156789-245156789 on Build GRCh38
rs943281850 Chr.1: 245156365-245156365 on Build GRCh38
rs576183812 Chr.1: 245156665-245156665 on Build GRCh38
rs958233804 Chr.1: 245156482-245156482 on Build GRCh38
rs1044232943 Chr.1: 245156721-245156721 on Build GRCh38
rs994627091 Chr.1: 245156395-245156395 on Build GRCh38
rs765943712 Chr.1: 245156491-245156491 on Build GRCh38
rs747869981 Chr.1: 245156566-245156566 on Build GRCh38
rs767207430 Chr.1: 245156548-245156548 on Build GRCh38
rs1021134496 Chr.1: 245156695-245156695 on Build GRCh38
rs751139875 Chr.1: 245156524-245156524 on Build GRCh38
rs530105063 Chr.1: 245156738-245156738 on Build GRCh38
rs773245496 Chr.1: 245156463-245156463 on Build GRCh38
rs1055480026 Chr.1: 245156794-245156795 on Build GRCh38
rs772276937 Chr.1: 245156560-245156560 on Build GRCh38
rs764301839 Chr.1: 245156535-245156536 on Build GRCh38
rs938041541 Chr.1: 245156797-245156797 on Build GRCh38
rs1002209932 Chr.1: 245156662-245156662 on Build GRCh38
rs940104132 Chr.1: 245156651-245156651 on Build GRCh38
rs972728971 Chr.1: 245156769-245156769 on Build GRCh38
rs919961859 Chr.1: 245156782-245156782 on Build GRCh38
rs940050719 Chr.1: 245156763-245156763 on Build GRCh38
rs1046957353 Chr.1: 245156779-245156779 on Build GRCh38
rs745950507 Chr.1: 245156559-245156559 on Build GRCh38
rs574362934 Chr.1: 245156377-245156377 on Build GRCh38
rs895726574 Chr.1: 245156468-245156468 on Build GRCh38
rs924884560 Chr.1: 245156539-245156539 on Build GRCh38
rs911536952 Chr.1: 245156354-245156354 on Build GRCh38
rs1025415000 Chr.1: 245156436-245156436 on Build GRCh38
rs761735254 Chr.1: 245156424-245156424 on Build GRCh38
rs949907329 Chr.1: 245156764-245156764 on Build GRCh38
rs554398118 Chr.1: 245156359-245156359 on Build GRCh38
rs1004270586 Chr.1: 245156431-245156431 on Build GRCh38
rs1057402055 Chr.1: 245156441-245156441 on Build GRCh38
rs999161612 Chr.1: 245156677-245156677 on Build GRCh38
rs756559815 Chr.1: 245156553-245156553 on Build GRCh38
rs533597039 Chr.1: 245156389-245156389 on Build GRCh38
rs1018692501 Chr.1: 245156715-245156715 on Build GRCh38
rs752444847 Chr.1: 245156653-245156653 on Build GRCh38
rs906561974 Chr.1: 245156660-245156660 on Build GRCh38
rs546503653 Chr.1: 245156646-245156646 on Build GRCh38
rs573451068 Chr.1: 245156699-245156702 on Build GRCh38
rs760640845 Chr.1: 245156528-245156548 on Build GRCh38
rs760917763 Chr.1: 245156663-245156663 on Build GRCh38
rs762519197 Chr.1: 245156489-245156489 on Build GRCh38
rs868036684 Chr.1: 245156543-245156543 on Build GRCh38
rs773933709 Chr.1: 245156615-245156615 on Build GRCh38
rs894051398 Chr.1: 245156675-245156675 on Build GRCh38
rs1011135362 Chr.1: 245156692-245156692 on Build GRCh38
rs920232018 Chr.1: 245156735-245156735 on Build GRCh38
rs966867509 Chr.1: 245156700-245156700 on Build GRCh38
rs771277250 Chr.1: 245156780-245156780 on Build GRCh38
rs983293637 Chr.1: 245156746-245156746 on Build GRCh38
rs753002645 Chr.1: 245156550-245156550 on Build GRCh38
rs906496262 Chr.1: 245156452-245156452 on Build GRCh38
rs754224933 Chr.1: 245156697-245156697 on Build GRCh38
rs184009471 Chr.1: 245156732-245156732 on Build GRCh38
rs577217671 Chr.1: 245156557-245156557 on Build GRCh38
rs865863102 Chr.1: 245156682-245156682 on Build GRCh38
rs930505849 Chr.1: 245156637-245156637 on Build GRCh38
rs888558652 Chr.1: 245156740-245156740 on Build GRCh38
rs1031933830 Chr.1: 245156678-245156678 on Build GRCh38
rs902247848 Chr.1: 245156667-245156667 on Build GRCh38
rs1020746679 Chr.1: 245156501-245156501 on Build GRCh38
rs1002877094 Chr.1: 245156464-245156464 on Build GRCh38
rs948573436 Chr.1: 245156476-245156476 on Build GRCh38
rs891173512 Chr.1: 245156416-245156416 on Build GRCh38
rs771009085 Chr.1: 245156596-245156596 on Build GRCh38
rs1033923891 Chr.1: 245156478-245156478 on Build GRCh38
rs964756002 Chr.1: 245156720-245156720 on Build GRCh38
rs759209166 Chr.1: 245156530-245156530 on Build GRCh38

View Full Product Details